Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 77039851-77040351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GNT6
Entrez Gene ID
192134
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6
Gene Aliases
B3Gn-T6, BGnT-6, beta-1,3-Gn-T6, beta3Gn-T6
UniGene
Hs.352622
Gene Chromosome Location
Chr.11: 77034341-77041973 on Build GRCh38
Transcripts
NM_138706.4

SNP Detail

SNP ID Chromosome Location
rs376893569 Chr.11: 77040013-77040013 on Build GRCh38
rs782037273 Chr.11: 77039970-77039970 on Build GRCh38
rs782062270 Chr.11: 77039970-77039970 on Build GRCh38
rs782536315 Chr.11: 77040030-77040030 on Build GRCh38
rs781809967 Chr.11: 77040023-77040023 on Build GRCh38
rs918748488 Chr.11: 77040096-77040096 on Build GRCh38
rs781800403 Chr.11: 77040092-77040092 on Build GRCh38
rs782445779 Chr.11: 77040094-77040094 on Build GRCh38
rs782422372 Chr.11: 77040265-77040265 on Build GRCh38
rs782075948 Chr.11: 77040172-77040172 on Build GRCh38
rs782520683 Chr.11: 77040104-77040104 on Build GRCh38
rs781990020 Chr.11: 77040078-77040078 on Build GRCh38
rs782264659 Chr.11: 77040234-77040234 on Build GRCh38
rs781897283 Chr.11: 77040034-77040034 on Build GRCh38
rs954181079 Chr.11: 77039876-77039876 on Build GRCh38
rs782357846 Chr.11: 77040040-77040040 on Build GRCh38
rs781853436 Chr.11: 77040174-77040174 on Build GRCh38
rs782349177 Chr.11: 77040142-77040142 on Build GRCh38
rs782319003 Chr.11: 77040242-77040242 on Build GRCh38
rs781938785 Chr.11: 77040274-77040274 on Build GRCh38
rs781972937 Chr.11: 77039894-77039894 on Build GRCh38
rs553141671 Chr.11: 77039887-77039887 on Build GRCh38
rs782106072 Chr.11: 77040178-77040179 on Build GRCh38
rs782096762 Chr.11: 77040012-77040012 on Build GRCh38
rs782386317 Chr.11: 77040004-77040004 on Build GRCh38
rs370071740 Chr.11: 77040010-77040010 on Build GRCh38
rs782714525 Chr.11: 77040194-77040194 on Build GRCh38
rs781826107 Chr.11: 77040284-77040284 on Build GRCh38
rs781800969 Chr.11: 77039906-77039906 on Build GRCh38
rs782499048 Chr.11: 77040197-77040197 on Build GRCh38
rs781831695 Chr.11: 77040028-77040028 on Build GRCh38
rs782615232 Chr.11: 77040002-77040002 on Build GRCh38
rs775222478 Chr.11: 77040238-77040238 on Build GRCh38
rs782676328 Chr.11: 77040005-77040005 on Build GRCh38
rs782704720 Chr.11: 77039972-77039972 on Build GRCh38
rs782419471 Chr.11: 77039883-77039883 on Build GRCh38
rs573228992 Chr.11: 77039895-77039895 on Build GRCh38
rs782689137 Chr.11: 77040296-77040296 on Build GRCh38
rs782056729 Chr.11: 77040024-77040024 on Build GRCh38
rs267603199 Chr.11: 77040182-77040182 on Build GRCh38
rs782801801 Chr.11: 77040090-77040090 on Build GRCh38
rs782378349 Chr.11: 77040044-77040044 on Build GRCh38
rs782288659 Chr.11: 77040240-77040240 on Build GRCh38
rs782775018 Chr.11: 77040173-77040173 on Build GRCh38
rs782598594 Chr.11: 77040041-77040041 on Build GRCh38
rs532474065 Chr.11: 77040147-77040147 on Build GRCh38
rs781794721 Chr.11: 77039993-77039993 on Build GRCh38
rs782582150 Chr.11: 77040129-77040129 on Build GRCh38
rs376838217 Chr.11: 77040144-77040144 on Build GRCh38
rs1017033463 Chr.11: 77040055-77040055 on Build GRCh38
rs782015746 Chr.11: 77039968-77039968 on Build GRCh38
rs969606190 Chr.11: 77040325-77040325 on Build GRCh38
rs782053863 Chr.11: 77040276-77040276 on Build GRCh38
rs782166923 Chr.11: 77040087-77040087 on Build GRCh38
rs782536931 Chr.11: 77039990-77039990 on Build GRCh38
rs782719043 Chr.11: 77039892-77039892 on Build GRCh38
rs782692000 Chr.11: 77040239-77040239 on Build GRCh38
rs782326955 Chr.11: 77039925-77039925 on Build GRCh38
rs782589139 Chr.11: 77040292-77040292 on Build GRCh38
rs781908212 Chr.11: 77039989-77039989 on Build GRCh38
rs782597921 Chr.11: 77040198-77040198 on Build GRCh38
rs774249444 Chr.11: 77040008-77040008 on Build GRCh38
rs782111857 Chr.11: 77039948-77039948 on Build GRCh38
rs760180919 Chr.11: 77040290-77040290 on Build GRCh38
rs200446406 Chr.11: 77040195-77040195 on Build GRCh38
rs199952262 Chr.11: 77040003-77040003 on Build GRCh38
rs782590846 Chr.11: 77039992-77039992 on Build GRCh38
rs201605703 Chr.11: 77039907-77039907 on Build GRCh38
rs782196204 Chr.11: 77040059-77040059 on Build GRCh38
rs782808940 Chr.11: 77040021-77040021 on Build GRCh38
rs999325119 Chr.11: 77039995-77039995 on Build GRCh38
rs782719005 Chr.11: 77040218-77040218 on Build GRCh38
rs781925100 Chr.11: 77039926-77039926 on Build GRCh38
rs782780998 Chr.11: 77040046-77040047 on Build GRCh38
rs782131925 Chr.11: 77040286-77040286 on Build GRCh38
rs782416119 Chr.11: 77039959-77039959 on Build GRCh38
rs374076293 Chr.11: 77040278-77040278 on Build GRCh38
rs782247197 Chr.11: 77039874-77039874 on Build GRCh38
rs376792782 Chr.11: 77039932-77039932 on Build GRCh38
rs782008657 Chr.11: 77039946-77039946 on Build GRCh38
rs782810391 Chr.11: 77040211-77040211 on Build GRCh38
rs557075861 Chr.11: 77040251-77040251 on Build GRCh38
rs112854071 Chr.11: 77040101-77040101 on Build GRCh38
rs781965154 Chr.11: 77040166-77040166 on Build GRCh38
rs782220216 Chr.11: 77040249-77040249 on Build GRCh38
rs190020175 Chr.11: 77040157-77040157 on Build GRCh38
rs901662991 Chr.11: 77040216-77040216 on Build GRCh38
rs935081233 Chr.11: 77040176-77040176 on Build GRCh38
rs782281194 Chr.11: 77040299-77040299 on Build GRCh38
rs782001104 Chr.11: 77040017-77040017 on Build GRCh38
rs1043333301 Chr.11: 77040164-77040164 on Build GRCh38
rs782625605 Chr.11: 77039908-77039908 on Build GRCh38
rs781826774 Chr.11: 77040114-77040114 on Build GRCh38
rs782212407 Chr.11: 77039909-77039909 on Build GRCh38
rs782228197 Chr.11: 77040136-77040136 on Build GRCh38
rs782277000 Chr.11: 77040006-77040006 on Build GRCh38
rs781821239 Chr.11: 77040103-77040103 on Build GRCh38
rs994640654 Chr.11: 77040273-77040273 on Build GRCh38
rs749101048 Chr.11: 77039962-77039962 on Build GRCh38
rs782021841 Chr.11: 77039886-77039886 on Build GRCh38
rs782282887 Chr.11: 77039917-77039917 on Build GRCh38
rs781973825 Chr.11: 77040324-77040324 on Build GRCh38
rs782313252 Chr.11: 77040061-77040061 on Build GRCh38
rs782312563 Chr.11: 77040314-77040314 on Build GRCh38
rs782007789 Chr.11: 77040143-77040143 on Build GRCh38
rs916807620 Chr.11: 77040152-77040152 on Build GRCh38
rs782634131 Chr.11: 77040031-77040031 on Build GRCh38
rs782486449 Chr.11: 77039996-77039996 on Build GRCh38
rs782572228 Chr.11: 77040248-77040248 on Build GRCh38
rs371112742 Chr.11: 77040019-77040019 on Build GRCh38
rs782781650 Chr.11: 77040288-77040288 on Build GRCh38
rs782505943 Chr.11: 77039911-77039911 on Build GRCh38
rs781834387 Chr.11: 77039973-77039973 on Build GRCh38
rs782683634 Chr.11: 77039916-77039916 on Build GRCh38
rs546848248 Chr.11: 77040046-77040046 on Build GRCh38
rs368100863 Chr.11: 77040291-77040291 on Build GRCh38
rs782785985 Chr.11: 77040069-77040069 on Build GRCh38
rs73493606 Chr.11: 77040122-77040122 on Build GRCh38
rs567787570 Chr.11: 77040256-77040256 on Build GRCh38
rs782344338 Chr.11: 77039941-77039941 on Build GRCh38
rs782549400 Chr.11: 77039900-77039900 on Build GRCh38
rs782448496 Chr.11: 77040124-77040124 on Build GRCh38
rs782633729 Chr.11: 77040119-77040119 on Build GRCh38
rs962826417 Chr.11: 77040269-77040269 on Build GRCh38
rs782522041 Chr.11: 77040037-77040037 on Build GRCh38
rs529741392 Chr.11: 77040039-77040039 on Build GRCh38
rs781996703 Chr.11: 77040252-77040252 on Build GRCh38
rs781812737 Chr.11: 77040299-77040299 on Build GRCh38
rs782780623 Chr.11: 77039902-77039902 on Build GRCh38
rs782553406 Chr.11: 77040175-77040175 on Build GRCh38
rs782742159 Chr.11: 77040100-77040100 on Build GRCh38
rs782751519 Chr.11: 77040025-77040025 on Build GRCh38

View Full Product Details