Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 77040230-77040717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GNT6
Entrez Gene ID
192134
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6
Gene Aliases
B3Gn-T6, BGnT-6, beta-1,3-Gn-T6, beta3Gn-T6
UniGene
Hs.352622
Gene Chromosome Location
Chr.11: 77034341-77041973 on Build GRCh38
Transcripts
NM_138706.4

SNP Detail

SNP ID Chromosome Location
rs782251537 Chr.11: 77040542-77040542 on Build GRCh38
rs782486897 Chr.11: 77040551-77040551 on Build GRCh38
rs184932010 Chr.11: 77040602-77040602 on Build GRCh38
rs898485626 Chr.11: 77040501-77040501 on Build GRCh38
rs373753540 Chr.11: 77040657-77040657 on Build GRCh38
rs200788398 Chr.11: 77040557-77040557 on Build GRCh38
rs34153015 Chr.11: 77040541-77040541 on Build GRCh38
rs782422372 Chr.11: 77040265-77040265 on Build GRCh38
rs782758540 Chr.11: 77040549-77040549 on Build GRCh38
rs781938785 Chr.11: 77040274-77040274 on Build GRCh38
rs572701335 Chr.11: 77040400-77040400 on Build GRCh38
rs782792955 Chr.11: 77040371-77040371 on Build GRCh38
rs77209527 Chr.11: 77040498-77040498 on Build GRCh38
rs782544598 Chr.11: 77040419-77040419 on Build GRCh38
rs781899944 Chr.11: 77040418-77040418 on Build GRCh38
rs781826107 Chr.11: 77040284-77040284 on Build GRCh38
rs782788518 Chr.11: 77040529-77040529 on Build GRCh38
rs569180595 Chr.11: 77040626-77040626 on Build GRCh38
rs544232471 Chr.11: 77040512-77040512 on Build GRCh38
rs782090275 Chr.11: 77040504-77040504 on Build GRCh38
rs782430090 Chr.11: 77040630-77040630 on Build GRCh38
rs782559900 Chr.11: 77040421-77040421 on Build GRCh38
rs782163214 Chr.11: 77040520-77040520 on Build GRCh38
rs782741188 Chr.11: 77040394-77040394 on Build GRCh38
rs782689137 Chr.11: 77040296-77040296 on Build GRCh38
rs573066585 Chr.11: 77040359-77040359 on Build GRCh38
rs368050760 Chr.11: 77040472-77040472 on Build GRCh38
rs782064366 Chr.11: 77040546-77040546 on Build GRCh38
rs781820303 Chr.11: 77040396-77040396 on Build GRCh38
rs559157215 Chr.11: 77040453-77040453 on Build GRCh38
rs969606190 Chr.11: 77040325-77040325 on Build GRCh38
rs182310862 Chr.11: 77040508-77040508 on Build GRCh38
rs782540707 Chr.11: 77040617-77040617 on Build GRCh38
rs781987873 Chr.11: 77040509-77040509 on Build GRCh38
rs782053863 Chr.11: 77040276-77040276 on Build GRCh38
rs782784317 Chr.11: 77040506-77040506 on Build GRCh38
rs796140348 Chr.11: 77040558-77040558 on Build GRCh38
rs782531366 Chr.11: 77040550-77040550 on Build GRCh38
rs782589139 Chr.11: 77040292-77040292 on Build GRCh38
rs782086741 Chr.11: 77040346-77040346 on Build GRCh38
rs782617267 Chr.11: 77040552-77040552 on Build GRCh38
rs782400171 Chr.11: 77040660-77040660 on Build GRCh38
rs782459907 Chr.11: 77040535-77040535 on Build GRCh38
rs144466809 Chr.11: 77040616-77040616 on Build GRCh38
rs782647418 Chr.11: 77040469-77040469 on Build GRCh38
rs782747442 Chr.11: 77040691-77040691 on Build GRCh38
rs760180919 Chr.11: 77040290-77040290 on Build GRCh38
rs796338514 Chr.11: 77040498-77040498 on Build GRCh38
rs539994853 Chr.11: 77040517-77040517 on Build GRCh38
rs781868444 Chr.11: 77040537-77040537 on Build GRCh38
rs933153099 Chr.11: 77040437-77040437 on Build GRCh38
rs782184727 Chr.11: 77040490-77040490 on Build GRCh38
rs782687719 Chr.11: 77040554-77040554 on Build GRCh38
rs782760943 Chr.11: 77040547-77040547 on Build GRCh38
rs560406872 Chr.11: 77040560-77040560 on Build GRCh38
rs782131925 Chr.11: 77040286-77040286 on Build GRCh38
rs782411068 Chr.11: 77040476-77040476 on Build GRCh38
rs782462783 Chr.11: 77040538-77040538 on Build GRCh38
rs374076293 Chr.11: 77040278-77040278 on Build GRCh38
rs782164686 Chr.11: 77040679-77040679 on Build GRCh38
rs202215709 Chr.11: 77040497-77040497 on Build GRCh38
rs782588970 Chr.11: 77040619-77040619 on Build GRCh38
rs532243431 Chr.11: 77040671-77040671 on Build GRCh38
rs1052626929 Chr.11: 77040415-77040415 on Build GRCh38
rs782639298 Chr.11: 77040416-77040416 on Build GRCh38
rs557075861 Chr.11: 77040251-77040251 on Build GRCh38
rs782220216 Chr.11: 77040249-77040249 on Build GRCh38
rs782178186 Chr.11: 77040540-77040540 on Build GRCh38
rs782469643 Chr.11: 77040621-77040621 on Build GRCh38
rs564286495 Chr.11: 77040586-77040586 on Build GRCh38
rs1037634832 Chr.11: 77040645-77040645 on Build GRCh38
rs782810735 Chr.11: 77040681-77040681 on Build GRCh38
rs575842932 Chr.11: 77040463-77040463 on Build GRCh38
rs782281194 Chr.11: 77040299-77040299 on Build GRCh38
rs782156666 Chr.11: 77040565-77040565 on Build GRCh38
rs782167194 Chr.11: 77040369-77040369 on Build GRCh38
rs986406752 Chr.11: 77040623-77040623 on Build GRCh38
rs781931238 Chr.11: 77040574-77040574 on Build GRCh38
rs782568909 Chr.11: 77040556-77040556 on Build GRCh38
rs782423354 Chr.11: 77040543-77040543 on Build GRCh38
rs979108404 Chr.11: 77040344-77040344 on Build GRCh38
rs782678259 Chr.11: 77040534-77040534 on Build GRCh38
rs782344123 Chr.11: 77040537-77040537 on Build GRCh38
rs889070560 Chr.11: 77040456-77040456 on Build GRCh38
rs782594923 Chr.11: 77040539-77040540 on Build GRCh38
rs545303916 Chr.11: 77040382-77040382 on Build GRCh38
rs782092571 Chr.11: 77040659-77040659 on Build GRCh38
rs994640654 Chr.11: 77040273-77040273 on Build GRCh38
rs782135224 Chr.11: 77040615-77040615 on Build GRCh38
rs782020010 Chr.11: 77040544-77040544 on Build GRCh38
rs781973825 Chr.11: 77040324-77040324 on Build GRCh38
rs935113896 Chr.11: 77040410-77040410 on Build GRCh38
rs201940118 Chr.11: 77040555-77040555 on Build GRCh38
rs550987490 Chr.11: 77040686-77040686 on Build GRCh38
rs782312563 Chr.11: 77040314-77040314 on Build GRCh38
rs782478587 Chr.11: 77040532-77040532 on Build GRCh38
rs782227194 Chr.11: 77040656-77040656 on Build GRCh38
rs781988853 Chr.11: 77040357-77040357 on Build GRCh38
rs782045806 Chr.11: 77040577-77040577 on Build GRCh38
rs782781650 Chr.11: 77040288-77040288 on Build GRCh38
rs781814394 Chr.11: 77040375-77040375 on Build GRCh38
rs782011076 Chr.11: 77040485-77040485 on Build GRCh38
rs368100863 Chr.11: 77040291-77040291 on Build GRCh38
rs782502339 Chr.11: 77040533-77040533 on Build GRCh38
rs782127311 Chr.11: 77040548-77040548 on Build GRCh38
rs567787570 Chr.11: 77040256-77040256 on Build GRCh38
rs782234558 Chr.11: 77040536-77040536 on Build GRCh38
rs553172183 Chr.11: 77040352-77040352 on Build GRCh38
rs781984195 Chr.11: 77040561-77040561 on Build GRCh38
rs782307701 Chr.11: 77040652-77040652 on Build GRCh38
rs781804770 Chr.11: 77040530-77040530 on Build GRCh38
rs962826417 Chr.11: 77040269-77040269 on Build GRCh38
rs782657432 Chr.11: 77040625-77040625 on Build GRCh38
rs781875528 Chr.11: 77040553-77040553 on Build GRCh38
rs782316745 Chr.11: 77040494-77040494 on Build GRCh38
rs781996703 Chr.11: 77040252-77040252 on Build GRCh38
rs781812737 Chr.11: 77040299-77040299 on Build GRCh38

View Full Product Details