Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 77821071-77821577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAMDC
Entrez Gene ID
28971
Gene Name
adipogenesis associated Mth938 domain containing
Gene Aliases
C11orf67, CK067, PTD015
UniGene
Hs.745040
Gene Chromosome Location
Chr.11: 77821114-77901281 on Build GRCh38
Transcripts
NM_001316957.1 NM_001316958.1 NM_001316960.1 NM_001316961.1 NM_001316962.1 NM_024684.3 NR_133637.1 XM_011544969.2 XM_017017923.1

Gene Symbol
RSF1
Entrez Gene ID
51773
Gene Name
remodeling and spacing factor 1
Gene Aliases
HBXAP, RSF-1, XAP8, p325
UniGene
Hs.420229
Gene Chromosome Location
Chr.11: 77660009-77872232 on Build GRCh38
Transcripts
NM_001316957.1 NM_001316958.1 NM_001316960.1 NM_001316961.1 NM_001316962.1 NM_024684.3 NR_133637.1 XM_011544969.2 XM_017017923.1

SNP Detail

SNP ID Chromosome Location
rs557539481 Chr.11: 77821345-77821345 on Build GRCh38
rs533066981 Chr.11: 77821104-77821104 on Build GRCh38
rs117804781 Chr.11: 77821121-77821121 on Build GRCh38
rs920686752 Chr.11: 77821516-77821516 on Build GRCh38
rs761041009 Chr.11: 77821136-77821136 on Build GRCh38
rs779331580 Chr.11: 77821461-77821461 on Build GRCh38
rs943416777 Chr.11: 77821115-77821115 on Build GRCh38
rs988129740 Chr.11: 77821368-77821368 on Build GRCh38
rs562399833 Chr.11: 77821390-77821390 on Build GRCh38
rs988186677 Chr.11: 77821177-77821177 on Build GRCh38
rs67606281 Chr.11: 77821417-77821417 on Build GRCh38
rs78196556 Chr.11: 77821260-77821260 on Build GRCh38
rs887666408 Chr.11: 77821445-77821445 on Build GRCh38
rs1048792210 Chr.11: 77821250-77821250 on Build GRCh38
rs572921715 Chr.11: 77821181-77821182 on Build GRCh38
rs576893069 Chr.11: 77821398-77821398 on Build GRCh38
rs1045455736 Chr.11: 77821346-77821346 on Build GRCh38
rs886098139 Chr.11: 77821490-77821490 on Build GRCh38
rs925279644 Chr.11: 77821139-77821139 on Build GRCh38
rs777681445 Chr.11: 77821395-77821395 on Build GRCh38
rs911885803 Chr.11: 77821270-77821270 on Build GRCh38
rs376469491 Chr.11: 77821119-77821119 on Build GRCh38
rs373364394 Chr.11: 77821410-77821410 on Build GRCh38
rs969800038 Chr.11: 77821171-77821171 on Build GRCh38
rs1018924667 Chr.11: 77821475-77821475 on Build GRCh38
rs756265333 Chr.11: 77821321-77821321 on Build GRCh38
rs1004519818 Chr.11: 77821252-77821252 on Build GRCh38
rs768798269 Chr.11: 77821275-77821275 on Build GRCh38
rs976347352 Chr.11: 77821406-77821406 on Build GRCh38
rs930484797 Chr.11: 77821327-77821327 on Build GRCh38
rs767918419 Chr.11: 77821128-77821128 on Build GRCh38
rs551179867 Chr.11: 77821440-77821440 on Build GRCh38
rs1048132196 Chr.11: 77821140-77821140 on Build GRCh38
rs905373090 Chr.11: 77821550-77821550 on Build GRCh38
rs746232582 Chr.11: 77821504-77821504 on Build GRCh38
rs1011496149 Chr.11: 77821427-77821427 on Build GRCh38
rs1001252996 Chr.11: 77821172-77821172 on Build GRCh38
rs546301848 Chr.11: 77821402-77821402 on Build GRCh38
rs989649208 Chr.11: 77821212-77821212 on Build GRCh38
rs375849077 Chr.11: 77821256-77821256 on Build GRCh38
rs1014372832 Chr.11: 77821155-77821155 on Build GRCh38
rs912656818 Chr.11: 77821375-77821375 on Build GRCh38
rs887437390 Chr.11: 77821251-77821251 on Build GRCh38
rs886345997 Chr.11: 77821153-77821153 on Build GRCh38
rs967399513 Chr.11: 77821459-77821459 on Build GRCh38
rs1024263898 Chr.11: 77821529-77821529 on Build GRCh38
rs1033863354 Chr.11: 77821175-77821175 on Build GRCh38
rs752849167 Chr.11: 77821256-77821256 on Build GRCh38
rs562414757 Chr.11: 77821525-77821525 on Build GRCh38
rs958214217 Chr.11: 77821189-77821189 on Build GRCh38
rs1032776744 Chr.11: 77821413-77821413 on Build GRCh38
rs961170116 Chr.11: 77821118-77821118 on Build GRCh38
rs912613367 Chr.11: 77821178-77821178 on Build GRCh38
rs912015325 Chr.11: 77821106-77821106 on Build GRCh38
rs914171626 Chr.11: 77821248-77821248 on Build GRCh38
rs565537614 Chr.11: 77821329-77821329 on Build GRCh38
rs1039077028 Chr.11: 77821311-77821311 on Build GRCh38
rs987727417 Chr.11: 77821466-77821466 on Build GRCh38
rs1023792107 Chr.11: 77821326-77821326 on Build GRCh38
rs535373455 Chr.11: 77821161-77821161 on Build GRCh38
rs944550716 Chr.11: 77821180-77821180 on Build GRCh38
rs191083087 Chr.11: 77821524-77821524 on Build GRCh38
rs138017952 Chr.11: 77821337-77821337 on Build GRCh38
rs935458038 Chr.11: 77821146-77821146 on Build GRCh38
rs540222090 Chr.11: 77821205-77821205 on Build GRCh38
rs900439945 Chr.11: 77821196-77821196 on Build GRCh38
rs576416348 Chr.11: 77821439-77821439 on Build GRCh38
rs1001179460 Chr.11: 77821411-77821411 on Build GRCh38
rs956712091 Chr.11: 77821364-77821364 on Build GRCh38
rs1032518746 Chr.11: 77821347-77821347 on Build GRCh38

View Full Product Details