Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 86809519-86810024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRSS23
Entrez Gene ID
11098
Gene Name
protease, serine 23
Gene Aliases
SIG13, SPUVE, ZSIG13
UniGene
Hs.25338
Gene Chromosome Location
Chr.11: 86791059-86952910 on Build GRCh38
Transcripts
NM_001293178.1 NM_001293179.1 NM_001293180.1 NM_007173.5 NR_120591.1 NR_120592.1 NR_120593.1

SNP Detail

SNP ID Chromosome Location
rs2155081 Chr.11: 86809710-86809710 on Build GRCh38
rs936556039 Chr.11: 86809596-86809596 on Build GRCh38
rs1003271910 Chr.11: 86809971-86809971 on Build GRCh38
rs75414855 Chr.11: 86809593-86809593 on Build GRCh38
rs995712465 Chr.11: 86809847-86809847 on Build GRCh38
rs556361020 Chr.11: 86809962-86809962 on Build GRCh38
rs989763042 Chr.11: 86809999-86809999 on Build GRCh38
rs1051618437 Chr.11: 86809685-86809685 on Build GRCh38
rs576195480 Chr.11: 86809970-86809970 on Build GRCh38
rs1023268509 Chr.11: 86809950-86809950 on Build GRCh38
rs1031200332 Chr.11: 86809996-86809996 on Build GRCh38
rs957977413 Chr.11: 86809985-86809985 on Build GRCh38
rs190388914 Chr.11: 86809557-86809557 on Build GRCh38
rs535140891 Chr.11: 86809859-86809859 on Build GRCh38
rs77287545 Chr.11: 86809553-86809553 on Build GRCh38
rs536301374 Chr.11: 86809887-86809887 on Build GRCh38
rs886248944 Chr.11: 86809712-86809712 on Build GRCh38
rs959811873 Chr.11: 86809876-86809876 on Build GRCh38
rs898773827 Chr.11: 86809818-86809818 on Build GRCh38
rs770120565 Chr.11: 86809901-86809901 on Build GRCh38
rs898870153 Chr.11: 86809895-86809897 on Build GRCh38
rs149983677 Chr.11: 86809998-86809998 on Build GRCh38
rs749505642 Chr.11: 86809949-86809949 on Build GRCh38
rs762873660 Chr.11: 86809650-86809651 on Build GRCh38
rs917638589 Chr.11: 86809583-86809583 on Build GRCh38
rs370825141 Chr.11: 86809653-86809653 on Build GRCh38
rs775965311 Chr.11: 86809853-86809853 on Build GRCh38
rs924078458 Chr.11: 86809567-86809567 on Build GRCh38
rs934057002 Chr.11: 86809705-86809705 on Build GRCh38
rs780311239 Chr.11: 86809903-86809903 on Build GRCh38
rs1013713194 Chr.11: 86809880-86809880 on Build GRCh38
rs1004635768 Chr.11: 86809779-86809779 on Build GRCh38
rs869107281 Chr.11: 86809915-86809916 on Build GRCh38
rs2155080 Chr.11: 86809590-86809590 on Build GRCh38
rs193211087 Chr.11: 86809656-86809656 on Build GRCh38
rs925221129 Chr.11: 86809595-86809595 on Build GRCh38
rs1036979197 Chr.11: 86809809-86809809 on Build GRCh38
rs369935331 Chr.11: 86809721-86809721 on Build GRCh38
rs933135487 Chr.11: 86809667-86809667 on Build GRCh38
rs757806225 Chr.11: 86809674-86809674 on Build GRCh38
rs554015539 Chr.11: 86809691-86809691 on Build GRCh38

View Full Product Details