Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 93167722-93168220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC36A4
Entrez Gene ID
120103
Gene Name
solute carrier family 36 member 4
Gene Aliases
PAT4
UniGene
Hs.148766
Gene Chromosome Location
Chr.11: 93144171-93197983 on Build GRCh38
Transcripts
NM_001286139.1 NM_152313.3 XM_017017174.1 XM_017017175.1 XM_017017176.1 XM_017017177.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2224378 c.633A>G p.I211M Chr.11: 93168079-93168079 on Build GRCh38
5927652 c.547G>A p.E183K Chr.11: 93168165-93168165 on Build GRCh38
4037644 c.764T>C p.V255A Chr.11: 93167948-93167948 on Build GRCh38
7157363 c.675C>T p.F225F Chr.11: 93168037-93168037 on Build GRCh38
1664212 c.757T>C p.Y253H Chr.11: 93167955-93167955 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371527871 Chr.11: 93168079-93168079 on Build GRCh38
rs762073615 Chr.11: 93168060-93168060 on Build GRCh38
rs760440376 Chr.11: 93167930-93167931 on Build GRCh38
rs754769165 Chr.11: 93168097-93168097 on Build GRCh38
rs369480830 Chr.11: 93167876-93167876 on Build GRCh38
rs201777152 Chr.11: 93168159-93168159 on Build GRCh38
rs143476889 Chr.11: 93168128-93168128 on Build GRCh38
rs772467424 Chr.11: 93168190-93168190 on Build GRCh38
rs1012025698 Chr.11: 93168047-93168047 on Build GRCh38
rs775840497 Chr.11: 93168032-93168032 on Build GRCh38
rs768391401 Chr.11: 93167900-93167900 on Build GRCh38
rs567112551 Chr.11: 93167814-93167814 on Build GRCh38
rs368143252 Chr.11: 93168163-93168163 on Build GRCh38
rs756031877 Chr.11: 93167985-93167985 on Build GRCh38
rs748618542 Chr.11: 93168174-93168175 on Build GRCh38
rs780680575 Chr.11: 93168153-93168153 on Build GRCh38
rs374736100 Chr.11: 93168093-93168093 on Build GRCh38
rs371169576 Chr.11: 93167993-93167993 on Build GRCh38
rs879203507 Chr.11: 93167970-93167970 on Build GRCh38
rs564532695 Chr.11: 93167992-93167992 on Build GRCh38
rs377658064 Chr.11: 93167936-93167936 on Build GRCh38
rs190929213 Chr.11: 93167833-93167833 on Build GRCh38
rs771563326 Chr.11: 93167911-93167911 on Build GRCh38
rs1028966700 Chr.11: 93167796-93167796 on Build GRCh38
rs112823142 Chr.11: 93168117-93168117 on Build GRCh38
rs778511831 Chr.11: 93167901-93167901 on Build GRCh38
rs376115076 Chr.11: 93168005-93168005 on Build GRCh38
rs964177974 Chr.11: 93167898-93167898 on Build GRCh38
rs764460873 Chr.11: 93167939-93167939 on Build GRCh38
rs760030683 Chr.11: 93167930-93167930 on Build GRCh38
rs367567712 Chr.11: 93168045-93168045 on Build GRCh38
rs762051458 Chr.11: 93167956-93167956 on Build GRCh38
rs112028381 Chr.11: 93167897-93167897 on Build GRCh38
rs774810348 Chr.11: 93168167-93168167 on Build GRCh38
rs183271008 Chr.11: 93167982-93167982 on Build GRCh38
rs542541567 Chr.11: 93167870-93167870 on Build GRCh38
rs773272275 Chr.11: 93168191-93168191 on Build GRCh38
rs775320791 Chr.11: 93167925-93167925 on Build GRCh38
rs767732185 Chr.11: 93168061-93168061 on Build GRCh38
rs575062361 Chr.11: 93167846-93167846 on Build GRCh38
rs527539896 Chr.11: 93168170-93168170 on Build GRCh38
rs912607805 Chr.11: 93167851-93167851 on Build GRCh38
rs146982814 Chr.11: 93168058-93168058 on Build GRCh38
rs538332481 Chr.11: 93167810-93167810 on Build GRCh38
rs746390824 Chr.11: 93168029-93168029 on Build GRCh38
rs751818878 Chr.11: 93167945-93167945 on Build GRCh38
rs116186359 Chr.11: 93167998-93167998 on Build GRCh38
rs570054127 Chr.11: 93168010-93168010 on Build GRCh38
rs113306357 Chr.11: 93167859-93167859 on Build GRCh38
rs915067617 Chr.11: 93167821-93167821 on Build GRCh38
rs764952824 Chr.11: 93168099-93168099 on Build GRCh38
rs966054856 Chr.11: 93167747-93167747 on Build GRCh38
rs143001920 Chr.11: 93168083-93168083 on Build GRCh38
rs764083872 Chr.11: 93167966-93167977 on Build GRCh38
rs974696746 Chr.11: 93167786-93167786 on Build GRCh38
rs112554756 Chr.11: 93167780-93167780 on Build GRCh38
rs769138426 Chr.11: 93168036-93168036 on Build GRCh38
rs200161055 Chr.11: 93168143-93168143 on Build GRCh38
rs757917081 Chr.11: 93167999-93167999 on Build GRCh38
rs767642095 Chr.11: 93167958-93167958 on Build GRCh38
rs374360768 Chr.11: 93167923-93167923 on Build GRCh38
rs771719551 Chr.11: 93167919-93167922 on Build GRCh38
rs770372713 Chr.11: 93168030-93168030 on Build GRCh38
rs200594142 Chr.11: 93167903-93167903 on Build GRCh38
rs148268542 Chr.11: 93167991-93167991 on Build GRCh38
rs749692751 Chr.11: 93168162-93168162 on Build GRCh38
rs762157383 Chr.11: 93167826-93167826 on Build GRCh38
rs1025765467 Chr.11: 93167761-93167761 on Build GRCh38
rs781676593 Chr.11: 93167979-93167980 on Build GRCh38
rs570967161 Chr.11: 93167749-93167749 on Build GRCh38
rs17854446 Chr.11: 93168087-93168087 on Build GRCh38
rs773305540 Chr.11: 93168076-93168076 on Build GRCh38
rs1025816105 Chr.11: 93168098-93168098 on Build GRCh38
rs748416732 Chr.11: 93168179-93168179 on Build GRCh38
rs756708847 Chr.11: 93168147-93168147 on Build GRCh38
rs770075806 Chr.11: 93167932-93167932 on Build GRCh38
rs777258856 Chr.11: 93168142-93168142 on Build GRCh38
rs989374559 Chr.11: 93167820-93167820 on Build GRCh38

View Full Product Details