Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 94546107-94546618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUT4
Entrez Gene ID
2526
Gene Name
fucosyltransferase 4
Gene Aliases
CD15, ELFT, FCT3A, FUC-TIV, FUTIV, LeX, SSEA-1
UniGene
Hs.390420
Gene Chromosome Location
Chr.11: 94543851-94549898 on Build GRCh38
Transcripts
NM_002033.3 NR_135093.1 NR_135094.1

Gene Symbol
LOC105369438
Entrez Gene ID
105369438
Gene Name
uncharacterized LOC105369438
Gene Aliases
-
UniGene
Hs.568921
Gene Chromosome Location
Chr.11: 94545332-94740355 on Build GRCh38
Transcripts
NM_002033.3 NR_135093.1 NR_135094.1

SNP Detail

SNP ID Chromosome Location
rs546679096 Chr.11: 94546380-94546380 on Build GRCh38
rs969524538 Chr.11: 94546197-94546197 on Build GRCh38
rs1013534369 Chr.11: 94546432-94546432 on Build GRCh38
rs1024799028 Chr.11: 94546438-94546438 on Build GRCh38
rs1039829425 Chr.11: 94546382-94546382 on Build GRCh38
rs930114917 Chr.11: 94546318-94546318 on Build GRCh38
rs565009470 Chr.11: 94546435-94546435 on Build GRCh38
rs945680865 Chr.11: 94546449-94546449 on Build GRCh38
rs139950114 Chr.11: 94546203-94546203 on Build GRCh38
rs969281381 Chr.11: 94546469-94546469 on Build GRCh38
rs551186512 Chr.11: 94546540-94546540 on Build GRCh38
rs114074063 Chr.11: 94546253-94546253 on Build GRCh38
rs575644812 Chr.11: 94546261-94546261 on Build GRCh38
rs906720392 Chr.11: 94546156-94546156 on Build GRCh38
rs897166332 Chr.11: 94546541-94546541 on Build GRCh38
rs1007379238 Chr.11: 94546346-94546346 on Build GRCh38
rs11574857 Chr.11: 94546509-94546509 on Build GRCh38
rs895397569 Chr.11: 94546233-94546233 on Build GRCh38
rs981227650 Chr.11: 94546455-94546455 on Build GRCh38
rs1039001075 Chr.11: 94546519-94546519 on Build GRCh38
rs867152061 Chr.11: 94546401-94546401 on Build GRCh38
rs1051221793 Chr.11: 94546319-94546319 on Build GRCh38
rs187124435 Chr.11: 94546180-94546180 on Build GRCh38
rs956494099 Chr.11: 94546178-94546178 on Build GRCh38
rs560913697 Chr.11: 94546311-94546311 on Build GRCh38
rs891324618 Chr.11: 94546321-94546321 on Build GRCh38
rs548957280 Chr.11: 94546563-94546563 on Build GRCh38
rs994560520 Chr.11: 94546561-94546561 on Build GRCh38
rs1018188846 Chr.11: 94546384-94546384 on Build GRCh38
rs980924945 Chr.11: 94546506-94546506 on Build GRCh38
rs772315100 Chr.11: 94546425-94546425 on Build GRCh38
rs758813259 Chr.11: 94546294-94546294 on Build GRCh38
rs1000879031 Chr.11: 94546404-94546404 on Build GRCh38
rs1030128793 Chr.11: 94546315-94546315 on Build GRCh38
rs957279899 Chr.11: 94546386-94546386 on Build GRCh38
rs1016943084 Chr.11: 94546274-94546274 on Build GRCh38
rs542636736 Chr.11: 94546290-94546290 on Build GRCh38
rs191641628 Chr.11: 94546325-94546325 on Build GRCh38
rs1055293079 Chr.11: 94546486-94546486 on Build GRCh38
rs528347683 Chr.11: 94546134-94546134 on Build GRCh38
rs878878784 Chr.11: 94546196-94546196 on Build GRCh38
rs939539122 Chr.11: 94546473-94546473 on Build GRCh38
rs532572594 Chr.11: 94546503-94546503 on Build GRCh38
rs1030896794 Chr.11: 94546405-94546405 on Build GRCh38
rs980574229 Chr.11: 94546201-94546201 on Build GRCh38
rs565861988 Chr.11: 94546458-94546459 on Build GRCh38
rs961633833 Chr.11: 94546286-94546286 on Build GRCh38
rs989993255 Chr.11: 94546391-94546391 on Build GRCh38

View Full Product Details