Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 100948263-100948764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP42
Entrez Gene ID
143872
Gene Name
Rho GTPase activating protein 42
Gene Aliases
GRAF3
UniGene
Hs.741465
Gene Chromosome Location
Chr.11: 100687676-100993941 on Build GRCh38
Transcripts
NM_152432.2 XM_011542615.2 XM_011542616.2 XM_011542617.2 XM_017017237.1 XM_017017238.1

SNP Detail

SNP ID Chromosome Location
rs1013199718 Chr.11: 100948592-100948592 on Build GRCh38
rs75461538 Chr.11: 100948677-100948677 on Build GRCh38
rs1027267274 Chr.11: 100948660-100948660 on Build GRCh38
rs914098187 Chr.11: 100948717-100948717 on Build GRCh38
rs1004797545 Chr.11: 100948435-100948435 on Build GRCh38
rs117118852 Chr.11: 100948610-100948610 on Build GRCh38
rs481991 Chr.11: 100948716-100948716 on Build GRCh38
rs796335006 Chr.11: 100948374-100948374 on Build GRCh38
rs761982384 Chr.11: 100948573-100948573 on Build GRCh38
rs533650168 Chr.11: 100948297-100948297 on Build GRCh38
rs926765208 Chr.11: 100948440-100948440 on Build GRCh38
rs943943800 Chr.11: 100948721-100948721 on Build GRCh38
rs890800037 Chr.11: 100948585-100948585 on Build GRCh38
rs760806302 Chr.11: 100948429-100948429 on Build GRCh38
rs1004538712 Chr.11: 100948667-100948667 on Build GRCh38
rs564797695 Chr.11: 100948295-100948295 on Build GRCh38
rs549216572 Chr.11: 100948362-100948362 on Build GRCh38
rs914147706 Chr.11: 100948458-100948458 on Build GRCh38
rs386756832 Chr.11: 100948699-100948716 on Build GRCh38
rs970746301 Chr.11: 100948372-100948372 on Build GRCh38
rs887244053 Chr.11: 100948432-100948432 on Build GRCh38
rs565823678 Chr.11: 100948377-100948377 on Build GRCh38
rs775296979 Chr.11: 100948416-100948416 on Build GRCh38
rs1054387334 Chr.11: 100948457-100948457 on Build GRCh38
rs1023294996 Chr.11: 100948598-100948598 on Build GRCh38
rs951401320 Chr.11: 100948291-100948291 on Build GRCh38
rs1007950327 Chr.11: 100948608-100948608 on Build GRCh38
rs529239065 Chr.11: 100948331-100948331 on Build GRCh38
rs995336653 Chr.11: 100948628-100948628 on Build GRCh38
rs980830466 Chr.11: 100948392-100948392 on Build GRCh38
rs527514762 Chr.11: 100948386-100948386 on Build GRCh38
rs11300533 Chr.11: 100948368-100948368 on Build GRCh38
rs190952077 Chr.11: 100948299-100948299 on Build GRCh38
rs633967 Chr.11: 100948574-100948574 on Build GRCh38
rs482050 Chr.11: 100948699-100948699 on Build GRCh38
rs572149265 Chr.11: 100948686-100948686 on Build GRCh38
rs767107658 Chr.11: 100948353-100948353 on Build GRCh38
rs7479352 Chr.11: 100948678-100948678 on Build GRCh38
rs970716819 Chr.11: 100948674-100948674 on Build GRCh38
rs541678411 Chr.11: 100948698-100948698 on Build GRCh38
rs374257795 Chr.11: 100948526-100948526 on Build GRCh38
rs899604993 Chr.11: 100948622-100948622 on Build GRCh38
rs1024897520 Chr.11: 100948669-100948669 on Build GRCh38
rs1039629146 Chr.11: 100948724-100948724 on Build GRCh38
rs563551893 Chr.11: 100948323-100948323 on Build GRCh38
rs504081 Chr.11: 100948661-100948661 on Build GRCh38
rs936873021 Chr.11: 100948446-100948446 on Build GRCh38
rs752155267 Chr.11: 100948570-100948570 on Build GRCh38
rs556378210 Chr.11: 100948637-100948637 on Build GRCh38

View Full Product Details