Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 101034612-101035111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGR
Entrez Gene ID
5241
Gene Name
progesterone receptor
Gene Aliases
NR3C3, PR
UniGene
Hs.32405
Gene Chromosome Location
Chr.11: 101029624-101130681 on Build GRCh38
Transcripts
NM_000926.4 NM_001202474.3 NM_001271161.2 NM_001271162.1 NR_073141.2 NR_073142.2 NR_073143.2

SNP Detail

SNP ID Chromosome Location
rs1039697797 Chr.11: 101035050-101035050 on Build GRCh38
rs576485973 Chr.11: 101034908-101034908 on Build GRCh38
rs947690389 Chr.11: 101034811-101034811 on Build GRCh38
rs1008272551 Chr.11: 101034715-101034715 on Build GRCh38
rs535554501 Chr.11: 101035020-101035020 on Build GRCh38
rs868375145 Chr.11: 101034751-101034751 on Build GRCh38
rs754339780 Chr.11: 101034990-101034990 on Build GRCh38
rs983427545 Chr.11: 101034774-101034774 on Build GRCh38
rs473409 Chr.11: 101034978-101034978 on Build GRCh38
rs768131616 Chr.11: 101034783-101034783 on Build GRCh38
rs1043501659 Chr.11: 101034971-101034971 on Build GRCh38
rs899853773 Chr.11: 101035056-101035056 on Build GRCh38
rs558473353 Chr.11: 101034740-101034740 on Build GRCh38
rs891117415 Chr.11: 101034634-101034634 on Build GRCh38
rs928715070 Chr.11: 101034763-101034763 on Build GRCh38
rs903566777 Chr.11: 101034938-101034939 on Build GRCh38
rs72563704 Chr.11: 101035065-101035065 on Build GRCh38
rs1043774067 Chr.11: 101034912-101034912 on Build GRCh38
rs186201243 Chr.11: 101034782-101034782 on Build GRCh38
rs999147633 Chr.11: 101034956-101034956 on Build GRCh38
rs116395595 Chr.11: 101034694-101034694 on Build GRCh38
rs1027546199 Chr.11: 101034755-101034755 on Build GRCh38
rs985184931 Chr.11: 101034675-101034675 on Build GRCh38
rs760137389 Chr.11: 101034762-101034762 on Build GRCh38
rs891009529 Chr.11: 101035027-101035027 on Build GRCh38
rs752049311 Chr.11: 101034705-101034705 on Build GRCh38
rs909249587 Chr.11: 101034677-101034677 on Build GRCh38
rs991744451 Chr.11: 101034813-101034813 on Build GRCh38
rs181867972 Chr.11: 101034667-101034667 on Build GRCh38
rs1008156468 Chr.11: 101035045-101035045 on Build GRCh38
rs190720887 Chr.11: 101034799-101034799 on Build GRCh38
rs1018290790 Chr.11: 101034726-101034726 on Build GRCh38
rs756537319 Chr.11: 101034935-101034935 on Build GRCh38
rs916253048 Chr.11: 101034837-101034837 on Build GRCh38
rs964090901 Chr.11: 101034734-101034734 on Build GRCh38
rs608995 Chr.11: 101035002-101035002 on Build GRCh38
rs753004389 Chr.11: 101034797-101034797 on Build GRCh38

View Full Product Details