Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 101914933-101915431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL5
Entrez Gene ID
253935
Gene Name
angiopoietin like 5
Gene Aliases
-
UniGene
Hs.318370
Gene Chromosome Location
Chr.11: 101890674-101916522 on Build GRCh38
Transcripts
NM_178127.4 XM_011542735.2 XM_017017466.1 NM_020802.3 XM_005271627.4

Gene Symbol
CEP126
Entrez Gene ID
57562
Gene Name
centrosomal protein 126
Gene Aliases
KIAA1377
UniGene
Hs.156352
Gene Chromosome Location
Chr.11: 101915015-102001065 on Build GRCh38
Transcripts
NM_178127.4 XM_011542735.2 XM_017017466.1 NM_020802.3 XM_005271627.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3935276 c.38G>A p.G13E Chr.11: 101915322-101915322 on Build GRCh38
7176965 c.59C>T p.S20L Chr.11: 101915343-101915343 on Build GRCh38
6694002 c.65A>T p.N22I Chr.11: 101915349-101915349 on Build GRCh38
6067035 c.96G>C p.E32D Chr.11: 101915380-101915380 on Build GRCh38
1745999 c.69C>T p.L23L Chr.11: 101915353-101915353 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146972831 Chr.11: 101915360-101915360 on Build GRCh38
rs536297070 Chr.11: 101915395-101915395 on Build GRCh38
rs779715132 Chr.11: 101915375-101915376 on Build GRCh38
rs200573063 Chr.11: 101915364-101915364 on Build GRCh38
rs939149970 Chr.11: 101915163-101915163 on Build GRCh38
rs911490223 Chr.11: 101915074-101915074 on Build GRCh38
rs374283089 Chr.11: 101915008-101915008 on Build GRCh38
rs150590684 Chr.11: 101915318-101915318 on Build GRCh38
rs761812843 Chr.11: 101915236-101915236 on Build GRCh38
rs926189454 Chr.11: 101915206-101915206 on Build GRCh38
rs577764251 Chr.11: 101915211-101915211 on Build GRCh38
rs751753088 Chr.11: 101915036-101915036 on Build GRCh38
rs149781214 Chr.11: 101915289-101915289 on Build GRCh38
rs57569155 Chr.11: 101915106-101915106 on Build GRCh38
rs951445862 Chr.11: 101915257-101915257 on Build GRCh38
rs755280571 Chr.11: 101915038-101915038 on Build GRCh38
rs776490810 Chr.11: 101915286-101915286 on Build GRCh38
rs994495203 Chr.11: 101915105-101915105 on Build GRCh38
rs1001644314 Chr.11: 101915346-101915346 on Build GRCh38
rs955523711 Chr.11: 101915062-101915062 on Build GRCh38
rs1051620477 Chr.11: 101915072-101915072 on Build GRCh38
rs779191649 Chr.11: 101915014-101915014 on Build GRCh38
rs751919013 Chr.11: 101915248-101915248 on Build GRCh38
rs148913485 Chr.11: 101915309-101915309 on Build GRCh38
rs181761204 Chr.11: 101915317-101915317 on Build GRCh38
rs769861010 Chr.11: 101915296-101915296 on Build GRCh38
rs934091173 Chr.11: 101915251-101915251 on Build GRCh38
rs967826207 Chr.11: 101915055-101915055 on Build GRCh38
rs993024948 Chr.11: 101914974-101914974 on Build GRCh38
rs1039000697 Chr.11: 101915280-101915280 on Build GRCh38
rs374652030 Chr.11: 101915256-101915256 on Build GRCh38
rs533605419 Chr.11: 101915190-101915190 on Build GRCh38
rs759755371 Chr.11: 101915316-101915316 on Build GRCh38
rs913198505 Chr.11: 101915250-101915250 on Build GRCh38
rs186513569 Chr.11: 101915056-101915056 on Build GRCh38
rs1012539266 Chr.11: 101915181-101915181 on Build GRCh38
rs543347915 Chr.11: 101914957-101914957 on Build GRCh38
rs115730613 Chr.11: 101915069-101915069 on Build GRCh38
rs925988987 Chr.11: 101915053-101915053 on Build GRCh38
rs935008107 Chr.11: 101915308-101915308 on Build GRCh38
rs765313268 Chr.11: 101915237-101915237 on Build GRCh38
rs758032724 Chr.11: 101915269-101915269 on Build GRCh38
rs980359582 Chr.11: 101915194-101915194 on Build GRCh38
rs778562178 Chr.11: 101915240-101915241 on Build GRCh38
rs749850977 Chr.11: 101915324-101915324 on Build GRCh38
rs757955825 Chr.11: 101915343-101915343 on Build GRCh38
rs776580879 Chr.11: 101915235-101915235 on Build GRCh38
rs1043438027 Chr.11: 101915189-101915189 on Build GRCh38
rs746741985 Chr.11: 101915285-101915285 on Build GRCh38
rs201779479 Chr.11: 101915302-101915302 on Build GRCh38
rs1038754710 Chr.11: 101914989-101914989 on Build GRCh38
rs865798047 Chr.11: 101915361-101915361 on Build GRCh38
rs756252505 Chr.11: 101915149-101915149 on Build GRCh38
rs745482845 Chr.11: 101915268-101915268 on Build GRCh38
rs1044020803 Chr.11: 101915298-101915298 on Build GRCh38
rs757955230 Chr.11: 101915360-101915361 on Build GRCh38
rs988782896 Chr.11: 101915225-101915225 on Build GRCh38
rs1050359388 Chr.11: 101915079-101915080 on Build GRCh38
rs368560519 Chr.11: 101915247-101915247 on Build GRCh38
rs756601462 Chr.11: 101915026-101915026 on Build GRCh38
rs190513535 Chr.11: 101915255-101915255 on Build GRCh38
rs919884351 Chr.11: 101915101-101915101 on Build GRCh38
rs1031148678 Chr.11: 101915212-101915212 on Build GRCh38
rs142643716 Chr.11: 101915016-101915016 on Build GRCh38
rs765042538 Chr.11: 101915018-101915018 on Build GRCh38
rs374130773 Chr.11: 101914992-101914992 on Build GRCh38
rs117883580 Chr.11: 101915283-101915283 on Build GRCh38
rs748433566 Chr.11: 101915301-101915301 on Build GRCh38
rs898857744 Chr.11: 101915091-101915091 on Build GRCh38
rs950515899 Chr.11: 101915111-101915111 on Build GRCh38
rs753263798 Chr.11: 101915259-101915259 on Build GRCh38
rs1056382134 Chr.11: 101915035-101915035 on Build GRCh38
rs974110709 Chr.11: 101915100-101915100 on Build GRCh38
rs1047431691 Chr.11: 101915293-101915293 on Build GRCh38
rs903507591 Chr.11: 101915207-101915207 on Build GRCh38
rs928723371 Chr.11: 101915267-101915267 on Build GRCh38
rs368939235 Chr.11: 101915265-101915265 on Build GRCh38
rs748048053 Chr.11: 101915366-101915366 on Build GRCh38
rs942683942 Chr.11: 101914972-101914972 on Build GRCh38
rs746685985 Chr.11: 101915344-101915344 on Build GRCh38
rs750002677 Chr.11: 101915293-101915293 on Build GRCh38
rs564510732 Chr.11: 101915172-101915172 on Build GRCh38
rs1007511092 Chr.11: 101915075-101915075 on Build GRCh38
rs1011931505 Chr.11: 101915042-101915042 on Build GRCh38
rs964237105 Chr.11: 101915081-101915081 on Build GRCh38
rs749770036 Chr.11: 101915176-101915176 on Build GRCh38
rs754752744 Chr.11: 101915349-101915349 on Build GRCh38
rs145606538 Chr.11: 101915297-101915297 on Build GRCh38
rs562134120 Chr.11: 101915057-101915057 on Build GRCh38
rs917381800 Chr.11: 101914998-101914998 on Build GRCh38
rs1003496468 Chr.11: 101915019-101915019 on Build GRCh38
rs764814199 Chr.11: 101915321-101915321 on Build GRCh38
rs768428583 Chr.11: 101915234-101915234 on Build GRCh38
rs941338537 Chr.11: 101915058-101915058 on Build GRCh38
rs1047583121 Chr.11: 101915113-101915113 on Build GRCh38
rs1022343088 Chr.11: 101915043-101915043 on Build GRCh38

View Full Product Details