Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 102772688-102773218 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMP10
Entrez Gene ID
4319
Gene Name
matrix metallopeptidase 10
Gene Aliases
SL-2, STMY2
UniGene
Hs.2258
Gene Chromosome Location
Chr.11: 102770502-102780628 on Build GRCh38
Transcripts
NM_002425.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5902342 c.1135G>A p.G379S Chr.11: 102772938-102772938 on Build GRCh38
5553352 c.1098G>T p.E366D Chr.11: 102772975-102772975 on Build GRCh38
7206058 c.1098G>A p.E366E Chr.11: 102772975-102772975 on Build GRCh38
4807413 c.1122C>A p.G374G Chr.11: 102772951-102772951 on Build GRCh38
4702664 c.1190A>C p.K397T Chr.11: 102772883-102772883 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550680862 Chr.11: 102772719-102772719 on Build GRCh38
rs139168339 Chr.11: 102772905-102772905 on Build GRCh38
rs998466011 Chr.11: 102773177-102773177 on Build GRCh38
rs774227794 Chr.11: 102772941-102772941 on Build GRCh38
rs953819732 Chr.11: 102772923-102772923 on Build GRCh38
rs949959809 Chr.11: 102773093-102773093 on Build GRCh38
rs138753435 Chr.11: 102772862-102772862 on Build GRCh38
rs937997284 Chr.11: 102773123-102773123 on Build GRCh38
rs190882512 Chr.11: 102773195-102773195 on Build GRCh38
rs1021200927 Chr.11: 102772881-102772881 on Build GRCh38
rs760619596 Chr.11: 102773057-102773057 on Build GRCh38
rs754035907 Chr.11: 102772955-102772955 on Build GRCh38
rs946761985 Chr.11: 102773160-102773160 on Build GRCh38
rs17359286 Chr.11: 102772987-102772987 on Build GRCh38
rs370589543 Chr.11: 102773088-102773088 on Build GRCh38
rs769229991 Chr.11: 102772999-102772999 on Build GRCh38
rs758757062 Chr.11: 102772983-102772983 on Build GRCh38
rs143648713 Chr.11: 102772867-102772867 on Build GRCh38
rs142994202 Chr.11: 102773025-102773025 on Build GRCh38
rs1049872685 Chr.11: 102772773-102772773 on Build GRCh38
rs998811827 Chr.11: 102772902-102772902 on Build GRCh38
rs764201572 Chr.11: 102772953-102772953 on Build GRCh38
rs111826228 Chr.11: 102772720-102772720 on Build GRCh38
rs767625727 Chr.11: 102772948-102772948 on Build GRCh38
rs17860992 Chr.11: 102772774-102772774 on Build GRCh38
rs770637354 Chr.11: 102773009-102773009 on Build GRCh38
rs375698563 Chr.11: 102772840-102772840 on Build GRCh38
rs188002289 Chr.11: 102773164-102773164 on Build GRCh38
rs975958791 Chr.11: 102772714-102772714 on Build GRCh38
rs751590059 Chr.11: 102772858-102772858 on Build GRCh38
rs746398386 Chr.11: 102772882-102772882 on Build GRCh38
rs928008349 Chr.11: 102773112-102773112 on Build GRCh38
rs756210031 Chr.11: 102772864-102772864 on Build GRCh38
rs918388605 Chr.11: 102773087-102773087 on Build GRCh38
rs772511075 Chr.11: 102772891-102772891 on Build GRCh38
rs879287804 Chr.11: 102772716-102772716 on Build GRCh38
rs372673583 Chr.11: 102772926-102772926 on Build GRCh38
rs761541280 Chr.11: 102772819-102772819 on Build GRCh38
rs377147064 Chr.11: 102772931-102772931 on Build GRCh38
rs745711567 Chr.11: 102773026-102773026 on Build GRCh38
rs769264520 Chr.11: 102772827-102772827 on Build GRCh38
rs1055249544 Chr.11: 102773124-102773124 on Build GRCh38
rs1046399818 Chr.11: 102772776-102772776 on Build GRCh38
rs370615545 Chr.11: 102773003-102773003 on Build GRCh38
rs949907095 Chr.11: 102772775-102772775 on Build GRCh38
rs147267769 Chr.11: 102772873-102772873 on Build GRCh38
rs771970013 Chr.11: 102773027-102773027 on Build GRCh38
rs867163970 Chr.11: 102772969-102772969 on Build GRCh38
rs777300384 Chr.11: 102773002-102773002 on Build GRCh38
rs752668633 Chr.11: 102772952-102772952 on Build GRCh38
rs368858787 Chr.11: 102772896-102772896 on Build GRCh38
rs985522711 Chr.11: 102773130-102773130 on Build GRCh38
rs779264298 Chr.11: 102772964-102772964 on Build GRCh38
rs537908479 Chr.11: 102772918-102772918 on Build GRCh38
rs547561 Chr.11: 102773163-102773163 on Build GRCh38
rs906496555 Chr.11: 102772782-102772782 on Build GRCh38
rs772918078 Chr.11: 102772829-102772829 on Build GRCh38
rs369950180 Chr.11: 102773061-102773061 on Build GRCh38
rs562311932 Chr.11: 102773178-102773178 on Build GRCh38
rs893889196 Chr.11: 102773134-102773134 on Build GRCh38
rs772793272 Chr.11: 102772922-102772922 on Build GRCh38
rs750720728 Chr.11: 102772978-102772978 on Build GRCh38
rs375465947 Chr.11: 102772863-102772863 on Build GRCh38
rs999161529 Chr.11: 102772970-102772970 on Build GRCh38
rs747532161 Chr.11: 102772995-102772995 on Build GRCh38
rs766177134 Chr.11: 102772847-102772847 on Build GRCh38
rs779207038 Chr.11: 102772876-102772876 on Build GRCh38
rs775329513 Chr.11: 102773038-102773038 on Build GRCh38
rs536226793 Chr.11: 102772807-102772807 on Build GRCh38
rs757454171 Chr.11: 102772960-102772960 on Build GRCh38
rs558198018 Chr.11: 102772861-102772861 on Build GRCh38
rs1036980475 Chr.11: 102772758-102772758 on Build GRCh38
rs1046035505 Chr.11: 102773099-102773099 on Build GRCh38
rs746199024 Chr.11: 102772878-102772878 on Build GRCh38
rs149436433 Chr.11: 102772943-102772943 on Build GRCh38
rs17860989 Chr.11: 102773166-102773167 on Build GRCh38

View Full Product Details