Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 105612185-105612683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIA4
Entrez Gene ID
2893
Gene Name
glutamate ionotropic receptor AMPA type subunit 4
Gene Aliases
GLUR4, GLUR4C, GLURD, GluA4
UniGene
Hs.503743
Gene Chromosome Location
Chr.11: 105609994-105982092 on Build GRCh38
Transcripts
NM_000829.3 NM_001077243.2 NM_001077244.1 NM_001112812.1 NR_046356.1 XM_005271518.3 XM_006718823.1 XM_011542775.2 XM_017017609.1 XM_017017610.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7370233 c.170C>A p.A57E Chr.11: 105612357-105612357 on Build GRCh38
7168387 c.190T>A p.L64M Chr.11: 105612377-105612377 on Build GRCh38
6865442 c.244G>A p.A82T Chr.11: 105612431-105612431 on Build GRCh38
7202906 c.191T>C p.L64S Chr.11: 105612378-105612378 on Build GRCh38
7078162 c.214G>C p.E72Q Chr.11: 105612401-105612401 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs398017426 Chr.11: 105612617-105612617 on Build GRCh38
rs756216312 Chr.11: 105612364-105612364 on Build GRCh38
rs1052151574 Chr.11: 105612253-105612253 on Build GRCh38
rs564247210 Chr.11: 105612259-105612259 on Build GRCh38
rs762714307 Chr.11: 105612291-105612291 on Build GRCh38
rs945508059 Chr.11: 105612600-105612600 on Build GRCh38
rs761365633 Chr.11: 105612281-105612281 on Build GRCh38
rs911750222 Chr.11: 105612606-105612607 on Build GRCh38
rs748195964 Chr.11: 105612233-105612233 on Build GRCh38
rs915428093 Chr.11: 105612588-105612588 on Build GRCh38
rs767509481 Chr.11: 105612360-105612360 on Build GRCh38
rs779280848 Chr.11: 105612453-105612453 on Build GRCh38
rs945165069 Chr.11: 105612596-105612596 on Build GRCh38
rs914960444 Chr.11: 105612573-105612573 on Build GRCh38
rs760839370 Chr.11: 105612240-105612240 on Build GRCh38
rs5794413 Chr.11: 105612608-105612608 on Build GRCh38
rs990110133 Chr.11: 105612585-105612585 on Build GRCh38
rs963152620 Chr.11: 105612400-105612400 on Build GRCh38
rs751328080 Chr.11: 105612341-105612341 on Build GRCh38
rs193111893 Chr.11: 105612583-105612583 on Build GRCh38
rs368282384 Chr.11: 105612449-105612449 on Build GRCh38
rs1016109337 Chr.11: 105612385-105612385 on Build GRCh38
rs769333448 Chr.11: 105612482-105612482 on Build GRCh38
rs993226604 Chr.11: 105612405-105612405 on Build GRCh38
rs981797165 Chr.11: 105612511-105612511 on Build GRCh38
rs970396828 Chr.11: 105612469-105612469 on Build GRCh38
rs199606404 Chr.11: 105612463-105612463 on Build GRCh38
rs754993716 Chr.11: 105612344-105612344 on Build GRCh38
rs777918745 Chr.11: 105612406-105612406 on Build GRCh38
rs772529297 Chr.11: 105612260-105612260 on Build GRCh38
rs749542799 Chr.11: 105612447-105612447 on Build GRCh38
rs1042523700 Chr.11: 105612609-105612609 on Build GRCh38
rs778028122 Chr.11: 105612247-105612247 on Build GRCh38
rs752871049 Chr.11: 105612361-105612361 on Build GRCh38
rs201288803 Chr.11: 105612617-105612617 on Build GRCh38
rs879157846 Chr.11: 105612473-105612473 on Build GRCh38
rs772912032 Chr.11: 105612289-105612289 on Build GRCh38
rs766289231 Chr.11: 105612329-105612329 on Build GRCh38
rs144198453 Chr.11: 105612266-105612266 on Build GRCh38
rs926267250 Chr.11: 105612529-105612529 on Build GRCh38
rs572356326 Chr.11: 105612472-105612472 on Build GRCh38
rs952775999 Chr.11: 105612367-105612367 on Build GRCh38
rs201112609 Chr.11: 105612618-105612618 on Build GRCh38
rs982407027 Chr.11: 105612514-105612514 on Build GRCh38
rs1004350206 Chr.11: 105612455-105612455 on Build GRCh38
rs989556153 Chr.11: 105612560-105612560 on Build GRCh38
rs760040690 Chr.11: 105612254-105612254 on Build GRCh38
rs531693868 Chr.11: 105612575-105612575 on Build GRCh38
rs749684779 Chr.11: 105612250-105612250 on Build GRCh38
rs926242574 Chr.11: 105612513-105612513 on Build GRCh38
rs149589990 Chr.11: 105612532-105612532 on Build GRCh38
rs774645253 Chr.11: 105612252-105612252 on Build GRCh38
rs775943675 Chr.11: 105612467-105612467 on Build GRCh38
rs769927999 Chr.11: 105612243-105612243 on Build GRCh38
rs571086344 Chr.11: 105612286-105612286 on Build GRCh38
rs199564574 Chr.11: 105612618-105612618 on Build GRCh38
rs143276365 Chr.11: 105612626-105612626 on Build GRCh38
rs554661076 Chr.11: 105612552-105612552 on Build GRCh38
rs768690195 Chr.11: 105612265-105612266 on Build GRCh38
rs117866514 Chr.11: 105612460-105612460 on Build GRCh38
rs1038922810 Chr.11: 105612622-105612622 on Build GRCh38
rs772110241 Chr.11: 105612397-105612397 on Build GRCh38
rs137983690 Chr.11: 105612267-105612267 on Build GRCh38

View Full Product Details