Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 107630613-107631112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMOD1
Entrez Gene ID
55531
Gene Name
ELMO domain containing 1
Gene Aliases
-
UniGene
Hs.495779
Gene Chromosome Location
Chr.11: 107591091-107666779 on Build GRCh38
Transcripts
NM_001130037.1 NM_001308018.1 NM_018712.3 XM_017017994.1 XM_017017995.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6067472 c.182C>G p.S61C Chr.11: 107630718-107630718 on Build GRCh38
274961 c.178G>A p.D60N Chr.11: 107630714-107630714 on Build GRCh38
6519556 c.189T>A p.S63R Chr.11: 107630725-107630725 on Build GRCh38
3443086 c.177G>A p.R59R Chr.11: 107630713-107630713 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755107181 Chr.11: 107630753-107630753 on Build GRCh38
rs552381590 Chr.11: 107630707-107630707 on Build GRCh38
rs183679099 Chr.11: 107631035-107631035 on Build GRCh38
rs533022167 Chr.11: 107630845-107630845 on Build GRCh38
rs746680638 Chr.11: 107630754-107630754 on Build GRCh38
rs759022154 Chr.11: 107631056-107631056 on Build GRCh38
rs56204863 Chr.11: 107631050-107631050 on Build GRCh38
rs577481740 Chr.11: 107631089-107631089 on Build GRCh38
rs866840825 Chr.11: 107630740-107630740 on Build GRCh38
rs747052545 Chr.11: 107630741-107630742 on Build GRCh38
rs760597246 Chr.11: 107630643-107630643 on Build GRCh38
rs117224984 Chr.11: 107630968-107630968 on Build GRCh38
rs946595253 Chr.11: 107631068-107631068 on Build GRCh38
rs199579878 Chr.11: 107630708-107630708 on Build GRCh38
rs1052242829 Chr.11: 107631045-107631045 on Build GRCh38
rs538815418 Chr.11: 107630882-107630882 on Build GRCh38
rs779860434 Chr.11: 107630669-107630672 on Build GRCh38
rs1011334254 Chr.11: 107630639-107630639 on Build GRCh38
rs779197750 Chr.11: 107630715-107630715 on Build GRCh38
rs982930257 Chr.11: 107630872-107630872 on Build GRCh38
rs953165162 Chr.11: 107630930-107630930 on Build GRCh38
rs780752220 Chr.11: 107630760-107630760 on Build GRCh38
rs940796230 Chr.11: 107631044-107631044 on Build GRCh38
rs757649712 Chr.11: 107630739-107630739 on Build GRCh38
rs935165159 Chr.11: 107631037-107631037 on Build GRCh38
rs1029867524 Chr.11: 107630859-107630859 on Build GRCh38
rs754383017 Chr.11: 107630755-107630755 on Build GRCh38
rs748419930 Chr.11: 107630754-107630754 on Build GRCh38
rs973060527 Chr.11: 107630907-107630907 on Build GRCh38
rs916230027 Chr.11: 107630938-107630938 on Build GRCh38
rs927859753 Chr.11: 107630981-107630981 on Build GRCh38
rs569906567 Chr.11: 107630695-107630695 on Build GRCh38
rs781387822 Chr.11: 107630753-107630754 on Build GRCh38
rs754171634 Chr.11: 107630714-107630714 on Build GRCh38
rs989827201 Chr.11: 107630857-107630857 on Build GRCh38
rs962963167 Chr.11: 107630902-107630902 on Build GRCh38
rs970452431 Chr.11: 107630817-107630817 on Build GRCh38
rs779637815 Chr.11: 107630747-107630747 on Build GRCh38
rs1031119387 Chr.11: 107630833-107630833 on Build GRCh38
rs372847998 Chr.11: 107630787-107630787 on Build GRCh38
rs762889330 Chr.11: 107630665-107630665 on Build GRCh38
rs981819448 Chr.11: 107630970-107630970 on Build GRCh38
rs751861529 Chr.11: 107630687-107630687 on Build GRCh38
rs747667925 Chr.11: 107630777-107630777 on Build GRCh38
rs56218117 Chr.11: 107630800-107630800 on Build GRCh38
rs766676566 Chr.11: 107630667-107630667 on Build GRCh38
rs1056420541 Chr.11: 107631091-107631091 on Build GRCh38
rs554558625 Chr.11: 107631085-107631085 on Build GRCh38
rs375813204 Chr.11: 107630892-107630892 on Build GRCh38
rs999196941 Chr.11: 107630818-107630818 on Build GRCh38
rs1024645849 Chr.11: 107630801-107630801 on Build GRCh38
rs951626257 Chr.11: 107630864-107630864 on Build GRCh38
rs998255401 Chr.11: 107630840-107630840 on Build GRCh38
rs201717101 Chr.11: 107630702-107630702 on Build GRCh38

View Full Product Details