Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111836087-111836600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG9
Entrez Gene ID
79796
Gene Name
ALG9, alpha-1,2-mannosyltransferase
Gene Aliases
CDG1L, DIBD1, GIKANIS, LOH11CR1J
UniGene
Hs.745155
Gene Chromosome Location
Chr.11: 111776096-111871581 on Build GRCh38
Transcripts
NM_001077690.1 NM_001077691.1 NM_001077692.1 NM_024740.2 XM_005277723.4 XM_005277724.4 XM_006718913.2 XM_011542992.1 XM_017018313.1 XM_017018314.1 XM_017018315.1 XM_017018316.1 XM_017018317.1 XM_017018318.1 XM_017018319.1 XM_017018320.1 XM_017018321.1 XM_017018322.1 XM_017018323.1 XM_017018324.1 XM_017018325.1 XM_017018326.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5739449 c.986A>G p.E329G Chr.11: 111836268-111836268 on Build GRCh38
1193262 c.976A>G p.I326V Chr.11: 111836278-111836278 on Build GRCh38
3443374 c.970C>T p.Q324* Chr.11: 111836284-111836284 on Build GRCh38
4760150 c.1055T>C p.M352T Chr.11: 111836199-111836199 on Build GRCh38
686398 c.1035C>A p.T345T Chr.11: 111836219-111836219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs967976781 Chr.11: 111836453-111836453 on Build GRCh38
rs782252831 Chr.11: 111836294-111836294 on Build GRCh38
rs782004306 Chr.11: 111836217-111836217 on Build GRCh38
rs376963147 Chr.11: 111836221-111836221 on Build GRCh38
rs782531411 Chr.11: 111836256-111836256 on Build GRCh38
rs138584071 Chr.11: 111836278-111836278 on Build GRCh38
rs782451691 Chr.11: 111836117-111836117 on Build GRCh38
rs541537158 Chr.11: 111836265-111836265 on Build GRCh38
rs782652453 Chr.11: 111836284-111836284 on Build GRCh38
rs782699331 Chr.11: 111836245-111836245 on Build GRCh38
rs139745447 Chr.11: 111836541-111836541 on Build GRCh38
rs143975084 Chr.11: 111836346-111836346 on Build GRCh38
rs782371858 Chr.11: 111836129-111836129 on Build GRCh38
rs782175992 Chr.11: 111836279-111836279 on Build GRCh38
rs782639398 Chr.11: 111836259-111836259 on Build GRCh38
rs782677859 Chr.11: 111836119-111836119 on Build GRCh38
rs1008067161 Chr.11: 111836216-111836216 on Build GRCh38
rs782248933 Chr.11: 111836239-111836240 on Build GRCh38
rs369596048 Chr.11: 111836267-111836267 on Build GRCh38
rs889705060 Chr.11: 111836205-111836205 on Build GRCh38
rs926023630 Chr.11: 111836191-111836191 on Build GRCh38
rs1043555793 Chr.11: 111836168-111836168 on Build GRCh38
rs369901475 Chr.11: 111836159-111836159 on Build GRCh38
rs782503370 Chr.11: 111836428-111836428 on Build GRCh38
rs782375085 Chr.11: 111836297-111836297 on Build GRCh38
rs782168420 Chr.11: 111836345-111836345 on Build GRCh38
rs1040960434 Chr.11: 111836218-111836218 on Build GRCh38
rs782615926 Chr.11: 111836144-111836144 on Build GRCh38
rs978926335 Chr.11: 111836471-111836471 on Build GRCh38
rs187507214 Chr.11: 111836308-111836308 on Build GRCh38
rs782752564 Chr.11: 111836226-111836226 on Build GRCh38
rs993524838 Chr.11: 111836410-111836410 on Build GRCh38
rs150673768 Chr.11: 111836493-111836493 on Build GRCh38
rs553047310 Chr.11: 111836381-111836381 on Build GRCh38
rs373098545 Chr.11: 111836116-111836116 on Build GRCh38
rs374442576 Chr.11: 111836273-111836273 on Build GRCh38
rs782412841 Chr.11: 111836282-111836282 on Build GRCh38
rs1050364950 Chr.11: 111836203-111836203 on Build GRCh38
rs782403977 Chr.11: 111836209-111836209 on Build GRCh38
rs781907541 Chr.11: 111836255-111836255 on Build GRCh38
rs781912775 Chr.11: 111836198-111836198 on Build GRCh38
rs542390814 Chr.11: 111836500-111836500 on Build GRCh38
rs121908022 Chr.11: 111836179-111836179 on Build GRCh38
rs782276266 Chr.11: 111836121-111836121 on Build GRCh38
rs548268879 Chr.11: 111836299-111836299 on Build GRCh38
rs191613774 Chr.11: 111836366-111836366 on Build GRCh38
rs377678568 Chr.11: 111836150-111836150 on Build GRCh38
rs782117315 Chr.11: 111836225-111836225 on Build GRCh38
rs377345625 Chr.11: 111836234-111836234 on Build GRCh38
rs932029764 Chr.11: 111836498-111836498 on Build GRCh38
rs534487519 Chr.11: 111836369-111836369 on Build GRCh38
rs781921370 Chr.11: 111836329-111836329 on Build GRCh38
rs185149177 Chr.11: 111836229-111836229 on Build GRCh38
rs782264822 Chr.11: 111836353-111836353 on Build GRCh38
rs781903223 Chr.11: 111836183-111836183 on Build GRCh38
rs573557743 Chr.11: 111836348-111836348 on Build GRCh38
rs374283538 Chr.11: 111836311-111836311 on Build GRCh38

View Full Product Details