Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 124919159-124919666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEPACAM
Entrez Gene ID
220296
Gene Name
hepatic and glial cell adhesion molecule
Gene Aliases
GlialCAM, MLC2A, MLC2B
UniGene
Hs.745294
Gene Chromosome Location
Chr.11: 124919244-124936412 on Build GRCh38
Transcripts
NM_152722.4 XM_005271449.2 XM_017017361.1 NM_001037558.2

Gene Symbol
HEPN1
Entrez Gene ID
641654
Gene Name
hepatocellular carcinoma, down-regulated 1
Gene Aliases
-
UniGene
Hs.159863
Gene Chromosome Location
Chr.11: 124919250-124920677 on Build GRCh38
Transcripts
NM_152722.4 XM_005271449.2 XM_017017361.1 NM_001037558.2

SNP Detail

SNP ID Chromosome Location
rs997388358 Chr.11: 124919212-124919212 on Build GRCh38
rs1033822977 Chr.11: 124919442-124919442 on Build GRCh38
rs202149246 Chr.11: 124919334-124919335 on Build GRCh38
rs992348645 Chr.11: 124919469-124919469 on Build GRCh38
rs769331413 Chr.11: 124919189-124919189 on Build GRCh38
rs1001543144 Chr.11: 124919418-124919418 on Build GRCh38
rs1016043523 Chr.11: 124919635-124919635 on Build GRCh38
rs899126537 Chr.11: 124919640-124919640 on Build GRCh38
rs886047917 Chr.11: 124919339-124919339 on Build GRCh38
rs375793524 Chr.11: 124919213-124919213 on Build GRCh38
rs560243987 Chr.11: 124919336-124919337 on Build GRCh38
rs891250803 Chr.11: 124919266-124919266 on Build GRCh38
rs544201825 Chr.11: 124919501-124919501 on Build GRCh38
rs756861875 Chr.11: 124919377-124919377 on Build GRCh38
rs374156953 Chr.11: 124919488-124919488 on Build GRCh38
rs571076673 Chr.11: 124919557-124919557 on Build GRCh38
rs947658025 Chr.11: 124919344-124919344 on Build GRCh38
rs753327871 Chr.11: 124919349-124919349 on Build GRCh38
rs959790154 Chr.11: 124919459-124919459 on Build GRCh38
rs559039197 Chr.11: 124919217-124919217 on Build GRCh38
rs900348534 Chr.11: 124919200-124919200 on Build GRCh38
rs1051582667 Chr.11: 124919641-124919641 on Build GRCh38
rs554825783 Chr.11: 124919494-124919494 on Build GRCh38
rs1020918223 Chr.11: 124919334-124919334 on Build GRCh38
rs200549730 Chr.11: 124919330-124919336 on Build GRCh38
rs1004636554 Chr.11: 124919581-124919581 on Build GRCh38
rs112607377 Chr.11: 124919286-124919286 on Build GRCh38
rs1051110692 Chr.11: 124919247-124919247 on Build GRCh38
rs370484067 Chr.11: 124919292-124919292 on Build GRCh38
rs368339941 Chr.11: 124919543-124919543 on Build GRCh38
rs914796300 Chr.11: 124919328-124919328 on Build GRCh38
rs886273672 Chr.11: 124919580-124919580 on Build GRCh38
rs538418045 Chr.11: 124919561-124919561 on Build GRCh38
rs904064000 Chr.11: 124919414-124919414 on Build GRCh38
rs921895525 Chr.11: 124919624-124919624 on Build GRCh38
rs1024725774 Chr.11: 124919497-124919497 on Build GRCh38
rs905511555 Chr.11: 124919464-124919464 on Build GRCh38
rs983078405 Chr.11: 124919502-124919503 on Build GRCh38
rs79225627 Chr.11: 124919524-124919524 on Build GRCh38
rs975055231 Chr.11: 124919604-124919604 on Build GRCh38
rs934793513 Chr.11: 124919226-124919226 on Build GRCh38

View Full Product Details