Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 125582341-125582858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EI24
Entrez Gene ID
9538
Gene Name
EI24, autophagy associated transmembrane protein
Gene Aliases
EPG4, PIG8, TP53I8
UniGene
Hs.643514
Gene Chromosome Location
Chr.11: 125569216-125584689 on Build GRCh38
Transcripts
NM_001290135.1 NM_004879.4 NR_110769.1 NR_110770.1 XM_011543069.1 XM_011543070.1 XM_017018582.1 NR_132372.1

Gene Symbol
STT3A-AS1
Entrez Gene ID
105369550
Gene Name
STT3A antisense RNA 1
Gene Aliases
-
UniGene
Hs.732083
Gene Chromosome Location
Chr.11: 125568108-125592568 on Build GRCh38
Transcripts
NM_001290135.1 NM_004879.4 NR_110769.1 NR_110770.1 XM_011543069.1 XM_011543070.1 XM_017018582.1 NR_132372.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4603370 c.843+1_843+2insC p.? Chr.11: 125582406-125582406 on Build GRCh38
5022172 c.844-1_844insC p.G283fs*>58 Chr.11: 125582406-125582407 on Build GRCh38
5989492 c.798C>T p.F266F Chr.11: 125582358-125582358 on Build GRCh38
7248127 c.855A>G p.A285A Chr.11: 125582418-125582418 on Build GRCh38
3666872 c.813T>G p.P271P Chr.11: 125582373-125582373 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772510307 Chr.11: 125582406-125582407 on Build GRCh38
rs940054973 Chr.11: 125582727-125582727 on Build GRCh38
rs916076566 Chr.11: 125582658-125582658 on Build GRCh38
rs957748630 Chr.11: 125582606-125582607 on Build GRCh38
rs200148421 Chr.11: 125582434-125582434 on Build GRCh38
rs148684485 Chr.11: 125582801-125582802 on Build GRCh38
rs898561678 Chr.11: 125582439-125582439 on Build GRCh38
rs995312606 Chr.11: 125582451-125582451 on Build GRCh38
rs768907071 Chr.11: 125582387-125582387 on Build GRCh38
rs542740303 Chr.11: 125582500-125582500 on Build GRCh38
rs541488011 Chr.11: 125582523-125582523 on Build GRCh38
rs868307053 Chr.11: 125582409-125582409 on Build GRCh38
rs754554338 Chr.11: 125582460-125582460 on Build GRCh38
rs762483784 Chr.11: 125582431-125582431 on Build GRCh38
rs4627097 Chr.11: 125582418-125582418 on Build GRCh38
rs551212807 Chr.11: 125582708-125582712 on Build GRCh38
rs959435951 Chr.11: 125582686-125582686 on Build GRCh38
rs1019732856 Chr.11: 125582482-125582482 on Build GRCh38
rs990317016 Chr.11: 125582612-125582612 on Build GRCh38
rs571041243 Chr.11: 125582599-125582599 on Build GRCh38
rs972394188 Chr.11: 125582416-125582416 on Build GRCh38
rs780577176 Chr.11: 125582700-125582700 on Build GRCh38
rs535423740 Chr.11: 125582488-125582488 on Build GRCh38
rs538655757 Chr.11: 125582604-125582604 on Build GRCh38
rs558287776 Chr.11: 125582704-125582704 on Build GRCh38
rs919621478 Chr.11: 125582807-125582807 on Build GRCh38
rs397812741 Chr.11: 125582408-125582409 on Build GRCh38
rs758879754 Chr.11: 125582627-125582627 on Build GRCh38
rs534388369 Chr.11: 125582424-125582424 on Build GRCh38
rs902113399 Chr.11: 125582499-125582499 on Build GRCh38
rs34775812 Chr.11: 125582828-125582828 on Build GRCh38
rs910989869 Chr.11: 125582822-125582822 on Build GRCh38
rs931358865 Chr.11: 125582803-125582803 on Build GRCh38
rs186567953 Chr.11: 125582749-125582749 on Build GRCh38
rs1028079665 Chr.11: 125582456-125582456 on Build GRCh38
rs972385931 Chr.11: 125582796-125582796 on Build GRCh38
rs201075869 Chr.11: 125582388-125582388 on Build GRCh38
rs910735404 Chr.11: 125582449-125582449 on Build GRCh38
rs562432902 Chr.11: 125582558-125582558 on Build GRCh38
rs961323344 Chr.11: 125582774-125582774 on Build GRCh38
rs903923125 Chr.11: 125582549-125582549 on Build GRCh38
rs762073143 Chr.11: 125582417-125582417 on Build GRCh38
rs548662841 Chr.11: 125582396-125582396 on Build GRCh38
rs1042410032 Chr.11: 125582528-125582528 on Build GRCh38
rs981703812 Chr.11: 125582703-125582703 on Build GRCh38
rs747494341 Chr.11: 125582772-125582772 on Build GRCh38
rs531417619 Chr.11: 125582499-125582500 on Build GRCh38
rs147164583 Chr.11: 125582812-125582812 on Build GRCh38
rs985590632 Chr.11: 125582818-125582818 on Build GRCh38
rs751230360 Chr.11: 125582457-125582457 on Build GRCh38
rs1032114782 Chr.11: 125582518-125582518 on Build GRCh38
rs765829350 Chr.11: 125582437-125582437 on Build GRCh38
rs767068810 Chr.11: 125582470-125582470 on Build GRCh38

View Full Product Details