Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 961735-962230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL17
Entrez Gene ID
339451
Gene Name
kelch like family member 17
Gene Aliases
-
UniGene
Hs.109212
Gene Chromosome Location
Chr.1: 960103-965719 on Build GRCh38
Transcripts
NM_198317.2 XM_006710600.3 XM_006710601.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6696568 c.538G>T p.A180S Chr.1: 961874-961874 on Build GRCh38
1492151 c.585_586GG>TT p.G196C Chr.1: 961921-961922 on Build GRCh38
302502 c.703C>A p.L235M Chr.1: 962039-962039 on Build GRCh38
5735196 c.568G>A p.D190N Chr.1: 961904-961904 on Build GRCh38
3361041 c.585G>T p.L195L Chr.1: 961921-961921 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141073078 Chr.1: 962060-962060 on Build GRCh38
rs572528335 Chr.1: 961761-961761 on Build GRCh38
rs576316712 Chr.1: 962074-962074 on Build GRCh38
rs752896220 Chr.1: 962063-962063 on Build GRCh38
rs764920249 Chr.1: 961931-961931 on Build GRCh38
rs761268341 Chr.1: 962038-962038 on Build GRCh38
rs779610967 Chr.1: 961824-961824 on Build GRCh38
rs755087830 Chr.1: 962059-962059 on Build GRCh38
rs781001873 Chr.1: 962016-962016 on Build GRCh38
rs781348880 Chr.1: 961904-961904 on Build GRCh38
rs771687125 Chr.1: 962079-962079 on Build GRCh38
rs767554091 Chr.1: 961817-961817 on Build GRCh38
rs767043015 Chr.1: 962085-962085 on Build GRCh38
rs774539979 Chr.1: 961791-961791 on Build GRCh38
rs747660114 Chr.1: 962174-962174 on Build GRCh38
rs368682182 Chr.1: 961956-961956 on Build GRCh38
rs150391309 Chr.1: 961950-961950 on Build GRCh38
rs374306425 Chr.1: 961790-961790 on Build GRCh38
rs778990282 Chr.1: 962005-962005 on Build GRCh38
rs150756317 Chr.1: 961871-961871 on Build GRCh38
rs746871827 Chr.1: 962011-962011 on Build GRCh38
rs775830816 Chr.1: 961907-961907 on Build GRCh38
rs766988185 Chr.1: 962047-962047 on Build GRCh38
rs751515985 Chr.1: 961766-961766 on Build GRCh38
rs371364043 Chr.1: 962176-962176 on Build GRCh38
rs763325166 Chr.1: 962193-962193 on Build GRCh38
rs776484757 Chr.1: 962175-962175 on Build GRCh38
rs941567586 Chr.1: 961837-961837 on Build GRCh38
rs759976649 Chr.1: 962139-962139 on Build GRCh38
rs765733602 Chr.1: 962000-962000 on Build GRCh38
rs761382592 Chr.1: 962077-962077 on Build GRCh38
rs773859170 Chr.1: 962076-962076 on Build GRCh38
rs750791656 Chr.1: 962114-962114 on Build GRCh38
rs758556605 Chr.1: 962100-962100 on Build GRCh38
rs762611286 Chr.1: 961985-961985 on Build GRCh38
rs765711602 Chr.1: 962140-962140 on Build GRCh38
rs773041226 Chr.1: 962075-962076 on Build GRCh38
rs372683639 Chr.1: 961828-961828 on Build GRCh38
rs762025110 Chr.1: 962201-962201 on Build GRCh38
rs377602573 Chr.1: 961961-961961 on Build GRCh38
rs373820131 Chr.1: 961929-961929 on Build GRCh38
rs748154276 Chr.1: 962073-962074 on Build GRCh38
rs769837786 Chr.1: 961863-961863 on Build GRCh38
rs770925041 Chr.1: 962144-962144 on Build GRCh38
rs770959542 Chr.1: 961970-961970 on Build GRCh38
rs1024915200 Chr.1: 961776-961776 on Build GRCh38
rs760674692 Chr.1: 961912-961912 on Build GRCh38
rs780280254 Chr.1: 961809-961810 on Build GRCh38
rs79271696 Chr.1: 961906-961906 on Build GRCh38
rs762455818 Chr.1: 962195-962196 on Build GRCh38
rs774935555 Chr.1: 961984-961984 on Build GRCh38
rs13303229 Chr.1: 962184-962184 on Build GRCh38
rs376949939 Chr.1: 961838-961838 on Build GRCh38
rs368211588 Chr.1: 961792-961792 on Build GRCh38
rs755867811 Chr.1: 962108-962108 on Build GRCh38
rs762700012 Chr.1: 961803-961803 on Build GRCh38
rs775104860 Chr.1: 962099-962100 on Build GRCh38
rs759829443 Chr.1: 961860-961860 on Build GRCh38
rs753120016 Chr.1: 961891-961891 on Build GRCh38
rs776287833 Chr.1: 962081-962082 on Build GRCh38
rs771298220 Chr.1: 961900-961900 on Build GRCh38
rs758172724 Chr.1: 961865-961865 on Build GRCh38
rs757418290 Chr.1: 962156-962156 on Build GRCh38
rs772022853 Chr.1: 961909-961909 on Build GRCh38
rs761107058 Chr.1: 962082-962082 on Build GRCh38
rs541095935 Chr.1: 961772-961772 on Build GRCh38
rs770603165 Chr.1: 962013-962013 on Build GRCh38
rs541134803 Chr.1: 962159-962159 on Build GRCh38
rs373943055 Chr.1: 962196-962196 on Build GRCh38
rs749662165 Chr.1: 961786-961786 on Build GRCh38
rs149416826 Chr.1: 961969-961969 on Build GRCh38
rs373185160 Chr.1: 961960-961960 on Build GRCh38
rs577713243 Chr.1: 962195-962195 on Build GRCh38
rs1020271774 Chr.1: 962206-962206 on Build GRCh38
rs768154140 Chr.1: 961777-961777 on Build GRCh38
rs751807287 Chr.1: 962146-962146 on Build GRCh38
rs747993667 Chr.1: 961908-961908 on Build GRCh38
rs747585285 Chr.1: 961899-961899 on Build GRCh38
rs759097083 Chr.1: 962094-962095 on Build GRCh38
rs916340930 Chr.1: 962112-962112 on Build GRCh38
rs776893739 Chr.1: 961802-961802 on Build GRCh38
rs752767694 Chr.1: 962092-962092 on Build GRCh38
rs753600798 Chr.1: 962163-962163 on Build GRCh38
rs548206227 Chr.1: 961928-961928 on Build GRCh38
rs776412372 Chr.1: 961972-961972 on Build GRCh38
rs879144111 Chr.1: 962080-962080 on Build GRCh38
rs1018375040 Chr.1: 961988-961988 on Build GRCh38
rs760049342 Chr.1: 962089-962089 on Build GRCh38
rs765936701 Chr.1: 961874-961874 on Build GRCh38
rs538369199 Chr.1: 962096-962101 on Build GRCh38
rs754030006 Chr.1: 962048-962048 on Build GRCh38
rs777142655 Chr.1: 962178-962178 on Build GRCh38
rs748432046 Chr.1: 961808-961808 on Build GRCh38
rs768085970 Chr.1: 962207-962207 on Build GRCh38
rs961254761 Chr.1: 961782-961782 on Build GRCh38
rs761484139 Chr.1: 962028-962028 on Build GRCh38
rs572314057 Chr.1: 962129-962129 on Build GRCh38
rs758740748 Chr.1: 961896-961896 on Build GRCh38
rs377056752 Chr.1: 961798-961798 on Build GRCh38
rs115985665 Chr.1: 961957-961957 on Build GRCh38
rs763985708 Chr.1: 961809-961809 on Build GRCh38
rs755624555 Chr.1: 961822-961822 on Build GRCh38
rs778896661 Chr.1: 962171-962171 on Build GRCh38
rs1056338646 Chr.1: 962105-962105 on Build GRCh38
rs112585496 Chr.1: 961818-961818 on Build GRCh38
rs984447124 Chr.1: 962191-962191 on Build GRCh38
rs752503574 Chr.1: 961762-961762 on Build GRCh38
rs146720000 Chr.1: 961919-961919 on Build GRCh38
rs746771831 Chr.1: 962076-962080 on Build GRCh38
rs757845370 Chr.1: 961948-961948 on Build GRCh38
rs751354441 Chr.1: 961812-961812 on Build GRCh38
rs753229154 Chr.1: 961875-961875 on Build GRCh38
rs994112590 Chr.1: 962143-962144 on Build GRCh38
rs998888660 Chr.1: 962135-962135 on Build GRCh38
rs769031275 Chr.1: 961789-961789 on Build GRCh38
rs758009322 Chr.1: 961940-961940 on Build GRCh38
rs771454741 Chr.1: 962133-962133 on Build GRCh38
rs778289416 Chr.1: 962115-962115 on Build GRCh38
rs776046757 Chr.1: 962190-962190 on Build GRCh38
rs4970441 Chr.1: 961945-961945 on Build GRCh38
rs941515377 Chr.1: 962054-962054 on Build GRCh38
rs555070541 Chr.1: 961927-961927 on Build GRCh38
rs1043246818 Chr.1: 961850-961850 on Build GRCh38
rs757224836 Chr.1: 961768-961768 on Build GRCh38
rs775195679 Chr.1: 962207-962207 on Build GRCh38
rs149866042 Chr.1: 961905-961905 on Build GRCh38
rs774988881 Chr.1: 962018-962018 on Build GRCh38
rs370709224 Chr.1: 961873-961873 on Build GRCh38
rs957195495 Chr.1: 961769-961769 on Build GRCh38
rs750922306 Chr.1: 961996-961996 on Build GRCh38
rs754996796 Chr.1: 962063-962076 on Build GRCh38
rs370568234 Chr.1: 961964-961964 on Build GRCh38
rs750280495 Chr.1: 962107-962107 on Build GRCh38
rs754936183 Chr.1: 962170-962170 on Build GRCh38
rs749244360 Chr.1: 962113-962113 on Build GRCh38
rs202222378 Chr.1: 961881-961881 on Build GRCh38
rs768401237 Chr.1: 962021-962021 on Build GRCh38
rs780866212 Chr.1: 962068-962068 on Build GRCh38
rs779940836 Chr.1: 962073-962073 on Build GRCh38
rs763990796 Chr.1: 961764-961764 on Build GRCh38
rs376633397 Chr.1: 962064-962064 on Build GRCh38
rs985552724 Chr.1: 961800-961800 on Build GRCh38
rs761842849 Chr.1: 961813-961813 on Build GRCh38
rs754367245 Chr.1: 962087-962087 on Build GRCh38
rs746242508 Chr.1: 962181-962181 on Build GRCh38
rs186126206 Chr.1: 961836-961836 on Build GRCh38
rs770492884 Chr.1: 962076-962085 on Build GRCh38
rs767609323 Chr.1: 961868-961868 on Build GRCh38
rs773284714 Chr.1: 961795-961795 on Build GRCh38
rs971643036 Chr.1: 962173-962173 on Build GRCh38
rs376251054 Chr.1: 961939-961939 on Build GRCh38
rs764760295 Chr.1: 961978-961978 on Build GRCh38
rs752015500 Chr.1: 961990-961990 on Build GRCh38
rs763674572 Chr.1: 961987-961987 on Build GRCh38
rs778532225 Chr.1: 961885-961885 on Build GRCh38
rs777334642 Chr.1: 961827-961828 on Build GRCh38
rs200189761 Chr.1: 961867-961867 on Build GRCh38
rs747108280 Chr.1: 961843-961843 on Build GRCh38
rs777089358 Chr.1: 962138-962138 on Build GRCh38
rs778954983 Chr.1: 962066-962067 on Build GRCh38
rs539689146 Chr.1: 962037-962037 on Build GRCh38
rs564354353 Chr.1: 961773-961773 on Build GRCh38
rs888334715 Chr.1: 962164-962164 on Build GRCh38
rs749312698 Chr.1: 962001-962001 on Build GRCh38
rs771014878 Chr.1: 961847-961847 on Build GRCh38
rs767721156 Chr.1: 962208-962208 on Build GRCh38
rs570662202 Chr.1: 961995-961995 on Build GRCh38
rs745830549 Chr.1: 962017-962017 on Build GRCh38
rs764335553 Chr.1: 962145-962145 on Build GRCh38
rs773090910 Chr.1: 961911-961911 on Build GRCh38
rs144788434 Chr.1: 961999-961999 on Build GRCh38
rs755318695 Chr.1: 962004-962004 on Build GRCh38
rs555753367 Chr.1: 962119-962119 on Build GRCh38
rs755786552 Chr.1: 961783-961783 on Build GRCh38
rs777554205 Chr.1: 961878-961878 on Build GRCh38
rs760901064 Chr.1: 961759-961759 on Build GRCh38
rs764178043 Chr.1: 962194-962194 on Build GRCh38
rs772976700 Chr.1: 962121-962121 on Build GRCh38
rs779741374 Chr.1: 961785-961785 on Build GRCh38
rs745599647 Chr.1: 962070-962070 on Build GRCh38
rs775503307 Chr.1: 962141-962141 on Build GRCh38
rs779781499 Chr.1: 962110-962110 on Build GRCh38
rs535365271 Chr.1: 962104-962104 on Build GRCh38
rs776536433 Chr.1: 961849-961849 on Build GRCh38
rs757536123 Chr.1: 961993-961993 on Build GRCh38
rs765256819 Chr.1: 962090-962090 on Build GRCh38
rs754538365 Chr.1: 961829-961829 on Build GRCh38
rs769793676 Chr.1: 962072-962072 on Build GRCh38
rs762450631 Chr.1: 962020-962020 on Build GRCh38
rs1012856809 Chr.1: 962149-962149 on Build GRCh38
rs771680353 Chr.1: 962120-962120 on Build GRCh38
rs910165348 Chr.1: 962052-962052 on Build GRCh38
rs759089843 Chr.1: 961973-961973 on Build GRCh38
rs767794160 Chr.1: 962030-962030 on Build GRCh38
rs749335838 Chr.1: 962049-962050 on Build GRCh38
rs370957996 Chr.1: 961834-961834 on Build GRCh38
rs1047309383 Chr.1: 961820-961820 on Build GRCh38
rs998697497 Chr.1: 961903-961903 on Build GRCh38

View Full Product Details