Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 963254-963752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL17
Entrez Gene ID
339451
Gene Name
kelch like family member 17
Gene Aliases
-
UniGene
Hs.109212
Gene Chromosome Location
Chr.1: 960103-965719 on Build GRCh38
Transcripts
NM_198317.2 XM_006710600.3 XM_006710601.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3977957 c.1212T>A p.A404A Chr.1: 963361-963361 on Build GRCh38
6453938 c.1233C>T p.P411P Chr.1: 963382-963382 on Build GRCh38
4995494 c.1258G>A p.V420M Chr.1: 963407-963407 on Build GRCh38
3985281 c.1219G>A p.E407K Chr.1: 963368-963368 on Build GRCh38
6567106 c.1350G>A p.L450L Chr.1: 963499-963499 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368223179 Chr.1: 963297-963297 on Build GRCh38
rs777044810 Chr.1: 963546-963546 on Build GRCh38
rs572603528 Chr.1: 963544-963544 on Build GRCh38
rs770047344 Chr.1: 963306-963306 on Build GRCh38
rs183082554 Chr.1: 963579-963579 on Build GRCh38
rs143920987 Chr.1: 963407-963407 on Build GRCh38
rs754557676 Chr.1: 963326-963326 on Build GRCh38
rs763016804 Chr.1: 963433-963433 on Build GRCh38
rs914329911 Chr.1: 963397-963397 on Build GRCh38
rs544458240 Chr.1: 963595-963595 on Build GRCh38
rs1022104728 Chr.1: 963404-963404 on Build GRCh38
rs545741601 Chr.1: 963716-963716 on Build GRCh38
rs376823140 Chr.1: 963547-963547 on Build GRCh38
rs567156376 Chr.1: 963723-963723 on Build GRCh38
rs201126895 Chr.1: 963403-963403 on Build GRCh38
rs762343181 Chr.1: 963408-963408 on Build GRCh38
rs369692068 Chr.1: 963520-963520 on Build GRCh38
rs567011450 Chr.1: 963621-963621 on Build GRCh38
rs759482935 Chr.1: 963364-963364 on Build GRCh38
rs896814687 Chr.1: 963717-963717 on Build GRCh38
rs190933350 Chr.1: 963554-963554 on Build GRCh38
rs944830572 Chr.1: 963512-963512 on Build GRCh38
rs1028350557 Chr.1: 963569-963569 on Build GRCh38
rs151038313 Chr.1: 963401-963401 on Build GRCh38
rs753447285 Chr.1: 963323-963323 on Build GRCh38
rs202077085 Chr.1: 963446-963446 on Build GRCh38
rs986683112 Chr.1: 963578-963578 on Build GRCh38
rs764225616 Chr.1: 963553-963553 on Build GRCh38
rs761461421 Chr.1: 963424-963424 on Build GRCh38
rs114880602 Chr.1: 963534-963534 on Build GRCh38
rs561134674 Chr.1: 963606-963606 on Build GRCh38
rs972281932 Chr.1: 963568-963568 on Build GRCh38
rs770306292 Chr.1: 963413-963413 on Build GRCh38
rs117269332 Chr.1: 963472-963472 on Build GRCh38
rs920776653 Chr.1: 963373-963373 on Build GRCh38
rs781197479 Chr.1: 963463-963463 on Build GRCh38
rs773852722 Chr.1: 963484-963484 on Build GRCh38
rs763618676 Chr.1: 963376-963376 on Build GRCh38
rs766153864 Chr.1: 963359-963359 on Build GRCh38
rs199888109 Chr.1: 963332-963332 on Build GRCh38
rs574461422 Chr.1: 963558-963558 on Build GRCh38
rs761950391 Chr.1: 963614-963614 on Build GRCh38
rs773102407 Chr.1: 963353-963353 on Build GRCh38
rs773776541 Chr.1: 963539-963539 on Build GRCh38
rs61743507 Chr.1: 963478-963478 on Build GRCh38
rs769889736 Chr.1: 963340-963340 on Build GRCh38
rs200489647 Chr.1: 963402-963402 on Build GRCh38
rs776019287 Chr.1: 963425-963425 on Build GRCh38
rs3813184 Chr.1: 963620-963620 on Build GRCh38
rs775587429 Chr.1: 963470-963470 on Build GRCh38
rs772708259 Chr.1: 963326-963341 on Build GRCh38
rs552833949 Chr.1: 963681-963681 on Build GRCh38
rs1013286830 Chr.1: 963695-963695 on Build GRCh38
rs369390543 Chr.1: 963383-963383 on Build GRCh38
rs144707534 Chr.1: 963365-963365 on Build GRCh38
rs138106672 Chr.1: 963489-963489 on Build GRCh38
rs200016628 Chr.1: 963337-963337 on Build GRCh38
rs747175868 Chr.1: 963524-963524 on Build GRCh38
rs952229196 Chr.1: 963577-963577 on Build GRCh38
rs762509334 Chr.1: 963275-963275 on Build GRCh38
rs755950369 Chr.1: 963280-963280 on Build GRCh38
rs761050303 Chr.1: 963496-963496 on Build GRCh38
rs768622403 Chr.1: 963537-963537 on Build GRCh38
rs767879373 Chr.1: 963426-963426 on Build GRCh38
rs377174000 Chr.1: 963299-963299 on Build GRCh38
rs780517667 Chr.1: 963438-963438 on Build GRCh38
rs779831344 Chr.1: 963530-963530 on Build GRCh38
rs907522800 Chr.1: 963729-963729 on Build GRCh38
rs776837077 Chr.1: 963303-963303 on Build GRCh38
rs759580735 Chr.1: 963304-963304 on Build GRCh38
rs542287402 Chr.1: 963290-963290 on Build GRCh38
rs138887470 Chr.1: 963348-963348 on Build GRCh38
rs903674267 Chr.1: 963543-963543 on Build GRCh38
rs372775936 Chr.1: 963331-963331 on Build GRCh38
rs750784198 Chr.1: 963430-963430 on Build GRCh38
rs551268560 Chr.1: 963329-963329 on Build GRCh38
rs751200941 Chr.1: 963422-963422 on Build GRCh38
rs140681689 Chr.1: 963450-963450 on Build GRCh38
rs368509581 Chr.1: 963313-963313 on Build GRCh38
rs1015331531 Chr.1: 963712-963712 on Build GRCh38
rs558979028 Chr.1: 963541-963541 on Build GRCh38
rs751999861 Chr.1: 963381-963381 on Build GRCh38
rs780175599 Chr.1: 963387-963387 on Build GRCh38
rs758570025 Chr.1: 963551-963551 on Build GRCh38
rs371486484 Chr.1: 963692-963692 on Build GRCh38
rs867193163 Chr.1: 963444-963444 on Build GRCh38
rs771522723 Chr.1: 963542-963542 on Build GRCh38
rs745623835 Chr.1: 963468-963468 on Build GRCh38
rs748334577 Chr.1: 963454-963454 on Build GRCh38
rs371775037 Chr.1: 963291-963291 on Build GRCh38
rs935528683 Chr.1: 963391-963391 on Build GRCh38
rs763244372 Chr.1: 963548-963548 on Build GRCh38
rs556181635 Chr.1: 963469-963469 on Build GRCh38
rs767126686 Chr.1: 963492-963492 on Build GRCh38
rs961068658 Chr.1: 963566-963566 on Build GRCh38
rs956515294 Chr.1: 963276-963276 on Build GRCh38
rs760258997 Chr.1: 963545-963545 on Build GRCh38
rs1028977778 Chr.1: 963499-963499 on Build GRCh38
rs778686903 Chr.1: 963294-963294 on Build GRCh38
rs752814994 Chr.1: 963521-963521 on Build GRCh38
rs886358911 Chr.1: 963575-963576 on Build GRCh38
rs369562577 Chr.1: 963317-963317 on Build GRCh38
rs768971932 Chr.1: 963394-963394 on Build GRCh38
rs367602312 Chr.1: 963275-963275 on Build GRCh38
rs778114781 Chr.1: 963522-963522 on Build GRCh38
rs201151830 Chr.1: 963314-963314 on Build GRCh38
rs542734356 Chr.1: 963582-963582 on Build GRCh38
rs937960821 Chr.1: 963688-963688 on Build GRCh38
rs559753801 Chr.1: 963592-963592 on Build GRCh38
rs930706281 Chr.1: 963377-963377 on Build GRCh38
rs200987171 Chr.1: 963322-963322 on Build GRCh38
rs148507945 Chr.1: 963382-963382 on Build GRCh38
rs573548988 Chr.1: 963513-963513 on Build GRCh38
rs192840610 Chr.1: 963630-963630 on Build GRCh38
rs113057758 Chr.1: 963349-963349 on Build GRCh38
rs776743794 Chr.1: 963360-963360 on Build GRCh38
rs564884548 Chr.1: 963333-963333 on Build GRCh38
rs555205753 Chr.1: 963693-963693 on Build GRCh38
rs752239466 Chr.1: 963367-963367 on Build GRCh38
rs754321508 Chr.1: 963445-963445 on Build GRCh38
rs760365107 Chr.1: 963355-963355 on Build GRCh38
rs763610879 Chr.1: 963409-963409 on Build GRCh38
rs375596188 Chr.1: 963493-963493 on Build GRCh38
rs752765535 Chr.1: 963550-963550 on Build GRCh38
rs766693373 Chr.1: 963497-963497 on Build GRCh38
rs749336528 Chr.1: 963393-963393 on Build GRCh38
rs772412429 Chr.1: 963457-963457 on Build GRCh38
rs569406064 Chr.1: 963685-963685 on Build GRCh38
rs113146153 Chr.1: 963525-963525 on Build GRCh38
rs758133453 Chr.1: 963372-963372 on Build GRCh38
rs769847093 Chr.1: 963405-963405 on Build GRCh38
rs756004059 Chr.1: 963527-963527 on Build GRCh38
rs187461410 Chr.1: 963613-963613 on Build GRCh38
rs771268378 Chr.1: 963325-963325 on Build GRCh38
rs142869836 Chr.1: 963475-963475 on Build GRCh38
rs868084410 Chr.1: 963663-963663 on Build GRCh38
rs150563961 Chr.1: 963495-963495 on Build GRCh38

View Full Product Details