Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1182639-1183147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTLL10
Entrez Gene ID
254173
Gene Name
tubulin tyrosine ligase like 10
Gene Aliases
TTLL5
UniGene
Hs.454835
Gene Chromosome Location
Chr.1: 1173709-1197936 on Build GRCh38
Transcripts
NM_001130045.1 NM_153254.2 XM_005244738.1 XM_011541177.2 XM_017000906.1 XM_017000907.1 XM_017000908.1 XM_017000909.1 XM_017000910.1 XM_017000911.1 XM_017000912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6527474 c.989A>C p.E330A Chr.1: 1182948-1182948 on Build GRCh38
3471172 c.836C>T p.P279L Chr.1: 1183014-1183014 on Build GRCh38
4934867 c.949T>C p.S317P Chr.1: 1182908-1182908 on Build GRCh38
4737301 c.1032C>T p.D344D Chr.1: 1182991-1182991 on Build GRCh38
5627158 c.927A>G p.I309M Chr.1: 1182886-1182886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772812931 Chr.1: 1183000-1183002 on Build GRCh38
rs750448128 Chr.1: 1182978-1182978 on Build GRCh38
rs556698095 Chr.1: 1182777-1182777 on Build GRCh38
rs769137130 Chr.1: 1183057-1183057 on Build GRCh38
rs138091862 Chr.1: 1182985-1182985 on Build GRCh38
rs773618340 Chr.1: 1183074-1183074 on Build GRCh38
rs190210386 Chr.1: 1182697-1182697 on Build GRCh38
rs140929937 Chr.1: 1182950-1182950 on Build GRCh38
rs772486594 Chr.1: 1183023-1183023 on Build GRCh38
rs752651634 Chr.1: 1183006-1183006 on Build GRCh38
rs959374200 Chr.1: 1182796-1182796 on Build GRCh38
rs994801486 Chr.1: 1182704-1182704 on Build GRCh38
rs546816658 Chr.1: 1182939-1182939 on Build GRCh38
rs750519272 Chr.1: 1182897-1182897 on Build GRCh38
rs776052443 Chr.1: 1182959-1182959 on Build GRCh38
rs770731314 Chr.1: 1182938-1182938 on Build GRCh38
rs61733845 Chr.1: 1182895-1182895 on Build GRCh38
rs149500516 Chr.1: 1183034-1183034 on Build GRCh38
rs575186968 Chr.1: 1182842-1182842 on Build GRCh38
rs906807331 Chr.1: 1182991-1182991 on Build GRCh38
rs564690970 Chr.1: 1182835-1182835 on Build GRCh38
rs1053930094 Chr.1: 1182914-1182914 on Build GRCh38
rs775929191 Chr.1: 1182868-1182868 on Build GRCh38
rs1036160564 Chr.1: 1182696-1182696 on Build GRCh38
rs552655548 Chr.1: 1183124-1183124 on Build GRCh38
rs748777928 Chr.1: 1183054-1183054 on Build GRCh38
rs1056580979 Chr.1: 1182662-1182662 on Build GRCh38
rs748724530 Chr.1: 1183065-1183065 on Build GRCh38
rs776709121 Chr.1: 1183085-1183085 on Build GRCh38
rs539223589 Chr.1: 1182718-1182718 on Build GRCh38
rs760072128 Chr.1: 1182864-1182864 on Build GRCh38
rs542384528 Chr.1: 1182791-1182791 on Build GRCh38
rs1037445794 Chr.1: 1183127-1183127 on Build GRCh38
rs779278930 Chr.1: 1183059-1183059 on Build GRCh38
rs747885373 Chr.1: 1182866-1182867 on Build GRCh38
rs1044419258 Chr.1: 1182813-1182813 on Build GRCh38
rs199544116 Chr.1: 1182871-1182871 on Build GRCh38
rs781004642 Chr.1: 1183012-1183012 on Build GRCh38
rs757104331 Chr.1: 1183043-1183043 on Build GRCh38
rs770367062 Chr.1: 1182958-1182958 on Build GRCh38
rs573566237 Chr.1: 1182778-1182778 on Build GRCh38
rs374332935 Chr.1: 1182862-1182862 on Build GRCh38
rs769064031 Chr.1: 1183016-1183016 on Build GRCh38
rs747983795 Chr.1: 1182837-1182837 on Build GRCh38
rs763435422 Chr.1: 1182960-1182960 on Build GRCh38
rs747186229 Chr.1: 1183080-1183080 on Build GRCh38
rs1049436351 Chr.1: 1182705-1182705 on Build GRCh38
rs975118879 Chr.1: 1182729-1182729 on Build GRCh38
rs777577506 Chr.1: 1183007-1183007 on Build GRCh38
rs572742200 Chr.1: 1182827-1182827 on Build GRCh38
rs376372471 Chr.1: 1182996-1182996 on Build GRCh38
rs371677298 Chr.1: 1182925-1182925 on Build GRCh38
rs779469524 Chr.1: 1183014-1183014 on Build GRCh38
rs939618520 Chr.1: 1183117-1183117 on Build GRCh38
rs371673936 Chr.1: 1182756-1182756 on Build GRCh38
rs566983549 Chr.1: 1182710-1182710 on Build GRCh38
rs71628944 Chr.1: 1183045-1183045 on Build GRCh38
rs1039285648 Chr.1: 1182749-1182749 on Build GRCh38
rs559152536 Chr.1: 1182739-1182739 on Build GRCh38
rs569439420 Chr.1: 1183032-1183032 on Build GRCh38
rs774424198 Chr.1: 1183099-1183099 on Build GRCh38
rs530154459 Chr.1: 1182688-1182688 on Build GRCh38
rs10907171 Chr.1: 1182832-1182832 on Build GRCh38
rs369266010 Chr.1: 1183037-1183037 on Build GRCh38
rs1056688904 Chr.1: 1183123-1183123 on Build GRCh38
rs770599987 Chr.1: 1183011-1183011 on Build GRCh38
rs548840586 Chr.1: 1183051-1183051 on Build GRCh38
rs1021162841 Chr.1: 1182758-1182758 on Build GRCh38
rs983275676 Chr.1: 1182707-1182707 on Build GRCh38
rs924058491 Chr.1: 1182772-1182772 on Build GRCh38
rs553028676 Chr.1: 1182810-1182810 on Build GRCh38
rs866630539 Chr.1: 1182957-1182957 on Build GRCh38
rs772397027 Chr.1: 1183066-1183066 on Build GRCh38
rs751801451 Chr.1: 1182873-1182873 on Build GRCh38
rs992410529 Chr.1: 1182981-1182981 on Build GRCh38
rs989567062 Chr.1: 1183067-1183067 on Build GRCh38
rs765878001 Chr.1: 1182865-1182865 on Build GRCh38
rs773422212 Chr.1: 1183024-1183024 on Build GRCh38
rs754953861 Chr.1: 1182836-1182836 on Build GRCh38
rs930504944 Chr.1: 1183022-1183022 on Build GRCh38
rs757457950 Chr.1: 1182887-1182887 on Build GRCh38
rs748096585 Chr.1: 1182910-1182910 on Build GRCh38
rs114587574 Chr.1: 1183035-1183035 on Build GRCh38
rs112838421 Chr.1: 1183111-1183111 on Build GRCh38
rs532600952 Chr.1: 1183019-1183019 on Build GRCh38
rs774624126 Chr.1: 1182967-1182967 on Build GRCh38
rs749189460 Chr.1: 1182833-1182833 on Build GRCh38
rs141341297 Chr.1: 1182977-1182977 on Build GRCh38
rs1053984109 Chr.1: 1182784-1182784 on Build GRCh38
rs950494106 Chr.1: 1182834-1182834 on Build GRCh38
rs181467899 Chr.1: 1182763-1182763 on Build GRCh38
rs764381242 Chr.1: 1182826-1182826 on Build GRCh38
rs1035448704 Chr.1: 1182689-1182689 on Build GRCh38
rs370878292 Chr.1: 1182750-1182750 on Build GRCh38
rs569571316 Chr.1: 1182751-1182751 on Build GRCh38
rs368616691 Chr.1: 1182906-1182906 on Build GRCh38
rs981370190 Chr.1: 1183110-1183110 on Build GRCh38
rs777472636 Chr.1: 1182924-1182924 on Build GRCh38
rs746709502 Chr.1: 1182843-1182843 on Build GRCh38
rs375676753 Chr.1: 1182898-1182898 on Build GRCh38
rs771578163 Chr.1: 1182984-1182985 on Build GRCh38
rs186578890 Chr.1: 1183020-1183020 on Build GRCh38
rs368642920 Chr.1: 1182870-1182870 on Build GRCh38
rs905531378 Chr.1: 1182824-1182824 on Build GRCh38
rs753887558 Chr.1: 1183033-1183033 on Build GRCh38
rs998868797 Chr.1: 1183061-1183061 on Build GRCh38
rs200505408 Chr.1: 1183095-1183095 on Build GRCh38
rs1007155817 Chr.1: 1182717-1182717 on Build GRCh38
rs147256311 Chr.1: 1182666-1182666 on Build GRCh38
rs752580428 Chr.1: 1182844-1182844 on Build GRCh38
rs572607586 Chr.1: 1183126-1183126 on Build GRCh38
rs771570382 Chr.1: 1182839-1182839 on Build GRCh38
rs146443945 Chr.1: 1182923-1182923 on Build GRCh38
rs746653227 Chr.1: 1182935-1182935 on Build GRCh38
rs371930110 Chr.1: 1183084-1183084 on Build GRCh38
rs766515271 Chr.1: 1182992-1182992 on Build GRCh38
rs201484894 Chr.1: 1183052-1183052 on Build GRCh38
rs879517367 Chr.1: 1182786-1182786 on Build GRCh38
rs1053169937 Chr.1: 1183073-1183073 on Build GRCh38

View Full Product Details