Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1234594-1235097 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GALT6
Entrez Gene ID
126792
Gene Name
beta-1,3-galactosyltransferase 6
Gene Aliases
EDSP2, SEMDJL1, beta3GalT6
UniGene
Hs.284284
Gene Chromosome Location
Chr.1: 1232249-1235041 on Build GRCh38
Transcripts
NM_080605.3

SNP Detail

SNP ID Chromosome Location
rs149066310 Chr.1: 1234904-1234904 on Build GRCh38
rs780606087 Chr.1: 1234848-1234848 on Build GRCh38
rs574225209 Chr.1: 1235027-1235027 on Build GRCh38
rs1027133948 Chr.1: 1234655-1234655 on Build GRCh38
rs117953951 Chr.1: 1234872-1234872 on Build GRCh38
rs972406433 Chr.1: 1234690-1234690 on Build GRCh38
rs993369240 Chr.1: 1234945-1234945 on Build GRCh38
rs946134583 Chr.1: 1234762-1234762 on Build GRCh38
rs773289595 Chr.1: 1235042-1235042 on Build GRCh38
rs1018488069 Chr.1: 1234680-1234680 on Build GRCh38
rs916504056 Chr.1: 1235038-1235038 on Build GRCh38
rs1018954473 Chr.1: 1234921-1234921 on Build GRCh38
rs967361696 Chr.1: 1234984-1234984 on Build GRCh38
rs748196490 Chr.1: 1234738-1234738 on Build GRCh38
rs964760358 Chr.1: 1235014-1235014 on Build GRCh38
rs147584042 Chr.1: 1234728-1234728 on Build GRCh38
rs1046042632 Chr.1: 1234843-1234843 on Build GRCh38
rs61353980 Chr.1: 1234768-1234768 on Build GRCh38
rs1046285908 Chr.1: 1235044-1235044 on Build GRCh38
rs1007023928 Chr.1: 1234905-1234905 on Build GRCh38
rs902865246 Chr.1: 1234619-1234619 on Build GRCh38
rs528705193 Chr.1: 1234632-1234632 on Build GRCh38
rs936940401 Chr.1: 1235050-1235050 on Build GRCh38
rs952124074 Chr.1: 1234734-1234734 on Build GRCh38
rs868318154 Chr.1: 1234974-1234974 on Build GRCh38
rs913335625 Chr.1: 1234748-1234748 on Build GRCh38
rs528644657 Chr.1: 1235034-1235034 on Build GRCh38
rs1048140102 Chr.1: 1235059-1235059 on Build GRCh38
rs1043012123 Chr.1: 1234830-1234830 on Build GRCh38
rs976213497 Chr.1: 1234809-1234809 on Build GRCh38
rs182976037 Chr.1: 1234858-1234858 on Build GRCh38
rs367678255 Chr.1: 1234788-1234788 on Build GRCh38
rs200607416 Chr.1: 1235061-1235061 on Build GRCh38
rs986155851 Chr.1: 1234678-1234678 on Build GRCh38
rs543807072 Chr.1: 1234992-1234992 on Build GRCh38
rs560033243 Chr.1: 1235032-1235032 on Build GRCh38
rs142047096 Chr.1: 1234927-1234927 on Build GRCh38
rs1017624439 Chr.1: 1234989-1234989 on Build GRCh38
rs552005567 Chr.1: 1234692-1234692 on Build GRCh38
rs894160021 Chr.1: 1235069-1235069 on Build GRCh38
rs958120799 Chr.1: 1235019-1235019 on Build GRCh38
rs1055045877 Chr.1: 1234624-1234624 on Build GRCh38
rs988815644 Chr.1: 1235057-1235057 on Build GRCh38
rs578161110 Chr.1: 1234951-1234951 on Build GRCh38
rs376136144 Chr.1: 1235033-1235033 on Build GRCh38
rs948452182 Chr.1: 1235070-1235070 on Build GRCh38
rs151147873 Chr.1: 1234928-1234928 on Build GRCh38
rs568962063 Chr.1: 1235016-1235016 on Build GRCh38
rs145265329 Chr.1: 1234637-1234637 on Build GRCh38
rs557659114 Chr.1: 1234650-1234650 on Build GRCh38
rs569896337 Chr.1: 1234769-1234769 on Build GRCh38
rs749535599 Chr.1: 1234673-1234673 on Build GRCh38
rs372447529 Chr.1: 1234812-1234812 on Build GRCh38
rs925522857 Chr.1: 1235049-1235049 on Build GRCh38
rs561880825 Chr.1: 1235046-1235046 on Build GRCh38
rs570244002 Chr.1: 1234717-1234717 on Build GRCh38
rs767525530 Chr.1: 1234835-1234835 on Build GRCh38
rs116449540 Chr.1: 1234625-1234625 on Build GRCh38
rs1013214869 Chr.1: 1234629-1234629 on Build GRCh38
rs551586720 Chr.1: 1235054-1235054 on Build GRCh38
rs952032145 Chr.1: 1234966-1234966 on Build GRCh38
rs935656878 Chr.1: 1234838-1234838 on Build GRCh38
rs896882896 Chr.1: 1234649-1234649 on Build GRCh38
rs1045439106 Chr.1: 1235075-1235075 on Build GRCh38
rs1033159909 Chr.1: 1234626-1234626 on Build GRCh38
rs985194731 Chr.1: 1234971-1234971 on Build GRCh38
rs971256147 Chr.1: 1234689-1234689 on Build GRCh38
rs1012629558 Chr.1: 1234873-1234873 on Build GRCh38
rs1055456120 Chr.1: 1234833-1234833 on Build GRCh38
rs993907246 Chr.1: 1234870-1234870 on Build GRCh38
rs774067103 Chr.1: 1234773-1234773 on Build GRCh38
rs998824943 Chr.1: 1234940-1234940 on Build GRCh38
rs956242187 Chr.1: 1235028-1235028 on Build GRCh38
rs370156207 Chr.1: 1235029-1235029 on Build GRCh38
rs1020324400 Chr.1: 1234976-1234976 on Build GRCh38
rs945624510 Chr.1: 1234827-1234827 on Build GRCh38
rs952868804 Chr.1: 1234656-1234656 on Build GRCh38

View Full Product Details