Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1598710-1599201 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNDC10
Entrez Gene ID
643988
Gene Name
fibronectin type III domain containing 10
Gene Aliases
C1orf233
UniGene
Hs.528816
Gene Chromosome Location
Chr.1: 1598008-1600096 on Build GRCh38
Transcripts
NM_001242659.1

SNP Detail

SNP ID Chromosome Location
rs968159209 Chr.1: 1598934-1598934 on Build GRCh38
rs767395515 Chr.1: 1599108-1599108 on Build GRCh38
rs750138802 Chr.1: 1599141-1599141 on Build GRCh38
rs918119526 Chr.1: 1599169-1599169 on Build GRCh38
rs994184303 Chr.1: 1598744-1598744 on Build GRCh38
rs889646006 Chr.1: 1598891-1598891 on Build GRCh38
rs1022197859 Chr.1: 1598916-1598916 on Build GRCh38
rs916442049 Chr.1: 1598897-1598897 on Build GRCh38
rs958017156 Chr.1: 1598865-1598865 on Build GRCh38
rs1043894201 Chr.1: 1598789-1598789 on Build GRCh38
rs538830535 Chr.1: 1599176-1599176 on Build GRCh38
rs117508919 Chr.1: 1599011-1599011 on Build GRCh38
rs891711405 Chr.1: 1599086-1599086 on Build GRCh38
rs556796187 Chr.1: 1598943-1598944 on Build GRCh38
rs1019561576 Chr.1: 1598913-1598913 on Build GRCh38
rs991048756 Chr.1: 1598896-1598896 on Build GRCh38
rs988202471 Chr.1: 1598778-1598778 on Build GRCh38
rs536510584 Chr.1: 1599046-1599046 on Build GRCh38
rs888688579 Chr.1: 1598991-1598991 on Build GRCh38
rs970515490 Chr.1: 1599115-1599115 on Build GRCh38
rs377019182 Chr.1: 1598953-1598953 on Build GRCh38
rs200342676 Chr.1: 1598811-1598812 on Build GRCh38
rs140360752 Chr.1: 1599014-1599014 on Build GRCh38
rs965997231 Chr.1: 1598917-1598917 on Build GRCh38
rs553155924 Chr.1: 1599152-1599152 on Build GRCh38
rs551551306 Chr.1: 1598818-1598818 on Build GRCh38
rs990730622 Chr.1: 1599105-1599105 on Build GRCh38
rs1032672884 Chr.1: 1598832-1598832 on Build GRCh38
rs1048429698 Chr.1: 1598981-1598981 on Build GRCh38
rs946842825 Chr.1: 1598788-1598788 on Build GRCh38
rs192696003 Chr.1: 1598912-1598912 on Build GRCh38
rs988150047 Chr.1: 1598927-1598927 on Build GRCh38
rs1030054419 Chr.1: 1598813-1598813 on Build GRCh38
rs998321420 Chr.1: 1599015-1599015 on Build GRCh38
rs528617250 Chr.1: 1598814-1598814 on Build GRCh38
rs898179582 Chr.1: 1599026-1599026 on Build GRCh38
rs761992683 Chr.1: 1599032-1599032 on Build GRCh38
rs1032038990 Chr.1: 1599068-1599068 on Build GRCh38
rs571699735 Chr.1: 1598830-1598830 on Build GRCh38
rs1008068191 Chr.1: 1599082-1599082 on Build GRCh38
rs373020728 Chr.1: 1598798-1598798 on Build GRCh38
rs999592332 Chr.1: 1599041-1599041 on Build GRCh38
rs1021410698 Chr.1: 1598931-1598931 on Build GRCh38
rs566698780 Chr.1: 1599160-1599160 on Build GRCh38
rs1007467470 Chr.1: 1598780-1598780 on Build GRCh38
rs549207649 Chr.1: 1598791-1598791 on Build GRCh38
rs764145472 Chr.1: 1598817-1598817 on Build GRCh38
rs997241964 Chr.1: 1598809-1598809 on Build GRCh38
rs1049088538 Chr.1: 1598751-1598751 on Build GRCh38
rs918672308 Chr.1: 1599150-1599150 on Build GRCh38
rs386627529 Chr.1: 1599168-1599169 on Build GRCh38
rs918683393 Chr.1: 1598944-1598944 on Build GRCh38
rs889605526 Chr.1: 1599081-1599081 on Build GRCh38
rs992293669 Chr.1: 1599168-1599168 on Build GRCh38
rs926795466 Chr.1: 1598947-1598947 on Build GRCh38
rs942993358 Chr.1: 1598992-1598992 on Build GRCh38
rs1036647770 Chr.1: 1599018-1599018 on Build GRCh38
rs1023571478 Chr.1: 1599112-1599112 on Build GRCh38
rs867304631 Chr.1: 1598743-1598743 on Build GRCh38
rs898252013 Chr.1: 1598819-1598819 on Build GRCh38
rs953985632 Chr.1: 1598731-1598731 on Build GRCh38
rs1034196354 Chr.1: 1599128-1599128 on Build GRCh38
rs888707984 Chr.1: 1598770-1598770 on Build GRCh38
rs560249189 Chr.1: 1598800-1598800 on Build GRCh38
rs1010198954 Chr.1: 1599101-1599101 on Build GRCh38
rs116277749 Chr.1: 1599016-1599016 on Build GRCh38
rs940274918 Chr.1: 1599173-1599173 on Build GRCh38
rs958178206 Chr.1: 1599074-1599074 on Build GRCh38
rs949218404 Chr.1: 1598909-1598909 on Build GRCh38

View Full Product Details