Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1626730-1627220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIB2
Entrez Gene ID
142678
Gene Name
mindbomb E3 ubiquitin protein ligase 2
Gene Aliases
ZZANK1, ZZZ5
UniGene
Hs.135805
Gene Chromosome Location
Chr.1: 1615188-1630610 on Build GRCh38
Transcripts
NM_001170686.1 NM_001170687.1 NM_001170688.1 NM_001170689.1 NM_080875.2 NR_033183.1 XM_006710372.1 XM_011540731.2 XM_011540736.2 XM_011540737.2 XM_011540741.2 XM_011540742.2 XM_017000349.1 XM_017000350.1 XM_017000351.1 XM_017000352.1 XM_017000353.1 XM_017000354.1 XM_017000355.1 XM_017000356.1 XM_017000357.1 XM_017000358.1 XM_017000359.1 XM_017000360.1 XM_017000361.1 XM_017000362.1 XM_017000363.1 XM_017000364.1 XM_017000365.1 XM_017000366.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1983225 c.1403G>A p.R468Q Chr.1: 1626988-1626988 on Build GRCh38
6030368 c.1596C>T p.S532S Chr.1: 1627084-1627084 on Build GRCh38
6321716 c.1395G>T p.E465D Chr.1: 1626980-1626980 on Build GRCh38
7251341 c.1668G>A p.A556A Chr.1: 1627156-1627156 on Build GRCh38
3930431 c.1497G>A p.A499A Chr.1: 1627156-1627156 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs943117674 Chr.1: 1626935-1626935 on Build GRCh38
rs746786514 Chr.1: 1626801-1626801 on Build GRCh38
rs775774891 Chr.1: 1627112-1627112 on Build GRCh38
rs757806716 Chr.1: 1627166-1627166 on Build GRCh38
rs746210044 Chr.1: 1627104-1627104 on Build GRCh38
rs753495989 Chr.1: 1627014-1627014 on Build GRCh38
rs745902686 Chr.1: 1626774-1626774 on Build GRCh38
rs771884695 Chr.1: 1626798-1626798 on Build GRCh38
rs767728996 Chr.1: 1626895-1626895 on Build GRCh38
rs747497684 Chr.1: 1627102-1627102 on Build GRCh38
rs147305689 Chr.1: 1627183-1627183 on Build GRCh38
rs952977646 Chr.1: 1627044-1627044 on Build GRCh38
rs778340867 Chr.1: 1627030-1627030 on Build GRCh38
rs776141141 Chr.1: 1626826-1626826 on Build GRCh38
rs771800762 Chr.1: 1626777-1626777 on Build GRCh38
rs766602333 Chr.1: 1626900-1626900 on Build GRCh38
rs780151717 Chr.1: 1627172-1627172 on Build GRCh38
rs757590718 Chr.1: 1626793-1626793 on Build GRCh38
rs992231678 Chr.1: 1626867-1626867 on Build GRCh38
rs374336653 Chr.1: 1626839-1626839 on Build GRCh38
rs750585131 Chr.1: 1626796-1626796 on Build GRCh38
rs763424169 Chr.1: 1626787-1626787 on Build GRCh38
rs377647692 Chr.1: 1627056-1627056 on Build GRCh38
rs751438966 Chr.1: 1626916-1626916 on Build GRCh38
rs1020858029 Chr.1: 1626844-1626844 on Build GRCh38
rs116614106 Chr.1: 1627150-1627150 on Build GRCh38
rs781654506 Chr.1: 1627167-1627167 on Build GRCh38
rs1042168881 Chr.1: 1627062-1627062 on Build GRCh38
rs757856017 Chr.1: 1626792-1626792 on Build GRCh38
rs760848171 Chr.1: 1626899-1626899 on Build GRCh38
rs756978924 Chr.1: 1626875-1626875 on Build GRCh38
rs753654765 Chr.1: 1627080-1627080 on Build GRCh38
rs11544452 Chr.1: 1627156-1627156 on Build GRCh38
rs986147768 Chr.1: 1627047-1627047 on Build GRCh38
rs182596783 Chr.1: 1626878-1626878 on Build GRCh38
rs773056464 Chr.1: 1627127-1627127 on Build GRCh38
rs370335685 Chr.1: 1626832-1626832 on Build GRCh38
rs753978799 Chr.1: 1626901-1626901 on Build GRCh38
rs757431037 Chr.1: 1626770-1626770 on Build GRCh38
rs781540255 Chr.1: 1626794-1626794 on Build GRCh38
rs768734622 Chr.1: 1627124-1627124 on Build GRCh38
rs375730682 Chr.1: 1626954-1626954 on Build GRCh38
rs945142667 Chr.1: 1627081-1627081 on Build GRCh38
rs888865113 Chr.1: 1626805-1626805 on Build GRCh38
rs766579165 Chr.1: 1626978-1626978 on Build GRCh38
rs766329475 Chr.1: 1626849-1626849 on Build GRCh38
rs775034527 Chr.1: 1627191-1627191 on Build GRCh38
rs757861473 Chr.1: 1627096-1627096 on Build GRCh38
rs976919227 Chr.1: 1627061-1627061 on Build GRCh38
rs772874602 Chr.1: 1626778-1626778 on Build GRCh38
rs192469390 Chr.1: 1626785-1626785 on Build GRCh38
rs765642355 Chr.1: 1626752-1626752 on Build GRCh38
rs368815922 Chr.1: 1626943-1626943 on Build GRCh38
rs747341119 Chr.1: 1627041-1627041 on Build GRCh38
rs888412873 Chr.1: 1626762-1626762 on Build GRCh38
rs374320129 Chr.1: 1626797-1626797 on Build GRCh38
rs752220483 Chr.1: 1626791-1626792 on Build GRCh38
rs542956464 Chr.1: 1626987-1626987 on Build GRCh38
rs750318431 Chr.1: 1627070-1627070 on Build GRCh38
rs951817188 Chr.1: 1627179-1627179 on Build GRCh38
rs143944834 Chr.1: 1627168-1627168 on Build GRCh38
rs751857535 Chr.1: 1626790-1626790 on Build GRCh38
rs537069361 Chr.1: 1626924-1626924 on Build GRCh38
rs369247456 Chr.1: 1626971-1626971 on Build GRCh38
rs778145398 Chr.1: 1626760-1626760 on Build GRCh38
rs748782576 Chr.1: 1626885-1626885 on Build GRCh38
rs138333872 Chr.1: 1626769-1626769 on Build GRCh38
rs774899672 Chr.1: 1626884-1626884 on Build GRCh38
rs769372591 Chr.1: 1626880-1626880 on Build GRCh38
rs771312259 Chr.1: 1627103-1627103 on Build GRCh38
rs528678824 Chr.1: 1626773-1626773 on Build GRCh38
rs777584906 Chr.1: 1626799-1626799 on Build GRCh38
rs903310022 Chr.1: 1626958-1626958 on Build GRCh38
rs755931314 Chr.1: 1627003-1627003 on Build GRCh38
rs372371160 Chr.1: 1626888-1626888 on Build GRCh38
rs201142750 Chr.1: 1627196-1627196 on Build GRCh38
rs761453337 Chr.1: 1626946-1626946 on Build GRCh38
rs750557291 Chr.1: 1626847-1626847 on Build GRCh38
rs1000715897 Chr.1: 1627135-1627135 on Build GRCh38
rs746870748 Chr.1: 1626874-1626874 on Build GRCh38
rs759623717 Chr.1: 1626982-1626982 on Build GRCh38
rs766237909 Chr.1: 1627074-1627074 on Build GRCh38
rs748629484 Chr.1: 1626963-1626963 on Build GRCh38
rs781688125 Chr.1: 1626809-1626815 on Build GRCh38
rs376776768 Chr.1: 1626989-1626989 on Build GRCh38
rs759403517 Chr.1: 1627149-1627149 on Build GRCh38
rs549282813 Chr.1: 1626846-1626846 on Build GRCh38
rs528885425 Chr.1: 1627178-1627178 on Build GRCh38
rs747673096 Chr.1: 1627024-1627024 on Build GRCh38
rs28510084 Chr.1: 1627057-1627057 on Build GRCh38
rs756622910 Chr.1: 1626818-1626818 on Build GRCh38
rs375958653 Chr.1: 1626909-1626909 on Build GRCh38
rs766397284 Chr.1: 1627137-1627137 on Build GRCh38
rs1033265861 Chr.1: 1626802-1626802 on Build GRCh38
rs773519482 Chr.1: 1626896-1626896 on Build GRCh38
rs758381540 Chr.1: 1626859-1626859 on Build GRCh38
rs543519928 Chr.1: 1627165-1627165 on Build GRCh38
rs752219521 Chr.1: 1627090-1627090 on Build GRCh38
rs1040299900 Chr.1: 1627026-1627026 on Build GRCh38
rs959728668 Chr.1: 1626772-1626772 on Build GRCh38
rs374415093 Chr.1: 1626977-1626977 on Build GRCh38
rs754731172 Chr.1: 1627084-1627084 on Build GRCh38
rs535775620 Chr.1: 1626833-1626833 on Build GRCh38
rs976912368 Chr.1: 1627192-1627192 on Build GRCh38
rs758684616 Chr.1: 1626755-1626755 on Build GRCh38
rs776255248 Chr.1: 1626784-1626784 on Build GRCh38
rs763626936 Chr.1: 1627194-1627194 on Build GRCh38
rs1000385433 Chr.1: 1626959-1626959 on Build GRCh38
rs763616930 Chr.1: 1627161-1627161 on Build GRCh38
rs754569876 Chr.1: 1627017-1627017 on Build GRCh38
rs771062308 Chr.1: 1627180-1627180 on Build GRCh38
rs765251943 Chr.1: 1626983-1626983 on Build GRCh38
rs775024898 Chr.1: 1626957-1626957 on Build GRCh38
rs779837113 Chr.1: 1626950-1626950 on Build GRCh38
rs776765127 Chr.1: 1626981-1626981 on Build GRCh38
rs933994195 Chr.1: 1627060-1627060 on Build GRCh38
rs770629597 Chr.1: 1626818-1626818 on Build GRCh38
rs1033649842 Chr.1: 1626969-1626970 on Build GRCh38
rs578020266 Chr.1: 1626889-1626889 on Build GRCh38
rs745462036 Chr.1: 1626879-1626879 on Build GRCh38
rs753078151 Chr.1: 1626786-1626786 on Build GRCh38
rs772513617 Chr.1: 1627025-1627025 on Build GRCh38
rs369689575 Chr.1: 1627018-1627018 on Build GRCh38
rs528290990 Chr.1: 1627193-1627193 on Build GRCh38
rs776087284 Chr.1: 1626830-1626830 on Build GRCh38
rs747983814 Chr.1: 1626776-1626776 on Build GRCh38
rs756754313 Chr.1: 1627197-1627197 on Build GRCh38
rs374305319 Chr.1: 1626972-1626972 on Build GRCh38
rs769342679 Chr.1: 1626887-1626887 on Build GRCh38
rs761692872 Chr.1: 1626988-1626988 on Build GRCh38
rs979691954 Chr.1: 1626765-1626765 on Build GRCh38
rs769127508 Chr.1: 1626828-1626828 on Build GRCh38
rs770059073 Chr.1: 1627107-1627107 on Build GRCh38
rs564164909 Chr.1: 1626779-1626779 on Build GRCh38
rs762266095 Chr.1: 1626842-1626842 on Build GRCh38
rs750239219 Chr.1: 1626996-1626996 on Build GRCh38
rs547225652 Chr.1: 1626775-1626775 on Build GRCh38
rs778699079 Chr.1: 1627085-1627085 on Build GRCh38
rs752278909 Chr.1: 1627173-1627173 on Build GRCh38
rs779868029 Chr.1: 1627011-1627011 on Build GRCh38
rs971253565 Chr.1: 1626754-1626754 on Build GRCh38
rs746402799 Chr.1: 1626817-1626818 on Build GRCh38
rs770541510 Chr.1: 1626780-1626780 on Build GRCh38
rs764022017 Chr.1: 1626910-1626910 on Build GRCh38
rs752338822 Chr.1: 1627155-1627155 on Build GRCh38
rs371407201 Chr.1: 1626951-1626951 on Build GRCh38
rs771268027 Chr.1: 1627043-1627043 on Build GRCh38
rs778154248 Chr.1: 1627101-1627101 on Build GRCh38
rs574649121 Chr.1: 1626788-1626788 on Build GRCh38
rs776629730 Chr.1: 1627147-1627147 on Build GRCh38
rs780809255 Chr.1: 1626945-1626945 on Build GRCh38
rs868488550 Chr.1: 1627119-1627119 on Build GRCh38
rs753162422 Chr.1: 1626753-1626753 on Build GRCh38
rs767420165 Chr.1: 1627067-1627067 on Build GRCh38
rs187253621 Chr.1: 1627190-1627190 on Build GRCh38
rs760707863 Chr.1: 1627132-1627132 on Build GRCh38
rs565775802 Chr.1: 1626851-1626851 on Build GRCh38
rs756244735 Chr.1: 1626848-1626848 on Build GRCh38
rs368048212 Chr.1: 1626827-1626827 on Build GRCh38
rs998215116 Chr.1: 1626956-1626956 on Build GRCh38
rs776859586 Chr.1: 1627052-1627052 on Build GRCh38
rs762262347 Chr.1: 1626791-1626791 on Build GRCh38
rs922983625 Chr.1: 1627071-1627071 on Build GRCh38
rs755551348 Chr.1: 1626751-1626751 on Build GRCh38
rs755767987 Chr.1: 1626947-1626947 on Build GRCh38

View Full Product Details