Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 134249076-134249579 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THYN1
Entrez Gene ID
29087
Gene Name
thymocyte nuclear protein 1
Gene Aliases
HSPC144, MDS012, MY105, THY28, THY28KD
UniGene
Hs.13645
Gene Chromosome Location
Chr.11: 134248278-134253401 on Build GRCh38
Transcripts
NM_001037304.1 NM_001037305.1 NM_014174.2 NM_199297.1 NM_199298.1 XM_005271527.3 XM_005271528.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6505222 c.388G>A p.V130M Chr.11: 134249259-134249259 on Build GRCh38
7197714 c.385-14_389del19 p.? Chr.11: 134249258-134249276 on Build GRCh38
5781270 c.480+9T>C p.? Chr.11: 134249158-134249158 on Build GRCh38
1229239 c.443A>C p.D148A Chr.11: 134249204-134249204 on Build GRCh38
3769249 c.419T>A p.F140Y Chr.11: 134249228-134249228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs921538393 Chr.11: 134249110-134249110 on Build GRCh38
rs574379640 Chr.11: 134249380-134249380 on Build GRCh38
rs760080424 Chr.11: 134249311-134249311 on Build GRCh38
rs755406528 Chr.11: 134249095-134249095 on Build GRCh38
rs770043651 Chr.11: 134249295-134249295 on Build GRCh38
rs960618778 Chr.11: 134249543-134249543 on Build GRCh38
rs773371953 Chr.11: 134249300-134249300 on Build GRCh38
rs143669769 Chr.11: 134249238-134249238 on Build GRCh38
rs201549226 Chr.11: 134249244-134249244 on Build GRCh38
rs774332980 Chr.11: 134249165-134249165 on Build GRCh38
rs975850401 Chr.11: 134249332-134249332 on Build GRCh38
rs1050722011 Chr.11: 134249317-134249317 on Build GRCh38
rs1038066192 Chr.11: 134249403-134249403 on Build GRCh38
rs770834034 Chr.11: 134249161-134249161 on Build GRCh38
rs765413239 Chr.11: 134249191-134249191 on Build GRCh38
rs755023212 Chr.11: 134249330-134249330 on Build GRCh38
rs1038116997 Chr.11: 134249150-134249150 on Build GRCh38
rs375806541 Chr.11: 134249142-134249142 on Build GRCh38
rs746922892 Chr.11: 134249413-134249413 on Build GRCh38
rs867101455 Chr.11: 134249356-134249356 on Build GRCh38
rs148055755 Chr.11: 134249260-134249260 on Build GRCh38
rs755774680 Chr.11: 134249458-134249458 on Build GRCh38
rs755765263 Chr.11: 134249239-134249239 on Build GRCh38
rs766510680 Chr.11: 134249310-134249310 on Build GRCh38
rs552853012 Chr.11: 134249214-134249214 on Build GRCh38
rs775169653 Chr.11: 134249171-134249171 on Build GRCh38
rs747362180 Chr.11: 134249112-134249112 on Build GRCh38
rs998932793 Chr.11: 134249272-134249272 on Build GRCh38
rs1013887548 Chr.11: 134249523-134249523 on Build GRCh38
rs1055175463 Chr.11: 134249479-134249479 on Build GRCh38
rs763274224 Chr.11: 134249309-134249309 on Build GRCh38
rs758606098 Chr.11: 134249200-134249200 on Build GRCh38
rs74953693 Chr.11: 134249293-134249293 on Build GRCh38
rs748820188 Chr.11: 134249245-134249245 on Build GRCh38
rs913662556 Chr.11: 134249270-134249274 on Build GRCh38
rs185577463 Chr.11: 134249202-134249202 on Build GRCh38
rs76175248 Chr.11: 134249157-134249157 on Build GRCh38
rs748470665 Chr.11: 134249137-134249138 on Build GRCh38
rs561173354 Chr.11: 134249302-134249302 on Build GRCh38
rs755558989 Chr.11: 134249218-134249218 on Build GRCh38
rs781696069 Chr.11: 134249138-134249138 on Build GRCh38
rs770359549 Chr.11: 134249256-134249256 on Build GRCh38
rs78493568 Chr.11: 134249136-134249136 on Build GRCh38
rs781668448 Chr.11: 134249097-134249097 on Build GRCh38
rs763346213 Chr.11: 134249231-134249232 on Build GRCh38
rs921289467 Chr.11: 134249354-134249354 on Build GRCh38
rs762054576 Chr.11: 134249187-134249187 on Build GRCh38
rs750490871 Chr.11: 134249193-134249193 on Build GRCh38
rs773725309 Chr.11: 134249226-134249227 on Build GRCh38
rs371259391 Chr.11: 134249271-134249271 on Build GRCh38
rs143788644 Chr.11: 134249395-134249395 on Build GRCh38
rs780751551 Chr.11: 134249109-134249109 on Build GRCh38
rs769358419 Chr.11: 134249147-134249147 on Build GRCh38
rs548062560 Chr.11: 134249521-134249521 on Build GRCh38
rs951958224 Chr.11: 134249162-134249163 on Build GRCh38
rs944756488 Chr.11: 134249372-134249372 on Build GRCh38
rs189130413 Chr.11: 134249378-134249378 on Build GRCh38
rs750496006 Chr.11: 134249103-134249103 on Build GRCh38
rs770128001 Chr.11: 134249216-134249217 on Build GRCh38
rs576658985 Chr.11: 134249169-134249169 on Build GRCh38
rs568052048 Chr.11: 134249113-134249113 on Build GRCh38
rs781575471 Chr.11: 134249232-134249232 on Build GRCh38
rs768730737 Chr.11: 134249097-134249098 on Build GRCh38
rs148110822 Chr.11: 134249414-134249414 on Build GRCh38
rs111688117 Chr.11: 134249466-134249466 on Build GRCh38
rs74834977 Chr.11: 134249124-134249124 on Build GRCh38
rs772025250 Chr.11: 134249275-134249275 on Build GRCh38
rs77562773 Chr.11: 134249153-134249153 on Build GRCh38
rs765312231 Chr.11: 134249101-134249101 on Build GRCh38
rs1051674057 Chr.11: 134249117-134249117 on Build GRCh38
rs181463829 Chr.11: 134249431-134249431 on Build GRCh38
rs758938429 Chr.11: 134249108-134249108 on Build GRCh38
rs776542958 Chr.11: 134249111-134249113 on Build GRCh38

View Full Product Details