Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 1028400-1028898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERC1
Entrez Gene ID
23085
Gene Name
ELKS/RAB6-interacting/CAST family member 1
Gene Aliases
Cast2, ELKS, ERC-1, RAB6IP2
UniGene
Hs.601216
Gene Chromosome Location
Chr.12: 990010-1495933 on Build GRCh38
Transcripts
NM_001301248.1 NM_178039.3 NM_178040.3 NR_027946.2 NR_027948.2 NR_027949.2 XM_011520941.2 XM_017019052.1 XM_017019053.1 XM_017019054.1 XM_017019055.1 XM_017019056.1 XM_017019057.1 XM_017019058.1 XM_017019059.1 XM_017019060.1 XM_017019061.1 XM_017019062.1 XM_017019063.1 XM_017019064.1 XM_017019065.1 XM_017019066.1 XM_017019067.1 XM_017019068.1 XM_017019069.1 XM_017019070.1 XM_017019071.1 XM_017019072.1 XM_017019073.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5633551 c.622A>G p.I208V Chr.12: 1028525-1028525 on Build GRCh38
7260636 c.509A>C p.K170T Chr.12: 1028412-1028412 on Build GRCh38
5429801 c.582G>A p.L194L Chr.12: 1028485-1028485 on Build GRCh38
5674080 c.560A>G p.K187R Chr.12: 1028463-1028463 on Build GRCh38
935426 c.561G>A p.K187K Chr.12: 1028464-1028464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891787584 Chr.12: 1028807-1028807 on Build GRCh38
rs756116359 Chr.12: 1028580-1028580 on Build GRCh38
rs4765690 Chr.12: 1028866-1028866 on Build GRCh38
rs750611588 Chr.12: 1028676-1028676 on Build GRCh38
rs778738170 Chr.12: 1028696-1028696 on Build GRCh38
rs560776616 Chr.12: 1028630-1028630 on Build GRCh38
rs776414325 Chr.12: 1028560-1028560 on Build GRCh38
rs138529974 Chr.12: 1028524-1028524 on Build GRCh38
rs141624941 Chr.12: 1028483-1028483 on Build GRCh38
rs1000665011 Chr.12: 1028465-1028465 on Build GRCh38
rs761486520 Chr.12: 1028454-1028454 on Build GRCh38
rs768042325 Chr.12: 1028617-1028617 on Build GRCh38
rs1012978614 Chr.12: 1028462-1028462 on Build GRCh38
rs746174200 Chr.12: 1028600-1028600 on Build GRCh38
rs1057402508 Chr.12: 1028768-1028768 on Build GRCh38
rs746480863 Chr.12: 1028548-1028548 on Build GRCh38
rs779824945 Chr.12: 1028611-1028611 on Build GRCh38
rs923302191 Chr.12: 1028739-1028739 on Build GRCh38
rs756664440 Chr.12: 1028482-1028482 on Build GRCh38
rs751768265 Chr.12: 1028477-1028477 on Build GRCh38
rs367850160 Chr.12: 1028583-1028583 on Build GRCh38
rs980284362 Chr.12: 1028713-1028713 on Build GRCh38
rs908969334 Chr.12: 1028680-1028680 on Build GRCh38
rs3748524 Chr.12: 1028665-1028665 on Build GRCh38
rs1031778175 Chr.12: 1028638-1028638 on Build GRCh38
rs1017448580 Chr.12: 1028511-1028511 on Build GRCh38
rs143021404 Chr.12: 1028562-1028562 on Build GRCh38
rs778101125 Chr.12: 1028495-1028495 on Build GRCh38
rs754717117 Chr.12: 1028667-1028667 on Build GRCh38
rs375602886 Chr.12: 1028564-1028564 on Build GRCh38
rs903658680 Chr.12: 1028461-1028461 on Build GRCh38
rs759770486 Chr.12: 1028573-1028573 on Build GRCh38
rs771781513 Chr.12: 1028518-1028518 on Build GRCh38
rs755117710 Chr.12: 1028479-1028479 on Build GRCh38
rs549420417 Chr.12: 1028767-1028767 on Build GRCh38
rs757540017 Chr.12: 1028503-1028503 on Build GRCh38
rs757704162 Chr.12: 1028586-1028586 on Build GRCh38
rs766676015 Chr.12: 1028455-1028455 on Build GRCh38
rs908345195 Chr.12: 1028782-1028782 on Build GRCh38
rs752803438 Chr.12: 1028579-1028579 on Build GRCh38
rs927467479 Chr.12: 1028721-1028721 on Build GRCh38
rs778762418 Chr.12: 1028507-1028507 on Build GRCh38
rs749701492 Chr.12: 1028501-1028501 on Build GRCh38
rs148016557 Chr.12: 1028428-1028428 on Build GRCh38
rs767676264 Chr.12: 1028575-1028575 on Build GRCh38
rs767590463 Chr.12: 1028480-1028480 on Build GRCh38
rs964865732 Chr.12: 1028693-1028693 on Build GRCh38
rs748620568 Chr.12: 1028851-1028852 on Build GRCh38
rs756572220 Chr.12: 1028538-1028538 on Build GRCh38
rs754283037 Chr.12: 1028584-1028584 on Build GRCh38
rs983566095 Chr.12: 1028663-1028663 on Build GRCh38
rs933854216 Chr.12: 1028758-1028759 on Build GRCh38
rs746581941 Chr.12: 1028616-1028616 on Build GRCh38
rs942147341 Chr.12: 1028689-1028689 on Build GRCh38
rs372323969 Chr.12: 1028450-1028450 on Build GRCh38
rs561230356 Chr.12: 1028833-1028833 on Build GRCh38
rs745507164 Chr.12: 1028508-1028508 on Build GRCh38
rs372854799 Chr.12: 1028559-1028559 on Build GRCh38
rs763892753 Chr.12: 1028448-1028448 on Build GRCh38
rs1047030883 Chr.12: 1028759-1028759 on Build GRCh38
rs770394260 Chr.12: 1028619-1028623 on Build GRCh38
rs946073373 Chr.12: 1028825-1028825 on Build GRCh38
rs756580033 Chr.12: 1028866-1028866 on Build GRCh38
rs117698549 Chr.12: 1028657-1028657 on Build GRCh38
rs938917580 Chr.12: 1028749-1028749 on Build GRCh38
rs990364156 Chr.12: 1028679-1028679 on Build GRCh38
rs772926711 Chr.12: 1028569-1028569 on Build GRCh38
rs775151618 Chr.12: 1028543-1028543 on Build GRCh38
rs758158876 Chr.12: 1028603-1028603 on Build GRCh38
rs1038229594 Chr.12: 1028869-1028869 on Build GRCh38

View Full Product Details