Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 1965379-1965885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCP1B
Entrez Gene ID
196513
Gene Name
decapping mRNA 1B
Gene Aliases
DCP1
UniGene
Hs.130934
Gene Chromosome Location
Chr.12: 1941591-2004535 on Build GRCh38
Transcripts
NM_152640.4 NR_135060.1 XM_011520927.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5027517 c.149A>G p.N50S Chr.12: 1965625-1965625 on Build GRCh38
2004506 c.510C>T p.D170D Chr.12: 1965570-1965570 on Build GRCh38
4205999 c.204C>T p.D68D Chr.12: 1965570-1965570 on Build GRCh38
6848990 c.188T>C p.L63P Chr.12: 1965586-1965586 on Build GRCh38
5027516 c.455A>G p.N152S Chr.12: 1965625-1965625 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374990195 Chr.12: 1965569-1965569 on Build GRCh38
rs773876733 Chr.12: 1965510-1965510 on Build GRCh38
rs752449451 Chr.12: 1965685-1965685 on Build GRCh38
rs748728615 Chr.12: 1965601-1965601 on Build GRCh38
rs999663207 Chr.12: 1965573-1965573 on Build GRCh38
rs1043341493 Chr.12: 1965718-1965718 on Build GRCh38
rs17854304 Chr.12: 1965639-1965639 on Build GRCh38
rs765896654 Chr.12: 1965552-1965552 on Build GRCh38
rs199521846 Chr.12: 1965513-1965513 on Build GRCh38
rs745499958 Chr.12: 1965636-1965636 on Build GRCh38
rs554954236 Chr.12: 1965745-1965745 on Build GRCh38
rs116094704 Chr.12: 1965717-1965717 on Build GRCh38
rs759909453 Chr.12: 1965646-1965646 on Build GRCh38
rs373737300 Chr.12: 1965638-1965638 on Build GRCh38
rs77248202 Chr.12: 1965707-1965707 on Build GRCh38
rs148356523 Chr.12: 1965656-1965656 on Build GRCh38
rs907578471 Chr.12: 1965714-1965714 on Build GRCh38
rs775317804 Chr.12: 1965741-1965741 on Build GRCh38
rs755994541 Chr.12: 1965592-1965592 on Build GRCh38
rs970004092 Chr.12: 1965614-1965614 on Build GRCh38
rs954354426 Chr.12: 1965671-1965671 on Build GRCh38
rs767747677 Chr.12: 1965678-1965678 on Build GRCh38
rs1046922517 Chr.12: 1965805-1965805 on Build GRCh38
rs777116091 Chr.12: 1965637-1965637 on Build GRCh38
rs371514448 Chr.12: 1965532-1965532 on Build GRCh38
rs745435659 Chr.12: 1965723-1965723 on Build GRCh38
rs1055027442 Chr.12: 1965523-1965523 on Build GRCh38
rs575223607 Chr.12: 1965832-1965832 on Build GRCh38
rs552788008 Chr.12: 1965798-1965798 on Build GRCh38
rs535844493 Chr.12: 1965424-1965424 on Build GRCh38
rs771578299 Chr.12: 1965729-1965729 on Build GRCh38
rs770577287 Chr.12: 1965507-1965507 on Build GRCh38
rs556386372 Chr.12: 1965588-1965588 on Build GRCh38
rs746997302 Chr.12: 1965740-1965743 on Build GRCh38
rs777719414 Chr.12: 1965600-1965600 on Build GRCh38
rs191847191 Chr.12: 1965415-1965415 on Build GRCh38
rs777112289 Chr.12: 1965801-1965801 on Build GRCh38
rs539950914 Chr.12: 1965658-1965658 on Build GRCh38
rs895134801 Chr.12: 1965529-1965529 on Build GRCh38
rs751744721 Chr.12: 1965585-1965585 on Build GRCh38
rs543785867 Chr.12: 1965749-1965749 on Build GRCh38
rs537730693 Chr.12: 1965504-1965504 on Build GRCh38
rs755906593 Chr.12: 1965688-1965688 on Build GRCh38
rs940533122 Chr.12: 1965435-1965435 on Build GRCh38
rs1013656314 Chr.12: 1965559-1965559 on Build GRCh38
rs774573624 Chr.12: 1965652-1965652 on Build GRCh38
rs1038025657 Chr.12: 1965443-1965443 on Build GRCh38
rs557690728 Chr.12: 1965578-1965578 on Build GRCh38
rs1013499231 Chr.12: 1965788-1965788 on Build GRCh38
rs372431121 Chr.12: 1965514-1965514 on Build GRCh38
rs570964218 Chr.12: 1965819-1965819 on Build GRCh38
rs574663522 Chr.12: 1965753-1965753 on Build GRCh38
rs890985539 Chr.12: 1965773-1965773 on Build GRCh38
rs531129662 Chr.12: 1965777-1965777 on Build GRCh38
rs374283465 Chr.12: 1965525-1965525 on Build GRCh38
rs368421095 Chr.12: 1965530-1965530 on Build GRCh38
rs35653089 Chr.12: 1965798-1965799 on Build GRCh38
rs546044828 Chr.12: 1965570-1965570 on Build GRCh38
rs777199782 Chr.12: 1965520-1965520 on Build GRCh38
rs770358025 Chr.12: 1965625-1965625 on Build GRCh38
rs116312871 Chr.12: 1965774-1965774 on Build GRCh38
rs773861780 Chr.12: 1965643-1965643 on Build GRCh38
rs142382421 Chr.12: 1965662-1965662 on Build GRCh38
rs773967226 Chr.12: 1965626-1965626 on Build GRCh38
rs749205798 Chr.12: 1965708-1965708 on Build GRCh38
rs747984575 Chr.12: 1965590-1965590 on Build GRCh38
rs766357928 Chr.12: 1965657-1965657 on Build GRCh38
rs757282883 Chr.12: 1965710-1965710 on Build GRCh38

View Full Product Details