Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 2920556-2921080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TULP3
Entrez Gene ID
7289
Gene Name
tubby like protein 3
Gene Aliases
TUBL3
UniGene
Hs.655333
Gene Chromosome Location
Chr.12: 2890867-2941140 on Build GRCh38
Transcripts
NM_001160408.1 NM_003324.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3460065 c.152C>T p.P51L Chr.12: 2920878-2920878 on Build GRCh38
4479270 c.230C>T p.P77L Chr.12: 2920899-2920899 on Build GRCh38
6814342 c.57G>A p.R19R Chr.12: 2920783-2920783 on Build GRCh38
7121847 c.186C>A p.V62V Chr.12: 2920912-2920912 on Build GRCh38
7259785 c.196+8delT p.? Chr.12: 2920930-2920930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779375609 Chr.12: 2920769-2920769 on Build GRCh38
rs780419258 Chr.12: 2920727-2920728 on Build GRCh38
rs752192533 Chr.12: 2920937-2920937 on Build GRCh38
rs759850323 Chr.12: 2920615-2920615 on Build GRCh38
rs180916982 Chr.12: 2920651-2920651 on Build GRCh38
rs539797999 Chr.12: 2920877-2920877 on Build GRCh38
rs772565838 Chr.12: 2920777-2920777 on Build GRCh38
rs747386968 Chr.12: 2920961-2920961 on Build GRCh38
rs999109516 Chr.12: 2920875-2920875 on Build GRCh38
rs146376218 Chr.12: 2920823-2920823 on Build GRCh38
rs939977693 Chr.12: 2920721-2920721 on Build GRCh38
rs540639970 Chr.12: 2920663-2920663 on Build GRCh38
rs1031804104 Chr.12: 2920881-2920881 on Build GRCh38
rs900686027 Chr.12: 2920984-2920984 on Build GRCh38
rs774229306 Chr.12: 2920926-2920926 on Build GRCh38
rs748211451 Chr.12: 2920966-2920966 on Build GRCh38
rs763792679 Chr.12: 2920947-2920947 on Build GRCh38
rs199601573 Chr.12: 2920953-2920953 on Build GRCh38
rs931118844 Chr.12: 2921041-2921041 on Build GRCh38
rs769997364 Chr.12: 2920915-2920915 on Build GRCh38
rs79398525 Chr.12: 2920758-2920758 on Build GRCh38
rs369808122 Chr.12: 2920837-2920837 on Build GRCh38
rs767580564 Chr.12: 2920934-2920934 on Build GRCh38
rs990228337 Chr.12: 2920888-2920888 on Build GRCh38
rs764551474 Chr.12: 2920997-2920997 on Build GRCh38
rs10128815 Chr.12: 2920697-2920697 on Build GRCh38
rs781712445 Chr.12: 2920965-2920965 on Build GRCh38
rs746242429 Chr.12: 2920856-2920856 on Build GRCh38
rs900716892 Chr.12: 2920619-2920619 on Build GRCh38
rs549126197 Chr.12: 2920798-2920798 on Build GRCh38
rs919745547 Chr.12: 2920998-2920998 on Build GRCh38
rs765604038 Chr.12: 2920719-2920719 on Build GRCh38
rs761573646 Chr.12: 2920718-2920718 on Build GRCh38
rs370858813 Chr.12: 2920909-2920909 on Build GRCh38
rs780967843 Chr.12: 2920749-2920749 on Build GRCh38
rs758758109 Chr.12: 2920949-2920949 on Build GRCh38
rs753484845 Chr.12: 2920948-2920948 on Build GRCh38
rs758297130 Chr.12: 2920738-2920738 on Build GRCh38
rs992579948 Chr.12: 2920763-2920763 on Build GRCh38
rs372007502 Chr.12: 2920714-2920714 on Build GRCh38
rs755410042 Chr.12: 2920963-2920963 on Build GRCh38
rs995236778 Chr.12: 2920790-2920790 on Build GRCh38
rs776803884 Chr.12: 2920799-2920799 on Build GRCh38
rs762722512 Chr.12: 2920830-2920830 on Build GRCh38
rs570541745 Chr.12: 2920920-2920920 on Build GRCh38
rs375307923 Chr.12: 2920736-2920736 on Build GRCh38
rs779858558 Chr.12: 2920742-2920742 on Build GRCh38
rs780590799 Chr.12: 2920873-2920873 on Build GRCh38
rs769598549 Chr.12: 2920764-2920764 on Build GRCh38
rs777297562 Chr.12: 2920853-2920853 on Build GRCh38
rs747838051 Chr.12: 2920757-2920757 on Build GRCh38
rs886554270 Chr.12: 2920611-2920611 on Build GRCh38
rs1047790824 Chr.12: 2920609-2920609 on Build GRCh38
rs550241450 Chr.12: 2920842-2920842 on Build GRCh38
rs764296834 Chr.12: 2920712-2920712 on Build GRCh38
rs971636883 Chr.12: 2920868-2920868 on Build GRCh38
rs996450983 Chr.12: 2920659-2920659 on Build GRCh38
rs955828215 Chr.12: 2920762-2920762 on Build GRCh38
rs373735109 Chr.12: 2920972-2920972 on Build GRCh38
rs751075342 Chr.12: 2920744-2920744 on Build GRCh38
rs139350068 Chr.12: 2920845-2920845 on Build GRCh38
rs775930305 Chr.12: 2920782-2920782 on Build GRCh38
rs761230656 Chr.12: 2920929-2920929 on Build GRCh38
rs930700763 Chr.12: 2920584-2920588 on Build GRCh38
rs746235539 Chr.12: 2920775-2920775 on Build GRCh38
rs529548720 Chr.12: 2920797-2920797 on Build GRCh38
rs755754398 Chr.12: 2920939-2920939 on Build GRCh38
rs765057866 Chr.12: 2920727-2920727 on Build GRCh38
rs373458913 Chr.12: 2920844-2920844 on Build GRCh38
rs1009064613 Chr.12: 2920613-2920613 on Build GRCh38
rs996770867 Chr.12: 2920985-2920985 on Build GRCh38
rs765458545 Chr.12: 2920819-2920819 on Build GRCh38
rs747555221 Chr.12: 2920877-2920877 on Build GRCh38
rs921248238 Chr.12: 2920608-2920608 on Build GRCh38
rs867816552 Chr.12: 2920695-2920695 on Build GRCh38
rs34569123 Chr.12: 2920757-2920757 on Build GRCh38
rs1040605495 Chr.12: 2920614-2920615 on Build GRCh38
rs761973948 Chr.12: 2920804-2920804 on Build GRCh38
rs200463632 Chr.12: 2920841-2920841 on Build GRCh38
rs990648120 Chr.12: 2920604-2920604 on Build GRCh38
rs369558550 Chr.12: 2920748-2920748 on Build GRCh38
rs921331249 Chr.12: 2920912-2920912 on Build GRCh38
rs202025723 Chr.12: 2920908-2920908 on Build GRCh38
rs932662785 Chr.12: 2920644-2920644 on Build GRCh38
rs773363701 Chr.12: 2920919-2920919 on Build GRCh38
rs376106577 Chr.12: 2920883-2920883 on Build GRCh38
rs546577846 Chr.12: 2921048-2921048 on Build GRCh38
rs999861969 Chr.12: 2921054-2921054 on Build GRCh38

View Full Product Details