Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4353344-4353839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TIGAR
Entrez Gene ID
57103
Gene Name
TP53 induced glycolysis regulatory phosphatase
Gene Aliases
C12orf5, FR2BP
UniGene
Hs.504545
Gene Chromosome Location
Chr.12: 4321193-4360028 on Build GRCh38
Transcripts
NM_020375.2

SNP Detail

SNP ID Chromosome Location
rs982677954 Chr.12: 4353761-4353761 on Build GRCh38
rs990803055 Chr.12: 4353608-4353608 on Build GRCh38
rs893485145 Chr.12: 4353709-4353709 on Build GRCh38
rs17700406 Chr.12: 4353432-4353432 on Build GRCh38
rs796383959 Chr.12: 4353514-4353515 on Build GRCh38
rs397960016 Chr.12: 4353513-4353514 on Build GRCh38
rs1053094927 Chr.12: 4353578-4353578 on Build GRCh38
rs144769894 Chr.12: 4353782-4353782 on Build GRCh38
rs757257306 Chr.12: 4353694-4353694 on Build GRCh38
rs769691958 Chr.12: 4353771-4353771 on Build GRCh38
rs768032613 Chr.12: 4353383-4353383 on Build GRCh38
rs957066473 Chr.12: 4353797-4353797 on Build GRCh38
rs58163835 Chr.12: 4353511-4353512 on Build GRCh38
rs183115585 Chr.12: 4353607-4353607 on Build GRCh38
rs932373328 Chr.12: 4353812-4353812 on Build GRCh38
rs201854938 Chr.12: 4353514-4353514 on Build GRCh38
rs1019786667 Chr.12: 4353433-4353433 on Build GRCh38
rs757476857 Chr.12: 4353811-4353811 on Build GRCh38
rs920614444 Chr.12: 4353613-4353613 on Build GRCh38
rs931190894 Chr.12: 4353632-4353632 on Build GRCh38
rs542407765 Chr.12: 4353529-4353529 on Build GRCh38
rs530873177 Chr.12: 4353573-4353573 on Build GRCh38
rs1021092510 Chr.12: 4353558-4353558 on Build GRCh38
rs900917775 Chr.12: 4353691-4353691 on Build GRCh38
rs928034160 Chr.12: 4353601-4353601 on Build GRCh38
rs530158984 Chr.12: 4353788-4353788 on Build GRCh38
rs539399297 Chr.12: 4353798-4353798 on Build GRCh38
rs553120145 Chr.12: 4353809-4353809 on Build GRCh38
rs939846539 Chr.12: 4353664-4353664 on Build GRCh38
rs953414732 Chr.12: 4353769-4353769 on Build GRCh38
rs1019552746 Chr.12: 4353473-4353473 on Build GRCh38
rs753074324 Chr.12: 4353401-4353401 on Build GRCh38
rs556966862 Chr.12: 4353791-4353791 on Build GRCh38
rs546798257 Chr.12: 4353789-4353789 on Build GRCh38
rs149407420 Chr.12: 4353687-4353687 on Build GRCh38
rs1048206958 Chr.12: 4353633-4353633 on Build GRCh38
rs386759756 Chr.12: 4353514-4353515 on Build GRCh38
rs965332833 Chr.12: 4353726-4353726 on Build GRCh38
rs995251355 Chr.12: 4353629-4353629 on Build GRCh38
rs192931781 Chr.12: 4353748-4353748 on Build GRCh38
rs951232632 Chr.12: 4353429-4353429 on Build GRCh38
rs989708776 Chr.12: 4353531-4353531 on Build GRCh38
rs979526862 Chr.12: 4353543-4353543 on Build GRCh38
rs60591544 Chr.12: 4353515-4353515 on Build GRCh38
rs777999378 Chr.12: 4353560-4353560 on Build GRCh38
rs188150911 Chr.12: 4353707-4353707 on Build GRCh38
rs138786908 Chr.12: 4353579-4353579 on Build GRCh38
rs1035220709 Chr.12: 4353764-4353764 on Build GRCh38
rs576913076 Chr.12: 4353451-4353451 on Build GRCh38
rs570357636 Chr.12: 4353795-4353795 on Build GRCh38
rs967286777 Chr.12: 4353563-4353563 on Build GRCh38
rs146638352 Chr.12: 4353811-4353812 on Build GRCh38
rs533308823 Chr.12: 4353658-4353658 on Build GRCh38
rs547963589 Chr.12: 4353749-4353749 on Build GRCh38
rs747260488 Chr.12: 4353693-4353693 on Build GRCh38
rs374376743 Chr.12: 4353740-4353740 on Build GRCh38
rs564318826 Chr.12: 4353647-4353647 on Build GRCh38
rs920562558 Chr.12: 4353783-4353783 on Build GRCh38
rs935544902 Chr.12: 4353576-4353576 on Build GRCh38
rs965198150 Chr.12: 4353455-4353455 on Build GRCh38
rs949041560 Chr.12: 4353612-4353612 on Build GRCh38
rs190971108 Chr.12: 4353518-4353518 on Build GRCh38
rs142462447 Chr.12: 4353385-4353385 on Build GRCh38
rs368890680 Chr.12: 4353645-4353645 on Build GRCh38
rs1026820812 Chr.12: 4353421-4353421 on Build GRCh38
rs573486573 Chr.12: 4353528-4353528 on Build GRCh38
rs544607990 Chr.12: 4353675-4353675 on Build GRCh38
rs904903834 Chr.12: 4353621-4353621 on Build GRCh38

View Full Product Details