Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4353908-4354428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TIGAR
Entrez Gene ID
57103
Gene Name
TP53 induced glycolysis regulatory phosphatase
Gene Aliases
C12orf5, FR2BP
UniGene
Hs.504545
Gene Chromosome Location
Chr.12: 4321193-4360028 on Build GRCh38
Transcripts
NM_020375.2

SNP Detail

SNP ID Chromosome Location
rs187761737 Chr.12: 4354125-4354125 on Build GRCh38
rs976709918 Chr.12: 4353978-4353978 on Build GRCh38
rs1033897755 Chr.12: 4354285-4354285 on Build GRCh38
rs566610951 Chr.12: 4354400-4354400 on Build GRCh38
rs1042425068 Chr.12: 4354051-4354051 on Build GRCh38
rs148167681 Chr.12: 4354120-4354120 on Build GRCh38
rs922405794 Chr.12: 4354000-4354000 on Build GRCh38
rs540204294 Chr.12: 4353974-4353974 on Build GRCh38
rs796436176 Chr.12: 4354340-4354340 on Build GRCh38
rs932455216 Chr.12: 4354372-4354372 on Build GRCh38
rs907892936 Chr.12: 4353932-4353932 on Build GRCh38
rs57273283 Chr.12: 4354234-4354234 on Build GRCh38
rs983845327 Chr.12: 4353931-4353931 on Build GRCh38
rs7962818 Chr.12: 4354038-4354038 on Build GRCh38
rs895439781 Chr.12: 4354158-4354158 on Build GRCh38
rs953996248 Chr.12: 4354308-4354308 on Build GRCh38
rs201483099 Chr.12: 4354358-4354359 on Build GRCh38
rs939712436 Chr.12: 4354359-4354359 on Build GRCh38
rs976512266 Chr.12: 4354368-4354368 on Build GRCh38
rs560243031 Chr.12: 4353990-4353990 on Build GRCh38
rs952944525 Chr.12: 4354301-4354301 on Build GRCh38
rs914983287 Chr.12: 4354037-4354037 on Build GRCh38
rs1012413598 Chr.12: 4354161-4354161 on Build GRCh38
rs976596906 Chr.12: 4354288-4354288 on Build GRCh38
rs922518570 Chr.12: 4354302-4354302 on Build GRCh38
rs961565499 Chr.12: 4353954-4353954 on Build GRCh38
rs144055031 Chr.12: 4354251-4354251 on Build GRCh38
rs1032345049 Chr.12: 4354145-4354145 on Build GRCh38
rs79946734 Chr.12: 4354332-4354332 on Build GRCh38
rs35875213 Chr.12: 4354368-4354368 on Build GRCh38
rs932487600 Chr.12: 4354009-4354009 on Build GRCh38
rs570693708 Chr.12: 4354250-4354250 on Build GRCh38
rs1041227876 Chr.12: 4354022-4354022 on Build GRCh38
rs538158555 Chr.12: 4354308-4354308 on Build GRCh38
rs77611907 Chr.12: 4353971-4353971 on Build GRCh38
rs1015310962 Chr.12: 4354231-4354231 on Build GRCh38
rs980949974 Chr.12: 4354252-4354252 on Build GRCh38
rs952209865 Chr.12: 4354168-4354168 on Build GRCh38
rs1049584971 Chr.12: 4354025-4354025 on Build GRCh38
rs1000493641 Chr.12: 4354398-4354411 on Build GRCh38
rs7297434 Chr.12: 4354163-4354163 on Build GRCh38
rs113179444 Chr.12: 4354130-4354130 on Build GRCh38
rs892499017 Chr.12: 4354157-4354157 on Build GRCh38
rs908249774 Chr.12: 4353940-4353940 on Build GRCh38
rs939785391 Chr.12: 4354018-4354018 on Build GRCh38
rs1004985838 Chr.12: 4354195-4354195 on Build GRCh38
rs900995018 Chr.12: 4354029-4354029 on Build GRCh38
rs902191123 Chr.12: 4354090-4354090 on Build GRCh38
rs922365461 Chr.12: 4354368-4354369 on Build GRCh38
rs985245832 Chr.12: 4354310-4354310 on Build GRCh38
rs1025508329 Chr.12: 4354177-4354177 on Build GRCh38
rs970867156 Chr.12: 4354222-4354222 on Build GRCh38
rs760301271 Chr.12: 4354137-4354137 on Build GRCh38
rs915096335 Chr.12: 4354324-4354324 on Build GRCh38

View Full Product Details