Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4911086-4911629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNA1
Entrez Gene ID
3736
Gene Name
potassium voltage-gated channel subfamily A member 1
Gene Aliases
AEMK, EA1, HBK1, HUK1, KV1.1, MBK1, MK1, RBK1
UniGene
Hs.416139
Gene Chromosome Location
Chr.12: 4909907-4918256 on Build GRCh38
Transcripts
NM_000217.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3398786 c.250C>T p.R84C Chr.12: 4911628-4911628 on Build GRCh38
7092400 c.247G>A p.D83N Chr.12: 4911625-4911625 on Build GRCh38
416218 c.162C>A p.T54T Chr.12: 4911540-4911540 on Build GRCh38
300881 c.205C>T p.R69C Chr.12: 4911583-4911583 on Build GRCh38
384251 c.14C>T p.S5F Chr.12: 4911392-4911392 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201504073 Chr.12: 4911438-4911438 on Build GRCh38
rs762038042 Chr.12: 4911383-4911383 on Build GRCh38
rs557615900 Chr.12: 4911281-4911281 on Build GRCh38
rs768682801 Chr.12: 4911369-4911369 on Build GRCh38
rs752293178 Chr.12: 4911418-4911418 on Build GRCh38
rs28388577 Chr.12: 4911284-4911284 on Build GRCh38
rs753446355 Chr.12: 4911363-4911363 on Build GRCh38
rs529968149 Chr.12: 4911397-4911397 on Build GRCh38
rs145042702 Chr.12: 4911546-4911546 on Build GRCh38
rs754549386 Chr.12: 4911394-4911394 on Build GRCh38
rs766476165 Chr.12: 4911396-4911396 on Build GRCh38
rs28666907 Chr.12: 4911283-4911283 on Build GRCh38
rs367921276 Chr.12: 4911431-4911431 on Build GRCh38
rs979789211 Chr.12: 4911132-4911132 on Build GRCh38
rs759476808 Chr.12: 4911496-4911496 on Build GRCh38
rs748311101 Chr.12: 4911594-4911594 on Build GRCh38
rs937284012 Chr.12: 4911513-4911513 on Build GRCh38
rs75906135 Chr.12: 4911350-4911350 on Build GRCh38
rs374515303 Chr.12: 4911567-4911567 on Build GRCh38
rs553150052 Chr.12: 4911303-4911303 on Build GRCh38
rs886042316 Chr.12: 4911522-4911522 on Build GRCh38
rs758976505 Chr.12: 4911315-4911315 on Build GRCh38
rs573930602 Chr.12: 4911255-4911255 on Build GRCh38
rs752344044 Chr.12: 4911358-4911358 on Build GRCh38
rs149959487 Chr.12: 4911514-4911514 on Build GRCh38
rs773128496 Chr.12: 4911390-4911390 on Build GRCh38
rs897263951 Chr.12: 4911602-4911602 on Build GRCh38
rs750848000 Chr.12: 4911423-4911423 on Build GRCh38
rs573191253 Chr.12: 4911307-4911307 on Build GRCh38
rs763846827 Chr.12: 4911534-4911534 on Build GRCh38
rs1043625587 Chr.12: 4911458-4911458 on Build GRCh38
rs904488099 Chr.12: 4911461-4911461 on Build GRCh38
rs139476802 Chr.12: 4911510-4911510 on Build GRCh38
rs1000246159 Chr.12: 4911275-4911275 on Build GRCh38
rs773463314 Chr.12: 4911358-4911358 on Build GRCh38
rs1024449916 Chr.12: 4911541-4911541 on Build GRCh38
rs904539145 Chr.12: 4911272-4911272 on Build GRCh38
rs925698547 Chr.12: 4911186-4911186 on Build GRCh38
rs35678382 Chr.12: 4911269-4911269 on Build GRCh38
rs375361885 Chr.12: 4911420-4911420 on Build GRCh38
rs1014419258 Chr.12: 4911540-4911540 on Build GRCh38
rs751176537 Chr.12: 4911329-4911329 on Build GRCh38
rs373645838 Chr.12: 4911454-4911454 on Build GRCh38
rs748174897 Chr.12: 4911462-4911462 on Build GRCh38
rs201149627 Chr.12: 4911366-4911366 on Build GRCh38
rs370288610 Chr.12: 4911537-4911537 on Build GRCh38
rs375647671 Chr.12: 4911471-4911471 on Build GRCh38
rs534276633 Chr.12: 4911222-4911222 on Build GRCh38
rs912944320 Chr.12: 4911214-4911214 on Build GRCh38
rs770814681 Chr.12: 4911486-4911486 on Build GRCh38
rs746212583 Chr.12: 4911362-4911362 on Build GRCh38
rs886049507 Chr.12: 4911149-4911149 on Build GRCh38
rs543514434 Chr.12: 4911227-4911227 on Build GRCh38
rs1050964510 Chr.12: 4911233-4911233 on Build GRCh38
rs766225126 Chr.12: 4911328-4911328 on Build GRCh38
rs754516764 Chr.12: 4911344-4911344 on Build GRCh38
rs1007499992 Chr.12: 4911267-4911267 on Build GRCh38
rs577510278 Chr.12: 4911289-4911289 on Build GRCh38
rs546357890 Chr.12: 4911293-4911293 on Build GRCh38
rs537723793 Chr.12: 4911277-4911277 on Build GRCh38
rs565679233 Chr.12: 4911208-4911208 on Build GRCh38
rs1063289 Chr.12: 4911456-4911456 on Build GRCh38
rs759152813 Chr.12: 4911407-4911407 on Build GRCh38
rs12817318 Chr.12: 4911245-4911245 on Build GRCh38
rs765802042 Chr.12: 4911290-4911290 on Build GRCh38
rs769483045 Chr.12: 4911384-4911384 on Build GRCh38
rs776861490 Chr.12: 4911379-4911379 on Build GRCh38
rs933828987 Chr.12: 4911217-4911217 on Build GRCh38
rs1049379872 Chr.12: 4911276-4911276 on Build GRCh38
rs891898566 Chr.12: 4911517-4911517 on Build GRCh38
rs777276806 Chr.12: 4911421-4911421 on Build GRCh38
rs1045641584 Chr.12: 4911263-4911263 on Build GRCh38
rs767073909 Chr.12: 4911417-4911417 on Build GRCh38
rs541828200 Chr.12: 4911361-4911361 on Build GRCh38
rs981121776 Chr.12: 4911495-4911495 on Build GRCh38
rs962697658 Chr.12: 4911395-4911395 on Build GRCh38
rs146308797 Chr.12: 4911492-4911492 on Build GRCh38
rs954199236 Chr.12: 4911124-4911124 on Build GRCh38
rs935779883 Chr.12: 4911195-4911195 on Build GRCh38
rs113587682 Chr.12: 4911144-4911144 on Build GRCh38
rs1024544676 Chr.12: 4911301-4911301 on Build GRCh38
rs747465523 Chr.12: 4911439-4911439 on Build GRCh38
rs941091631 Chr.12: 4911193-4911193 on Build GRCh38
rs752067203 Chr.12: 4911557-4911557 on Build GRCh38
rs773288048 Chr.12: 4911477-4911477 on Build GRCh38
rs1037207049 Chr.12: 4911194-4911194 on Build GRCh38
rs755064500 Chr.12: 4911452-4911452 on Build GRCh38
rs1014875835 Chr.12: 4911299-4911299 on Build GRCh38
rs749152092 Chr.12: 4911485-4911485 on Build GRCh38
rs757572774 Chr.12: 4911359-4911359 on Build GRCh38
rs552253837 Chr.12: 4911574-4911574 on Build GRCh38
rs886049508 Chr.12: 4911352-4911352 on Build GRCh38
rs543311674 Chr.12: 4911422-4911422 on Build GRCh38

View Full Product Details