Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6550617-6551124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFFO1
Entrez Gene ID
25900
Gene Name
intermediate filament family orphan 1
Gene Aliases
HOM-TES-103, IFFO
UniGene
Hs.15243
Gene Chromosome Location
Chr.12: 6539528-6556102 on Build GRCh38
Transcripts
NM_001039670.2 NM_001193457.1 NM_080730.4 NR_036467.1 XM_006718975.3 XM_006718976.3 XM_006718977.3 XM_006718981.3 XM_011520947.2 XM_011520948.2 XM_011520949.2 XM_011520952.2 XM_017019169.1 XM_017019170.1 XM_017019171.1 XM_017019172.1 XM_017019173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5470075 c.930+1G>A p.? Chr.12: 6550694-6550694 on Build GRCh38
111379 c.812_825del14 p.D271fs*9 Chr.12: 6550950-6550963 on Build GRCh38
5443634 c.850G>A p.D284N Chr.12: 6550775-6550775 on Build GRCh38
4147400 c.814C>T p.R272C Chr.12: 6550961-6550961 on Build GRCh38
1363714 c.898G>A p.E300K Chr.12: 6550727-6550727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755049146 Chr.12: 6550810-6550810 on Build GRCh38
rs1028368164 Chr.12: 6550706-6550706 on Build GRCh38
rs754175858 Chr.12: 6550661-6550661 on Build GRCh38
rs758729876 Chr.12: 6550786-6550786 on Build GRCh38
rs553407822 Chr.12: 6550719-6550719 on Build GRCh38
rs375866509 Chr.12: 6550960-6550960 on Build GRCh38
rs768497993 Chr.12: 6550703-6550703 on Build GRCh38
rs201471948 Chr.12: 6550961-6550961 on Build GRCh38
rs542587824 Chr.12: 6550773-6550773 on Build GRCh38
rs939977616 Chr.12: 6550913-6550913 on Build GRCh38
rs370112336 Chr.12: 6550739-6550739 on Build GRCh38
rs773709812 Chr.12: 6551036-6551036 on Build GRCh38
rs948570963 Chr.12: 6550877-6550877 on Build GRCh38
rs751851967 Chr.12: 6550796-6550796 on Build GRCh38
rs989006159 Chr.12: 6551041-6551041 on Build GRCh38
rs150862109 Chr.12: 6550984-6550984 on Build GRCh38
rs867564741 Chr.12: 6550866-6550866 on Build GRCh38
rs745527883 Chr.12: 6550927-6550927 on Build GRCh38
rs773206304 Chr.12: 6550672-6550672 on Build GRCh38
rs760154205 Chr.12: 6550931-6550931 on Build GRCh38
rs909717559 Chr.12: 6550895-6550895 on Build GRCh38
rs759145020 Chr.12: 6550645-6550645 on Build GRCh38
rs7312991 Chr.12: 6551053-6551053 on Build GRCh38
rs1048754608 Chr.12: 6550865-6550865 on Build GRCh38
rs763226796 Chr.12: 6550979-6550979 on Build GRCh38
rs1036877925 Chr.12: 6550916-6550916 on Build GRCh38
rs954361326 Chr.12: 6550695-6550695 on Build GRCh38
rs539508978 Chr.12: 6550671-6550671 on Build GRCh38
rs773208565 Chr.12: 6550722-6550722 on Build GRCh38
rs538555054 Chr.12: 6550872-6550872 on Build GRCh38
rs750778667 Chr.12: 6550782-6550782 on Build GRCh38
rs771782016 Chr.12: 6550929-6550929 on Build GRCh38
rs376062108 Chr.12: 6550687-6550687 on Build GRCh38
rs181390413 Chr.12: 6551070-6551070 on Build GRCh38
rs926153778 Chr.12: 6550718-6550718 on Build GRCh38
rs868631000 Chr.12: 6551089-6551089 on Build GRCh38
rs762241648 Chr.12: 6550775-6550775 on Build GRCh38
rs533053290 Chr.12: 6551048-6551048 on Build GRCh38
rs761803226 Chr.12: 6550871-6550871 on Build GRCh38
rs767057674 Chr.12: 6551047-6551047 on Build GRCh38
rs570940366 Chr.12: 6550655-6550655 on Build GRCh38
rs765374647 Chr.12: 6550924-6550925 on Build GRCh38
rs777722429 Chr.12: 6551011-6551011 on Build GRCh38
rs763372317 Chr.12: 6551037-6551037 on Build GRCh38
rs768968893 Chr.12: 6550704-6550704 on Build GRCh38
rs937944577 Chr.12: 6550834-6550834 on Build GRCh38
rs767906387 Chr.12: 6550983-6550983 on Build GRCh38
rs773939106 Chr.12: 6550912-6550912 on Build GRCh38
rs771692285 Chr.12: 6551025-6551025 on Build GRCh38
rs754265635 Chr.12: 6550774-6550774 on Build GRCh38
rs371399810 Chr.12: 6550934-6550934 on Build GRCh38
rs778677350 Chr.12: 6551021-6551021 on Build GRCh38
rs112892336 Chr.12: 6550811-6550811 on Build GRCh38
rs756184110 Chr.12: 6550997-6550997 on Build GRCh38
rs747266024 Chr.12: 6550694-6550694 on Build GRCh38
rs764277123 Chr.12: 6550758-6550760 on Build GRCh38
rs749200957 Chr.12: 6551012-6551012 on Build GRCh38
rs773814755 Chr.12: 6550750-6550750 on Build GRCh38
rs921257711 Chr.12: 6550657-6550657 on Build GRCh38
rs372490626 Chr.12: 6550828-6550828 on Build GRCh38
rs749075467 Chr.12: 6550899-6550899 on Build GRCh38
rs1015368932 Chr.12: 6550894-6550895 on Build GRCh38
rs867673669 Chr.12: 6551056-6551056 on Build GRCh38
rs762745121 Chr.12: 6550726-6550726 on Build GRCh38
rs751763714 Chr.12: 6550978-6550978 on Build GRCh38
rs7301426 Chr.12: 6551059-6551059 on Build GRCh38
rs768072966 Chr.12: 6551031-6551031 on Build GRCh38
rs200021779 Chr.12: 6550930-6550930 on Build GRCh38
rs746487859 Chr.12: 6551030-6551030 on Build GRCh38
rs369782446 Chr.12: 6551022-6551022 on Build GRCh38
rs368335004 Chr.12: 6550814-6550814 on Build GRCh38
rs770658500 Chr.12: 6550906-6550906 on Build GRCh38
rs774840970 Chr.12: 6551049-6551049 on Build GRCh38
rs368129065 Chr.12: 6550890-6550890 on Build GRCh38
rs769690036 Chr.12: 6550721-6550721 on Build GRCh38
rs766583734 Chr.12: 6550790-6550790 on Build GRCh38
rs775164551 Chr.12: 6551028-6551028 on Build GRCh38
rs372763837 Chr.12: 6550682-6550682 on Build GRCh38
rs781183172 Chr.12: 6550710-6550710 on Build GRCh38
rs759231295 Chr.12: 6550761-6550761 on Build GRCh38
rs560024002 Chr.12: 6550869-6550869 on Build GRCh38
rs979052412 Chr.12: 6551000-6551000 on Build GRCh38
rs776673041 Chr.12: 6550783-6550783 on Build GRCh38
rs757014162 Chr.12: 6551015-6551015 on Build GRCh38
rs978336247 Chr.12: 6550823-6550823 on Build GRCh38
rs750796306 Chr.12: 6551047-6551062 on Build GRCh38
rs762086963 Chr.12: 6550794-6550794 on Build GRCh38
rs376803395 Chr.12: 6550829-6550829 on Build GRCh38
rs765460399 Chr.12: 6550779-6550779 on Build GRCh38
rs867397656 Chr.12: 6550681-6550681 on Build GRCh38

View Full Product Details