Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6769405-6769913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTMS
Entrez Gene ID
5763
Gene Name
parathymosin
Gene Aliases
ParaT
UniGene
Hs.504613
Gene Chromosome Location
Chr.12: 6766139-6770952 on Build GRCh38
Transcripts
NM_002824.4 XM_011520987.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2099141 c.117G>T p.E39D Chr.12: 6769674-6769674 on Build GRCh38
942872 c.60G>A p.K20K Chr.12: 6769617-6769617 on Build GRCh38
3812877 c.71T>G p.V24G Chr.12: 6769628-6769628 on Build GRCh38
3739849 c.62A>G p.K21R Chr.12: 6769619-6769619 on Build GRCh38
5832117 c.71T>G p.V24G Chr.12: 6769628-6769628 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368939007 Chr.12: 6769605-6769605 on Build GRCh38
rs1025342196 Chr.12: 6769650-6769650 on Build GRCh38
rs973805007 Chr.12: 6769613-6769615 on Build GRCh38
rs771690069 Chr.12: 6769607-6769607 on Build GRCh38
rs755762459 Chr.12: 6769817-6769817 on Build GRCh38
rs999346260 Chr.12: 6769464-6769464 on Build GRCh38
rs202102069 Chr.12: 6769557-6769557 on Build GRCh38
rs372653257 Chr.12: 6769655-6769655 on Build GRCh38
rs1034873577 Chr.12: 6769471-6769471 on Build GRCh38
rs111982897 Chr.12: 6769767-6769767 on Build GRCh38
rs756954035 Chr.12: 6769585-6769585 on Build GRCh38
rs751124604 Chr.12: 6769583-6769584 on Build GRCh38
rs748363383 Chr.12: 6769724-6769724 on Build GRCh38
rs757188367 Chr.12: 6769718-6769718 on Build GRCh38
rs542451468 Chr.12: 6769669-6769669 on Build GRCh38
rs777780138 Chr.12: 6769831-6769831 on Build GRCh38
rs767999155 Chr.12: 6769814-6769814 on Build GRCh38
rs1034706368 Chr.12: 6769454-6769454 on Build GRCh38
rs572553716 Chr.12: 6769834-6769834 on Build GRCh38
rs374959763 Chr.12: 6769588-6769588 on Build GRCh38
rs914865849 Chr.12: 6769732-6769732 on Build GRCh38
rs538826575 Chr.12: 6769801-6769801 on Build GRCh38
rs1001644715 Chr.12: 6769798-6769798 on Build GRCh38
rs898411763 Chr.12: 6769883-6769883 on Build GRCh38
rs776748371 Chr.12: 6769887-6769887 on Build GRCh38
rs777507246 Chr.12: 6769558-6769558 on Build GRCh38
rs182171171 Chr.12: 6769428-6769428 on Build GRCh38
rs552426374 Chr.12: 6769460-6769460 on Build GRCh38
rs112569458 Chr.12: 6769600-6769600 on Build GRCh38
rs1045528439 Chr.12: 6769775-6769775 on Build GRCh38
rs759914872 Chr.12: 6769876-6769876 on Build GRCh38
rs1012550650 Chr.12: 6769482-6769482 on Build GRCh38
rs780802546 Chr.12: 6769669-6769676 on Build GRCh38
rs772591681 Chr.12: 6769758-6769758 on Build GRCh38
rs745767749 Chr.12: 6769725-6769725 on Build GRCh38
rs746674425 Chr.12: 6769808-6769808 on Build GRCh38
rs768213418 Chr.12: 6769646-6769646 on Build GRCh38
rs986913722 Chr.12: 6769668-6769668 on Build GRCh38
rs942719342 Chr.12: 6769628-6769628 on Build GRCh38
rs555110018 Chr.12: 6769716-6769716 on Build GRCh38
rs780781097 Chr.12: 6769820-6769820 on Build GRCh38
rs73053705 Chr.12: 6769503-6769503 on Build GRCh38
rs372087097 Chr.12: 6769878-6769878 on Build GRCh38
rs534649050 Chr.12: 6769723-6769723 on Build GRCh38
rs764263216 Chr.12: 6769880-6769880 on Build GRCh38
rs144752841 Chr.12: 6769776-6769776 on Build GRCh38
rs771458445 Chr.12: 6769837-6769837 on Build GRCh38
rs1041768083 Chr.12: 6769770-6769770 on Build GRCh38
rs772579493 Chr.12: 6769877-6769877 on Build GRCh38
rs775067851 Chr.12: 6769870-6769870 on Build GRCh38
rs567965771 Chr.12: 6769686-6769686 on Build GRCh38
rs764256982 Chr.12: 6769708-6769708 on Build GRCh38
rs867073491 Chr.12: 6769645-6769645 on Build GRCh38
rs891520304 Chr.12: 6769734-6769734 on Build GRCh38
rs779032083 Chr.12: 6769677-6769677 on Build GRCh38
rs749023407 Chr.12: 6769830-6769831 on Build GRCh38
rs973919092 Chr.12: 6769486-6769486 on Build GRCh38
rs753828000 Chr.12: 6769567-6769567 on Build GRCh38
rs1001438853 Chr.12: 6769437-6769437 on Build GRCh38
rs774838869 Chr.12: 6769680-6769680 on Build GRCh38
rs760614592 Chr.12: 6769699-6769699 on Build GRCh38
rs576112852 Chr.12: 6769659-6769659 on Build GRCh38
rs754481529 Chr.12: 6769666-6769668 on Build GRCh38
rs746514929 Chr.12: 6769637-6769637 on Build GRCh38
rs896337811 Chr.12: 6769473-6769473 on Build GRCh38
rs896275391 Chr.12: 6769830-6769830 on Build GRCh38
rs912582474 Chr.12: 6769593-6769593 on Build GRCh38
rs573363956 Chr.12: 6769806-6769806 on Build GRCh38
rs752813922 Chr.12: 6769555-6769555 on Build GRCh38
rs971695364 Chr.12: 6769434-6769434 on Build GRCh38
rs149379625 Chr.12: 6769446-6769446 on Build GRCh38
rs767851017 Chr.12: 6769685-6769685 on Build GRCh38
rs779738380 Chr.12: 6769608-6769608 on Build GRCh38
rs938723540 Chr.12: 6769782-6769782 on Build GRCh38
rs779989386 Chr.12: 6769779-6769779 on Build GRCh38
rs752415138 Chr.12: 6769690-6769691 on Build GRCh38
rs377468902 Chr.12: 6769682-6769682 on Build GRCh38

View Full Product Details