Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7470620-7471127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD163
Entrez Gene ID
9332
Gene Name
CD163 molecule
Gene Aliases
M130, MM130, SCARI1
UniGene
Hs.504641
Gene Chromosome Location
Chr.12: 7470816-7503818 on Build GRCh38
Transcripts
NM_004244.5 NM_203416.3 XM_005253528.4 XM_005253529.4

SNP Detail

SNP ID Chromosome Location
rs374210755 Chr.12: 7470736-7470736 on Build GRCh38
rs983475867 Chr.12: 7471076-7471076 on Build GRCh38
rs1045550780 Chr.12: 7471062-7471062 on Build GRCh38
rs867585097 Chr.12: 7470834-7470834 on Build GRCh38
rs932467870 Chr.12: 7470852-7470852 on Build GRCh38
rs950716364 Chr.12: 7470792-7470792 on Build GRCh38
rs145776905 Chr.12: 7470804-7470804 on Build GRCh38
rs1027923765 Chr.12: 7471046-7471046 on Build GRCh38
rs1033399072 Chr.12: 7470964-7470964 on Build GRCh38
rs138294305 Chr.12: 7470891-7470891 on Build GRCh38
rs1040923879 Chr.12: 7470836-7470836 on Build GRCh38
rs181880710 Chr.12: 7470955-7470955 on Build GRCh38
rs893695940 Chr.12: 7470860-7470860 on Build GRCh38
rs563227835 Chr.12: 7470945-7470945 on Build GRCh38
rs994994889 Chr.12: 7471008-7471008 on Build GRCh38
rs996063150 Chr.12: 7471095-7471095 on Build GRCh38
rs920809744 Chr.12: 7470776-7470776 on Build GRCh38
rs951655179 Chr.12: 7470890-7470890 on Build GRCh38
rs781076546 Chr.12: 7470947-7470947 on Build GRCh38
rs113590996 Chr.12: 7470817-7470817 on Build GRCh38
rs73043456 Chr.12: 7470670-7470670 on Build GRCh38
rs773062606 Chr.12: 7470825-7470825 on Build GRCh38
rs935303414 Chr.12: 7470715-7470715 on Build GRCh38
rs186899245 Chr.12: 7471044-7471044 on Build GRCh38
rs749717393 Chr.12: 7471030-7471032 on Build GRCh38
rs780312753 Chr.12: 7471068-7471068 on Build GRCh38
rs924557190 Chr.12: 7470679-7470679 on Build GRCh38
rs921089435 Chr.12: 7470851-7470851 on Build GRCh38
rs1019253143 Chr.12: 7470920-7470920 on Build GRCh38
rs761125366 Chr.12: 7470894-7470894 on Build GRCh38
rs914398969 Chr.12: 7470653-7470653 on Build GRCh38
rs543552000 Chr.12: 7470909-7470909 on Build GRCh38
rs1056501253 Chr.12: 7470779-7470779 on Build GRCh38
rs1009215482 Chr.12: 7470880-7470880 on Build GRCh38
rs139277569 Chr.12: 7470802-7470802 on Build GRCh38
rs917930234 Chr.12: 7471051-7471051 on Build GRCh38
rs190560190 Chr.12: 7470798-7470798 on Build GRCh38
rs772763178 Chr.12: 7470677-7470677 on Build GRCh38
rs924501701 Chr.12: 7471015-7471015 on Build GRCh38
rs896547158 Chr.12: 7470782-7470782 on Build GRCh38
rs948120114 Chr.12: 7471053-7471053 on Build GRCh38
rs1034259824 Chr.12: 7470657-7470657 on Build GRCh38
rs1012756961 Chr.12: 7470799-7470799 on Build GRCh38
rs914365810 Chr.12: 7470948-7470948 on Build GRCh38
rs912010108 Chr.12: 7470829-7470829 on Build GRCh38
rs974069051 Chr.12: 7470691-7470691 on Build GRCh38
rs950933655 Chr.12: 7471073-7471073 on Build GRCh38
rs766299368 Chr.12: 7470921-7470921 on Build GRCh38
rs1054131249 Chr.12: 7470859-7470859 on Build GRCh38
rs1003737152 Chr.12: 7470646-7470646 on Build GRCh38
rs7305868 Chr.12: 7471001-7471001 on Build GRCh38
rs781002839 Chr.12: 7470659-7470659 on Build GRCh38
rs938596592 Chr.12: 7471024-7471024 on Build GRCh38
rs1130416 Chr.12: 7470985-7470985 on Build GRCh38
rs911894536 Chr.12: 7471091-7471091 on Build GRCh38

View Full Product Details