Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10158753-10159248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OLR1
Entrez Gene ID
4973
Gene Name
oxidized low density lipoprotein receptor 1
Gene Aliases
CLEC8A, LOX1, LOXIN, SCARE1, SLOX1
UniGene
Hs.412484
Gene Chromosome Location
Chr.12: 10158300-10172191 on Build GRCh38
Transcripts
NM_001172632.1 NM_001172633.1 NM_002543.3

SNP Detail

SNP ID Chromosome Location
rs201636631 Chr.12: 10158810-10158810 on Build GRCh38
rs921813417 Chr.12: 10159047-10159047 on Build GRCh38
rs890342284 Chr.12: 10159065-10159065 on Build GRCh38
rs552716540 Chr.12: 10158945-10158945 on Build GRCh38
rs1035238383 Chr.12: 10159195-10159195 on Build GRCh38
rs1023839474 Chr.12: 10158893-10158893 on Build GRCh38
rs962280756 Chr.12: 10159131-10159131 on Build GRCh38
rs1003634337 Chr.12: 10159171-10159171 on Build GRCh38
rs928212585 Chr.12: 10158784-10158784 on Build GRCh38
rs1031000466 Chr.12: 10158973-10158973 on Build GRCh38
rs982989387 Chr.12: 10159215-10159215 on Build GRCh38
rs999193802 Chr.12: 10159072-10159072 on Build GRCh38
rs1010684674 Chr.12: 10159102-10159102 on Build GRCh38
rs533021686 Chr.12: 10159205-10159205 on Build GRCh38
rs181429704 Chr.12: 10159013-10159013 on Build GRCh38
rs940950941 Chr.12: 10158845-10158845 on Build GRCh38
rs918248595 Chr.12: 10158869-10158869 on Build GRCh38
rs147287871 Chr.12: 10159067-10159067 on Build GRCh38
rs75688283 Chr.12: 10158918-10158918 on Build GRCh38
rs958068096 Chr.12: 10158979-10158979 on Build GRCh38
rs972764606 Chr.12: 10159150-10159150 on Build GRCh38
rs901057240 Chr.12: 10158957-10158957 on Build GRCh38
rs35678209 Chr.12: 10158796-10158797 on Build GRCh38
rs143980830 Chr.12: 10159020-10159020 on Build GRCh38
rs568346386 Chr.12: 10158846-10158846 on Build GRCh38
rs1023781996 Chr.12: 10159115-10159115 on Build GRCh38
rs766688088 Chr.12: 10158887-10158887 on Build GRCh38
rs1037580454 Chr.12: 10158800-10158801 on Build GRCh38
rs969597132 Chr.12: 10159128-10159128 on Build GRCh38
rs36051049 Chr.12: 10158944-10158944 on Build GRCh38
rs900328681 Chr.12: 10158830-10158830 on Build GRCh38
rs557031762 Chr.12: 10158907-10158907 on Build GRCh38
rs377485340 Chr.12: 10159086-10159086 on Build GRCh38
rs544245192 Chr.12: 10159062-10159062 on Build GRCh38
rs373801260 Chr.12: 10158796-10158796 on Build GRCh38
rs185807023 Chr.12: 10159113-10159113 on Build GRCh38
rs893454575 Chr.12: 10159095-10159095 on Build GRCh38
rs34938789 Chr.12: 10159012-10159012 on Build GRCh38
rs957734255 Chr.12: 10158892-10158892 on Build GRCh38
rs1050286 Chr.12: 10158964-10158964 on Build GRCh38
rs931047379 Chr.12: 10158872-10158872 on Build GRCh38
rs35738181 Chr.12: 10158855-10158855 on Build GRCh38

View Full Product Details