Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10191655-10192193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM52B
Entrez Gene ID
120939
Gene Name
transmembrane protein 52B
Gene Aliases
C12orf59
UniGene
Hs.226422
Gene Chromosome Location
Chr.12: 10170599-10191804 on Build GRCh38
Transcripts
NM_001079815.1 NM_153022.3 XM_006719029.2

SNP Detail

SNP ID Chromosome Location
rs972838448 Chr.12: 10192062-10192062 on Build GRCh38
rs1042132172 Chr.12: 10192126-10192126 on Build GRCh38
rs867033840 Chr.12: 10191868-10191868 on Build GRCh38
rs1038945554 Chr.12: 10192100-10192100 on Build GRCh38
rs959851640 Chr.12: 10191968-10191968 on Build GRCh38
rs7316627 Chr.12: 10192091-10192091 on Build GRCh38
rs940787164 Chr.12: 10192135-10192135 on Build GRCh38
rs995070446 Chr.12: 10191881-10191881 on Build GRCh38
rs564371865 Chr.12: 10191810-10191810 on Build GRCh38
rs746802288 Chr.12: 10191965-10191965 on Build GRCh38
rs532988066 Chr.12: 10191820-10191820 on Build GRCh38
rs779756620 Chr.12: 10191938-10191938 on Build GRCh38
rs957900301 Chr.12: 10192058-10192058 on Build GRCh38
rs943050646 Chr.12: 10191861-10191861 on Build GRCh38
rs187600720 Chr.12: 10191932-10191932 on Build GRCh38
rs142167538 Chr.12: 10192083-10192083 on Build GRCh38
rs73262640 Chr.12: 10191823-10191823 on Build GRCh38
rs148679675 Chr.12: 10191939-10191939 on Build GRCh38
rs566592492 Chr.12: 10191835-10191835 on Build GRCh38
rs899387456 Chr.12: 10191875-10191875 on Build GRCh38
rs1031269685 Chr.12: 10192051-10192051 on Build GRCh38
rs997258022 Chr.12: 10191894-10191894 on Build GRCh38
rs1042102895 Chr.12: 10191900-10191900 on Build GRCh38
rs111229255 Chr.12: 10191832-10191832 on Build GRCh38
rs918013479 Chr.12: 10191789-10191789 on Build GRCh38
rs571039482 Chr.12: 10192078-10192078 on Build GRCh38
rs766328252 Chr.12: 10191741-10191741 on Build GRCh38
rs537244665 Chr.12: 10191989-10191989 on Build GRCh38
rs977922713 Chr.12: 10191997-10191997 on Build GRCh38
rs949467997 Chr.12: 10191795-10191795 on Build GRCh38
rs1037011317 Chr.12: 10191842-10191842 on Build GRCh38
rs936597362 Chr.12: 10191766-10191766 on Build GRCh38
rs192413744 Chr.12: 10192110-10192110 on Build GRCh38
rs978393197 Chr.12: 10191702-10191702 on Build GRCh38
rs902246466 Chr.12: 10192161-10192161 on Build GRCh38
rs374129830 Chr.12: 10191877-10191877 on Build GRCh38
rs779298141 Chr.12: 10191819-10191819 on Build GRCh38
rs984582134 Chr.12: 10192080-10192080 on Build GRCh38
rs550902512 Chr.12: 10192077-10192077 on Build GRCh38
rs902718160 Chr.12: 10191922-10191922 on Build GRCh38
rs982174710 Chr.12: 10192107-10192107 on Build GRCh38
rs1026572174 Chr.12: 10191892-10191892 on Build GRCh38
rs940894837 Chr.12: 10191840-10191840 on Build GRCh38
rs909095771 Chr.12: 10191855-10191855 on Build GRCh38
rs1006590007 Chr.12: 10191964-10191964 on Build GRCh38
rs959412650 Chr.12: 10191777-10191777 on Build GRCh38
rs918663912 Chr.12: 10192065-10192065 on Build GRCh38
rs7316488 Chr.12: 10191977-10191977 on Build GRCh38
rs1039074386 Chr.12: 10191866-10191866 on Build GRCh38
rs950546258 Chr.12: 10192066-10192066 on Build GRCh38
rs530832066 Chr.12: 10191696-10191696 on Build GRCh38
rs989457643 Chr.12: 10191769-10191769 on Build GRCh38
rs769483872 Chr.12: 10192131-10192131 on Build GRCh38
rs889482843 Chr.12: 10191926-10191926 on Build GRCh38
rs923910022 Chr.12: 10191737-10191737 on Build GRCh38
rs757499413 Chr.12: 10191907-10191907 on Build GRCh38
rs4545645 Chr.12: 10191788-10191788 on Build GRCh38
rs561829213 Chr.12: 10191679-10191679 on Build GRCh38
rs916578668 Chr.12: 10192085-10192085 on Build GRCh38
rs747672510 Chr.12: 10192109-10192109 on Build GRCh38

View Full Product Details