Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 11021794-11022324 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRH1
Entrez Gene ID
5554
Gene Name
proline rich protein HaeIII subfamily 1
Gene Aliases
Db-s, PA, PIF-S, PRH2, Pr1/Pr2
UniGene
Hs.656965
Gene Chromosome Location
Chr.12: 10880961-11171623 on Build GRCh38
Transcripts
NM_001291314.1 NM_001291315.1 NR_133575.1 NR_037918.2 NM_001316893.1 NM_176888.2

Gene Symbol
PRH1-PRR4
Entrez Gene ID
100533464
Gene Name
PRH1-PRR4 readthrough
Gene Aliases
PRH1
UniGene
Hs.732031
Gene Chromosome Location
Chr.12: 10845849-11171625 on Build GRCh38
Transcripts
NM_001291314.1 NM_001291315.1 NR_133575.1 NR_037918.2 NM_001316893.1 NM_176888.2

Gene Symbol
PRH1-TAS2R14
Entrez Gene ID
106707243
Gene Name
PRH1-TAS2R14 readthrough
Gene Aliases
-
UniGene
Hs.731575
Gene Chromosome Location
Chr.12: 10937405-11171623 on Build GRCh38
Transcripts
NM_001291314.1 NM_001291315.1 NR_133575.1 NR_037918.2 NM_001316893.1 NM_176888.2

Gene Symbol
TAS2R19
Entrez Gene ID
259294
Gene Name
taste 2 receptor member 19
Gene Aliases
MSTP058, T2R19, T2R23, T2R48, TAS2R23, TAS2R48
UniGene
Hs.687025
Gene Chromosome Location
Chr.12: 11021619-11022620 on Build GRCh38
Transcripts
NM_001291314.1 NM_001291315.1 NR_133575.1 NR_037918.2 NM_001316893.1 NM_176888.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6189625 c.254C>T p.S85F Chr.12: 11022318-11022318 on Build GRCh38
6135313 c.278A>G p.H93R Chr.12: 11022294-11022294 on Build GRCh38
4146561 c.679G>C p.A227P Chr.12: 11021893-11021893 on Build GRCh38
6519973 c.652G>T p.D218Y Chr.12: 11021920-11021920 on Build GRCh38
4567410 c.702_703CC>TT p.L235F Chr.12: 11021869-11021870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200760616 Chr.12: 11021996-11021996 on Build GRCh38
rs767677694 Chr.12: 11022030-11022030 on Build GRCh38
rs796729478 Chr.12: 11021949-11021949 on Build GRCh38
rs199722277 Chr.12: 11022166-11022166 on Build GRCh38
rs75356565 Chr.12: 11022116-11022116 on Build GRCh38
rs890254573 Chr.12: 11021843-11021843 on Build GRCh38
rs550008134 Chr.12: 11022300-11022300 on Build GRCh38
rs375238408 Chr.12: 11021902-11021902 on Build GRCh38
rs778787132 Chr.12: 11022288-11022288 on Build GRCh38
rs199592952 Chr.12: 11021867-11021867 on Build GRCh38
rs756376706 Chr.12: 11021829-11021830 on Build GRCh38
rs760881964 Chr.12: 11022131-11022131 on Build GRCh38
rs769607556 Chr.12: 11022000-11022000 on Build GRCh38
rs750823944 Chr.12: 11021956-11021956 on Build GRCh38
rs763119571 Chr.12: 11021868-11021868 on Build GRCh38
rs376656728 Chr.12: 11022124-11022124 on Build GRCh38
rs796467634 Chr.12: 11021967-11021967 on Build GRCh38
rs202167741 Chr.12: 11022295-11022295 on Build GRCh38
rs373198792 Chr.12: 11021995-11021995 on Build GRCh38
rs754855824 Chr.12: 11022281-11022281 on Build GRCh38
rs760972910 Chr.12: 11022044-11022044 on Build GRCh38
rs142089691 Chr.12: 11022165-11022165 on Build GRCh38
rs983623900 Chr.12: 11021981-11021981 on Build GRCh38
rs112657873 Chr.12: 11022268-11022268 on Build GRCh38
rs747042594 Chr.12: 11022206-11022206 on Build GRCh38
rs911626925 Chr.12: 11022147-11022147 on Build GRCh38
rs200095330 Chr.12: 11021911-11021911 on Build GRCh38
rs775321672 Chr.12: 11022011-11022011 on Build GRCh38
rs772144970 Chr.12: 11022127-11022127 on Build GRCh38
rs375526057 Chr.12: 11022026-11022026 on Build GRCh38
rs780754068 Chr.12: 11022094-11022094 on Build GRCh38
rs112900131 Chr.12: 11021817-11021817 on Build GRCh38
rs777564748 Chr.12: 11022299-11022299 on Build GRCh38
rs777991764 Chr.12: 11021944-11021945 on Build GRCh38
rs540613976 Chr.12: 11021935-11021935 on Build GRCh38
rs374667272 Chr.12: 11021866-11021866 on Build GRCh38
rs199639701 Chr.12: 11021977-11021977 on Build GRCh38
rs796827624 Chr.12: 11022133-11022133 on Build GRCh38
rs759361007 Chr.12: 11021927-11021927 on Build GRCh38
rs753181094 Chr.12: 11022173-11022173 on Build GRCh38
rs753494301 Chr.12: 11022278-11022278 on Build GRCh38
rs535353524 Chr.12: 11022043-11022043 on Build GRCh38
rs770713225 Chr.12: 11021919-11021919 on Build GRCh38
rs138697707 Chr.12: 11021925-11021925 on Build GRCh38
rs115193179 Chr.12: 11022246-11022246 on Build GRCh38
rs115299813 Chr.12: 11021899-11021899 on Build GRCh38
rs12578654 Chr.12: 11022255-11022255 on Build GRCh38
rs567585743 Chr.12: 11022261-11022261 on Build GRCh38
rs201112051 Chr.12: 11021916-11021916 on Build GRCh38
rs774523719 Chr.12: 11021992-11021993 on Build GRCh38
rs73047160 Chr.12: 11022187-11022187 on Build GRCh38
rs777422257 Chr.12: 11021913-11021913 on Build GRCh38
rs115229396 Chr.12: 11021853-11021853 on Build GRCh38
rs751784326 Chr.12: 11021879-11021879 on Build GRCh38
rs773464415 Chr.12: 11022031-11022031 on Build GRCh38
rs73047158 Chr.12: 11022182-11022182 on Build GRCh38
rs529522791 Chr.12: 11021943-11021943 on Build GRCh38
rs138364937 Chr.12: 11021838-11021838 on Build GRCh38
rs766846237 Chr.12: 11021959-11021959 on Build GRCh38
rs985624326 Chr.12: 11022077-11022077 on Build GRCh38
rs144643889 Chr.12: 11021958-11021958 on Build GRCh38
rs796520365 Chr.12: 11022019-11022019 on Build GRCh38
rs200081075 Chr.12: 11021877-11021877 on Build GRCh38
rs796367001 Chr.12: 11022037-11022037 on Build GRCh38
rs1868769 Chr.12: 11022154-11022154 on Build GRCh38
rs766664896 Chr.12: 11022132-11022132 on Build GRCh38
rs771205863 Chr.12: 11022214-11022214 on Build GRCh38
rs12424373 Chr.12: 11022196-11022196 on Build GRCh38
rs776659604 Chr.12: 11021849-11021849 on Build GRCh38
rs139249106 Chr.12: 11022250-11022250 on Build GRCh38
rs201694653 Chr.12: 11021991-11021991 on Build GRCh38
rs755854239 Chr.12: 11022185-11022185 on Build GRCh38
rs774904403 Chr.12: 11022074-11022074 on Build GRCh38
rs746674128 Chr.12: 11021976-11021976 on Build GRCh38
rs79906789 Chr.12: 11022099-11022099 on Build GRCh38
rs779018202 Chr.12: 11022195-11022195 on Build GRCh38
rs202189737 Chr.12: 11021944-11021944 on Build GRCh38
rs776007129 Chr.12: 11022237-11022237 on Build GRCh38
rs370728247 Chr.12: 11021934-11021934 on Build GRCh38
rs749173411 Chr.12: 11022016-11022016 on Build GRCh38
rs759942495 Chr.12: 11022233-11022233 on Build GRCh38
rs796724738 Chr.12: 11022119-11022119 on Build GRCh38
rs143675824 Chr.12: 11022097-11022097 on Build GRCh38
rs372163504 Chr.12: 11021983-11021983 on Build GRCh38
rs760514884 Chr.12: 11021826-11021826 on Build GRCh38
rs745475288 Chr.12: 11022096-11022096 on Build GRCh38
rs772702281 Chr.12: 11022185-11022187 on Build GRCh38
rs755415850 Chr.12: 11022070-11022070 on Build GRCh38
rs75155989 Chr.12: 11022073-11022073 on Build GRCh38
rs139744503 Chr.12: 11022061-11022061 on Build GRCh38
rs776324145 Chr.12: 11022191-11022192 on Build GRCh38
rs563064379 Chr.12: 11021960-11021960 on Build GRCh38
rs767767305 Chr.12: 11021952-11021952 on Build GRCh38
rs749429214 Chr.12: 11021963-11021964 on Build GRCh38
rs755234275 Chr.12: 11022240-11022240 on Build GRCh38
rs201930669 Chr.12: 11022048-11022048 on Build GRCh38
rs780611250 Chr.12: 11021907-11021907 on Build GRCh38
rs772046457 Chr.12: 11022201-11022201 on Build GRCh38
rs376754166 Chr.12: 11022126-11022126 on Build GRCh38
rs768782461 Chr.12: 11022122-11022122 on Build GRCh38
rs560445580 Chr.12: 11021937-11021937 on Build GRCh38
rs759798918 Chr.12: 11022088-11022088 on Build GRCh38
rs745690071 Chr.12: 11021999-11021999 on Build GRCh38
rs373764737 Chr.12: 11022090-11022090 on Build GRCh38
rs200043527 Chr.12: 11022045-11022045 on Build GRCh38
rs771333377 Chr.12: 11021965-11021966 on Build GRCh38
rs777194969 Chr.12: 11021973-11021973 on Build GRCh38
rs776801944 Chr.12: 11022226-11022226 on Build GRCh38
rs757931642 Chr.12: 11021972-11021972 on Build GRCh38
rs754802603 Chr.12: 11022193-11022193 on Build GRCh38
rs768870640 Chr.12: 11022020-11022020 on Build GRCh38
rs766986683 Chr.12: 11022180-11022180 on Build GRCh38
rs200715232 Chr.12: 11021893-11021893 on Build GRCh38
rs796988286 Chr.12: 11022219-11022219 on Build GRCh38
rs771551352 Chr.12: 11021912-11021912 on Build GRCh38
rs753730046 Chr.12: 11022190-11022190 on Build GRCh38
rs796717526 Chr.12: 11022227-11022227 on Build GRCh38
rs752585725 Chr.12: 11022289-11022289 on Build GRCh38
rs530176245 Chr.12: 11022036-11022036 on Build GRCh38
rs149328292 Chr.12: 11021946-11021946 on Build GRCh38
rs746444973 Chr.12: 11022035-11022036 on Build GRCh38
rs963594671 Chr.12: 11022036-11022037 on Build GRCh38
rs750954456 Chr.12: 11021901-11021901 on Build GRCh38

View Full Product Details