Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 11061228-11061741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRH1
Entrez Gene ID
5554
Gene Name
proline rich protein HaeIII subfamily 1
Gene Aliases
Db-s, PA, PIF-S, PRH2, Pr1/Pr2
UniGene
Hs.656965
Gene Chromosome Location
Chr.12: 10880961-11171623 on Build GRCh38
Transcripts
NM_001291314.1 NM_001291315.1 NR_133575.1 NR_037918.2 NM_001316893.1 NM_176887.2

Gene Symbol
PRH1-PRR4
Entrez Gene ID
100533464
Gene Name
PRH1-PRR4 readthrough
Gene Aliases
PRH1
UniGene
Hs.732031
Gene Chromosome Location
Chr.12: 10845849-11171625 on Build GRCh38
Transcripts
NM_001291314.1 NM_001291315.1 NR_133575.1 NR_037918.2 NM_001316893.1 NM_176887.2

Gene Symbol
PRH1-TAS2R14
Entrez Gene ID
106707243
Gene Name
PRH1-TAS2R14 readthrough
Gene Aliases
-
UniGene
Hs.731575
Gene Chromosome Location
Chr.12: 10937405-11171623 on Build GRCh38
Transcripts
NM_001291314.1 NM_001291315.1 NR_133575.1 NR_037918.2 NM_001316893.1 NM_176887.2

Gene Symbol
TAS2R46
Entrez Gene ID
259292
Gene Name
taste 2 receptor member 46
Gene Aliases
T2R46, T2R54
UniGene
Hs.688193
Gene Chromosome Location
Chr.12: 11061365-11062294 on Build GRCh38
Transcripts
NM_001291314.1 NM_001291315.1 NR_133575.1 NR_037918.2 NM_001316893.1 NM_176887.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5903837 c.597C>T p.I199I Chr.12: 11061698-11061698 on Build GRCh38
263972 c.602C>A p.S201Y Chr.12: 11061693-11061693 on Build GRCh38
4594641 c.828A>G p.P276P Chr.12: 11061467-11061467 on Build GRCh38
4602839 c.888T>A p.H296Q Chr.12: 11061407-11061407 on Build GRCh38
6352601 c.834C>G p.I278M Chr.12: 11061461-11061461 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553647348 Chr.12: 11061410-11061410 on Build GRCh38
rs764685134 Chr.12: 11061452-11061452 on Build GRCh38
rs76147157 Chr.12: 11061538-11061538 on Build GRCh38
rs775964550 Chr.12: 11061443-11061443 on Build GRCh38
rs757249666 Chr.12: 11061488-11061488 on Build GRCh38
rs776950484 Chr.12: 11061336-11061336 on Build GRCh38
rs532799811 Chr.12: 11061285-11061285 on Build GRCh38
rs756108410 Chr.12: 11061562-11061562 on Build GRCh38
rs370408044 Chr.12: 11061493-11061493 on Build GRCh38
rs772352695 Chr.12: 11061658-11061658 on Build GRCh38
rs749575510 Chr.12: 11061510-11061510 on Build GRCh38
rs35801645 Chr.12: 11061691-11061692 on Build GRCh38
rs375025784 Chr.12: 11061597-11061597 on Build GRCh38
rs376068759 Chr.12: 11061545-11061545 on Build GRCh38
rs372278369 Chr.12: 11061426-11061426 on Build GRCh38
rs78022839 Chr.12: 11061291-11061291 on Build GRCh38
rs769125646 Chr.12: 11061601-11061601 on Build GRCh38
rs200812670 Chr.12: 11061600-11061600 on Build GRCh38
rs750226246 Chr.12: 11061715-11061715 on Build GRCh38
rs762373246 Chr.12: 11061509-11061509 on Build GRCh38
rs768445839 Chr.12: 11061682-11061682 on Build GRCh38
rs781254430 Chr.12: 11061557-11061557 on Build GRCh38
rs754611567 Chr.12: 11061395-11061395 on Build GRCh38
rs898894334 Chr.12: 11061324-11061324 on Build GRCh38
rs751399496 Chr.12: 11061554-11061554 on Build GRCh38
rs894953585 Chr.12: 11061257-11061257 on Build GRCh38
rs553357638 Chr.12: 11061474-11061474 on Build GRCh38
rs747529394 Chr.12: 11061669-11061669 on Build GRCh38
rs893683777 Chr.12: 11061620-11061621 on Build GRCh38
rs753858957 Chr.12: 11061392-11061405 on Build GRCh38
rs749695359 Chr.12: 11061499-11061499 on Build GRCh38
rs1051577281 Chr.12: 11061596-11061596 on Build GRCh38
rs755574779 Chr.12: 11061361-11061361 on Build GRCh38
rs747114723 Chr.12: 11061418-11061418 on Build GRCh38
rs768916818 Chr.12: 11061568-11061568 on Build GRCh38
rs553708509 Chr.12: 11061464-11061464 on Build GRCh38
rs370643592 Chr.12: 11061350-11061350 on Build GRCh38
rs947909567 Chr.12: 11061284-11061284 on Build GRCh38
rs780329354 Chr.12: 11061563-11061563 on Build GRCh38
rs376020270 Chr.12: 11061700-11061700 on Build GRCh38
rs765582301 Chr.12: 11061347-11061347 on Build GRCh38
rs777881796 Chr.12: 11061411-11061411 on Build GRCh38
rs774107032 Chr.12: 11061698-11061698 on Build GRCh38
rs2708381 Chr.12: 11061546-11061546 on Build GRCh38
rs750263341 Chr.12: 11061628-11061628 on Build GRCh38
rs796631017 Chr.12: 11061402-11061402 on Build GRCh38
rs2599402 Chr.12: 11061370-11061370 on Build GRCh38
rs778899748 Chr.12: 11061344-11061344 on Build GRCh38
rs185280425 Chr.12: 11061381-11061381 on Build GRCh38
rs781493121 Chr.12: 11061492-11061492 on Build GRCh38
rs368038360 Chr.12: 11061420-11061420 on Build GRCh38
rs374109594 Chr.12: 11061322-11061322 on Build GRCh38
rs374074547 Chr.12: 11061616-11061616 on Build GRCh38
rs752221868 Chr.12: 11061542-11061542 on Build GRCh38
rs778835386 Chr.12: 11061401-11061401 on Build GRCh38
rs139412224 Chr.12: 11061461-11061461 on Build GRCh38
rs775054715 Chr.12: 11061605-11061605 on Build GRCh38
rs542162895 Chr.12: 11061496-11061496 on Build GRCh38
rs76020872 Chr.12: 11061534-11061534 on Build GRCh38
rs915999455 Chr.12: 11061582-11061582 on Build GRCh38
rs777519851 Chr.12: 11061413-11061413 on Build GRCh38
rs780502162 Chr.12: 11061591-11061591 on Build GRCh38
rs557044065 Chr.12: 11061398-11061398 on Build GRCh38
rs768843374 Chr.12: 11061502-11061502 on Build GRCh38
rs137920081 Chr.12: 11061310-11061310 on Build GRCh38
rs941700841 Chr.12: 11061494-11061494 on Build GRCh38
rs377226761 Chr.12: 11061583-11061583 on Build GRCh38
rs371099707 Chr.12: 11061593-11061593 on Build GRCh38
rs999387607 Chr.12: 11061277-11061277 on Build GRCh38
rs746939972 Chr.12: 11061644-11061644 on Build GRCh38
rs746887895 Chr.12: 11061483-11061483 on Build GRCh38
rs994614992 Chr.12: 11061328-11061328 on Build GRCh38
rs769907597 Chr.12: 11061440-11061440 on Build GRCh38
rs749985491 Chr.12: 11061351-11061351 on Build GRCh38
rs561938546 Chr.12: 11061503-11061503 on Build GRCh38
rs773665473 Chr.12: 11061599-11061599 on Build GRCh38
rs200461484 Chr.12: 11061615-11061615 on Build GRCh38
rs745982573 Chr.12: 11061560-11061560 on Build GRCh38
rs771070419 Chr.12: 11061421-11061421 on Build GRCh38
rs546616630 Chr.12: 11061307-11061307 on Build GRCh38
rs754699113 Chr.12: 11061467-11061467 on Build GRCh38
rs554807484 Chr.12: 11061340-11061340 on Build GRCh38
rs375412933 Chr.12: 11061639-11061639 on Build GRCh38
rs370059980 Chr.12: 11061592-11061592 on Build GRCh38
rs371366678 Chr.12: 11061316-11061316 on Build GRCh38
rs1036012764 Chr.12: 11061610-11061610 on Build GRCh38
rs1035752661 Chr.12: 11061652-11061652 on Build GRCh38
rs199588605 Chr.12: 11061270-11061270 on Build GRCh38
rs763206168 Chr.12: 11061449-11061449 on Build GRCh38
rs764041640 Chr.12: 11061550-11061550 on Build GRCh38
rs150894148 Chr.12: 11061433-11061433 on Build GRCh38
rs748489547 Chr.12: 11061595-11061595 on Build GRCh38
rs753921956 Chr.12: 11061635-11061635 on Build GRCh38
rs369943612 Chr.12: 11061296-11061296 on Build GRCh38
rs376359978 Chr.12: 11061675-11061675 on Build GRCh38
rs377505981 Chr.12: 11061674-11061674 on Build GRCh38
rs747861564 Chr.12: 11061406-11061406 on Build GRCh38
rs374212046 Chr.12: 11061665-11061665 on Build GRCh38
rs764769965 Chr.12: 11061470-11061470 on Build GRCh38
rs2708380 Chr.12: 11061613-11061613 on Build GRCh38
rs760549574 Chr.12: 11061324-11061325 on Build GRCh38
rs772092030 Chr.12: 11061407-11061407 on Build GRCh38
rs753429374 Chr.12: 11061394-11061394 on Build GRCh38
rs200528237 Chr.12: 11061475-11061475 on Build GRCh38
rs774802527 Chr.12: 11061453-11061453 on Build GRCh38
rs776162761 Chr.12: 11061524-11061524 on Build GRCh38
rs748540248 Chr.12: 11061651-11061651 on Build GRCh38
rs749529200 Chr.12: 11061566-11061566 on Build GRCh38
rs777855752 Chr.12: 11061481-11061481 on Build GRCh38
rs757540705 Chr.12: 11061399-11061400 on Build GRCh38
rs1001601109 Chr.12: 11061640-11061640 on Build GRCh38
rs773393543 Chr.12: 11061516-11061516 on Build GRCh38
rs371448520 Chr.12: 11061629-11061629 on Build GRCh38
rs377726228 Chr.12: 11061260-11061260 on Build GRCh38
rs757304666 Chr.12: 11061556-11061556 on Build GRCh38
rs780063069 Chr.12: 11061634-11061634 on Build GRCh38
rs758867798 Chr.12: 11061484-11061485 on Build GRCh38
rs750628525 Chr.12: 11061444-11061445 on Build GRCh38
rs778984342 Chr.12: 11061421-11061421 on Build GRCh38
rs760811935 Chr.12: 11061321-11061321 on Build GRCh38
rs372334609 Chr.12: 11061670-11061670 on Build GRCh38
rs796513572 Chr.12: 11061309-11061309 on Build GRCh38
rs760189646 Chr.12: 11061457-11061457 on Build GRCh38
rs755242162 Chr.12: 11061638-11061638 on Build GRCh38
rs377680929 Chr.12: 11061348-11061348 on Build GRCh38
rs977446658 Chr.12: 11061468-11061468 on Build GRCh38
rs369672636 Chr.12: 11061701-11061701 on Build GRCh38

View Full Product Details