Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 11091733-11092234 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRH1
Entrez Gene ID
5554
Gene Name
proline rich protein HaeIII subfamily 1
Gene Aliases
Db-s, PA, PIF-S, PRH2, Pr1/Pr2
UniGene
Hs.656965
Gene Chromosome Location
Chr.12: 10880961-11171623 on Build GRCh38
Transcripts
NM_001291314.1 NM_001291315.1 NR_133575.1 NR_037918.2 NM_001316893.1 NM_176884.2

Gene Symbol
PRH1-PRR4
Entrez Gene ID
100533464
Gene Name
PRH1-PRR4 readthrough
Gene Aliases
PRH1
UniGene
Hs.732031
Gene Chromosome Location
Chr.12: 10845849-11171625 on Build GRCh38
Transcripts
NM_001291314.1 NM_001291315.1 NR_133575.1 NR_037918.2 NM_001316893.1 NM_176884.2

Gene Symbol
PRH1-TAS2R14
Entrez Gene ID
106707243
Gene Name
PRH1-TAS2R14 readthrough
Gene Aliases
-
UniGene
Hs.731575
Gene Chromosome Location
Chr.12: 10937405-11171623 on Build GRCh38
Transcripts
NM_001291314.1 NM_001291315.1 NR_133575.1 NR_037918.2 NM_001316893.1 NM_176884.2

Gene Symbol
TAS2R43
Entrez Gene ID
259289
Gene Name
taste 2 receptor member 43
Gene Aliases
T2R43, T2R52
UniGene
Hs.688195
Gene Chromosome Location
Chr.12: 11091287-11092313 on Build GRCh38
Transcripts
NM_001291314.1 NM_001291315.1 NR_133575.1 NR_037918.2 NM_001316893.1 NM_176884.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3811039 c.142C>G p.L48V Chr.12: 11092088-11092088 on Build GRCh38
3811035 c.195C>T p.N65N Chr.12: 11092035-11092035 on Build GRCh38
5802490 c.400_402delTTG p.L134delL Chr.12: 11091828-11091830 on Build GRCh38
3931521 c.438+1A>G p.? Chr.12: 11091791-11091791 on Build GRCh38
6384823 c.95C>G p.S32C Chr.12: 11092135-11092135 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751937135 Chr.12: 11091769-11091770 on Build GRCh38
rs769548101 Chr.12: 11091834-11091834 on Build GRCh38
rs535413221 Chr.12: 11091825-11091825 on Build GRCh38
rs748648257 Chr.12: 11091772-11091773 on Build GRCh38
rs777754923 Chr.12: 11091762-11091762 on Build GRCh38
rs367920076 Chr.12: 11092052-11092052 on Build GRCh38
rs376983401 Chr.12: 11091968-11091968 on Build GRCh38
rs201819808 Chr.12: 11092145-11092145 on Build GRCh38
rs747017404 Chr.12: 11092089-11092089 on Build GRCh38
rs371095380 Chr.12: 11091900-11091900 on Build GRCh38
rs770843906 Chr.12: 11092090-11092090 on Build GRCh38
rs199777806 Chr.12: 11092198-11092198 on Build GRCh38
rs750704685 Chr.12: 11091804-11091804 on Build GRCh38
rs774288758 Chr.12: 11091855-11091855 on Build GRCh38
rs111982292 Chr.12: 11091833-11091833 on Build GRCh38
rs770491026 Chr.12: 11091775-11091775 on Build GRCh38
rs745982356 Chr.12: 11092155-11092155 on Build GRCh38
rs573442217 Chr.12: 11092097-11092097 on Build GRCh38
rs758957841 Chr.12: 11091983-11091983 on Build GRCh38
rs200893955 Chr.12: 11092035-11092035 on Build GRCh38
rs755635590 Chr.12: 11091996-11091996 on Build GRCh38
rs767706122 Chr.12: 11091859-11091859 on Build GRCh38
rs753109540 Chr.12: 11092048-11092048 on Build GRCh38
rs200108786 Chr.12: 11091937-11091937 on Build GRCh38
rs779787864 Chr.12: 11091933-11091933 on Build GRCh38
rs766050964 Chr.12: 11092060-11092060 on Build GRCh38
rs773418501 Chr.12: 11092114-11092115 on Build GRCh38
rs757443713 Chr.12: 11091766-11091766 on Build GRCh38
rs11526470 Chr.12: 11091836-11091836 on Build GRCh38
rs73064966 Chr.12: 11091791-11091791 on Build GRCh38
rs144614950 Chr.12: 11091960-11091960 on Build GRCh38
rs753311828 Chr.12: 11091923-11091923 on Build GRCh38
rs749586928 Chr.12: 11092184-11092184 on Build GRCh38
rs749601715 Chr.12: 11091815-11091815 on Build GRCh38
rs547520631 Chr.12: 11091787-11091787 on Build GRCh38
rs758404927 Chr.12: 11091761-11091761 on Build GRCh38
rs373154103 Chr.12: 11091769-11091769 on Build GRCh38
rs201085601 Chr.12: 11091958-11091958 on Build GRCh38
rs775393495 Chr.12: 11091786-11091786 on Build GRCh38
rs770530290 Chr.12: 11091831-11091831 on Build GRCh38
rs555009223 Chr.12: 11091783-11091783 on Build GRCh38
rs757805454 Chr.12: 11092055-11092055 on Build GRCh38
rs763944038 Chr.12: 11091842-11091842 on Build GRCh38
rs768075873 Chr.12: 11092125-11092125 on Build GRCh38
rs763243676 Chr.12: 11092116-11092116 on Build GRCh38
rs200369046 Chr.12: 11092146-11092146 on Build GRCh38
rs746511503 Chr.12: 11091824-11091824 on Build GRCh38
rs775525781 Chr.12: 11092115-11092115 on Build GRCh38
rs986733368 Chr.12: 11092004-11092005 on Build GRCh38
rs113197337 Chr.12: 11092088-11092088 on Build GRCh38
rs866591649 Chr.12: 11091799-11091799 on Build GRCh38
rs776287488 Chr.12: 11091776-11091776 on Build GRCh38
rs191086711 Chr.12: 11092124-11092124 on Build GRCh38
rs200999522 Chr.12: 11092004-11092004 on Build GRCh38
rs200922417 Chr.12: 11092080-11092097 on Build GRCh38
rs759671855 Chr.12: 11091976-11091976 on Build GRCh38
rs779406969 Chr.12: 11091819-11091819 on Build GRCh38
rs750474264 Chr.12: 11091860-11091860 on Build GRCh38
rs201583586 Chr.12: 11092047-11092047 on Build GRCh38
rs778823151 Chr.12: 11092017-11092017 on Build GRCh38
rs779687712 Chr.12: 11092002-11092002 on Build GRCh38
rs776469455 Chr.12: 11092012-11092012 on Build GRCh38
rs766682476 Chr.12: 11092132-11092133 on Build GRCh38
rs778393759 Chr.12: 11092075-11092075 on Build GRCh38
rs374071892 Chr.12: 11091771-11091771 on Build GRCh38
rs201455884 Chr.12: 11091779-11091779 on Build GRCh38
rs531024084 Chr.12: 11092134-11092134 on Build GRCh38
rs866530303 Chr.12: 11091941-11091941 on Build GRCh38
rs751636330 Chr.12: 11091765-11091765 on Build GRCh38
rs200257456 Chr.12: 11091757-11091757 on Build GRCh38
rs774865839 Chr.12: 11092196-11092196 on Build GRCh38
rs781570675 Chr.12: 11091771-11091772 on Build GRCh38
rs766914576 Chr.12: 11091768-11091768 on Build GRCh38
rs768452582 Chr.12: 11091957-11091960 on Build GRCh38
rs369289861 Chr.12: 11091802-11091802 on Build GRCh38
rs766598967 Chr.12: 11091967-11091967 on Build GRCh38
rs73064968 Chr.12: 11091992-11091992 on Build GRCh38
rs771710605 Chr.12: 11091863-11091866 on Build GRCh38
rs367840367 Chr.12: 11092068-11092068 on Build GRCh38
rs754114537 Chr.12: 11091969-11091969 on Build GRCh38
rs201622094 Chr.12: 11091928-11091928 on Build GRCh38
rs200586631 Chr.12: 11091770-11091770 on Build GRCh38
rs202243775 Chr.12: 11092022-11092022 on Build GRCh38
rs761216323 Chr.12: 11092206-11092206 on Build GRCh38
rs761646001 Chr.12: 11091856-11091856 on Build GRCh38
rs778335774 Chr.12: 11091779-11091780 on Build GRCh38
rs199546416 Chr.12: 11092054-11092054 on Build GRCh38
rs376628544 Chr.12: 11092165-11092165 on Build GRCh38
rs758044045 Chr.12: 11092133-11092133 on Build GRCh38
rs11535673 Chr.12: 11092003-11092003 on Build GRCh38
rs186718859 Chr.12: 11091839-11091839 on Build GRCh38
rs749052007 Chr.12: 11091935-11091935 on Build GRCh38
rs780440350 Chr.12: 11091809-11091809 on Build GRCh38
rs745322342 Chr.12: 11091930-11091930 on Build GRCh38
rs763305130 Chr.12: 11092130-11092131 on Build GRCh38
rs746749985 Chr.12: 11092139-11092139 on Build GRCh38
rs68157013 Chr.12: 11092126-11092126 on Build GRCh38
rs760678007 Chr.12: 11091966-11091966 on Build GRCh38
rs775304193 Chr.12: 11091952-11091953 on Build GRCh38
rs368569282 Chr.12: 11092194-11092194 on Build GRCh38
rs756327766 Chr.12: 11091808-11091808 on Build GRCh38
rs199553429 Chr.12: 11091955-11091955 on Build GRCh38
rs773644057 Chr.12: 11092199-11092199 on Build GRCh38
rs775607159 Chr.12: 11092183-11092183 on Build GRCh38
rs758242131 Chr.12: 11092084-11092084 on Build GRCh38
rs770086595 Chr.12: 11092110-11092110 on Build GRCh38
rs764043343 Chr.12: 11091788-11091788 on Build GRCh38
rs775998182 Chr.12: 11091832-11091832 on Build GRCh38
rs201393002 Chr.12: 11091886-11091886 on Build GRCh38
rs756415369 Chr.12: 11091768-11091768 on Build GRCh38
rs138563991 Chr.12: 11092032-11092032 on Build GRCh38
rs758799232 Chr.12: 11091927-11091927 on Build GRCh38
rs774015696 Chr.12: 11091938-11091938 on Build GRCh38
rs757296915 Chr.12: 11091797-11091797 on Build GRCh38
rs755467594 Chr.12: 11091816-11091816 on Build GRCh38
rs773307005 Chr.12: 11091962-11091962 on Build GRCh38
rs374815304 Chr.12: 11092067-11092067 on Build GRCh38
rs759861245 Chr.12: 11092043-11092043 on Build GRCh38
rs761745827 Chr.12: 11092067-11092068 on Build GRCh38
rs745869855 Chr.12: 11092104-11092104 on Build GRCh38
rs755553616 Chr.12: 11091771-11091771 on Build GRCh38
rs763187564 Chr.12: 11092050-11092050 on Build GRCh38
rs201281372 Chr.12: 11092122-11092122 on Build GRCh38
rs199862459 Chr.12: 11092131-11092131 on Build GRCh38
rs748002106 Chr.12: 11092076-11092076 on Build GRCh38
rs201210705 Chr.12: 11091884-11091884 on Build GRCh38
rs761853947 Chr.12: 11091946-11091946 on Build GRCh38
rs758677584 Chr.12: 11091768-11091769 on Build GRCh38
rs760586402 Chr.12: 11092034-11092034 on Build GRCh38
rs769602644 Chr.12: 11091932-11091932 on Build GRCh38
rs201460452 Chr.12: 11091871-11091871 on Build GRCh38
rs966587470 Chr.12: 11092111-11092111 on Build GRCh38
rs201645619 Chr.12: 11092132-11092132 on Build GRCh38
rs577504710 Chr.12: 11091982-11091982 on Build GRCh38

View Full Product Details