Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 13067063-13067562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM234B
Entrez Gene ID
57613
Gene Name
family with sequence similarity 234 member B
Gene Aliases
KIAA1467
UniGene
Hs.132660
Gene Chromosome Location
Chr.12: 12927680-13083449 on Build GRCh38
Transcripts
NM_020853.1 XM_017019706.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5760676 c.1008A>G p.I336M Chr.12: 13067162-13067162 on Build GRCh38
7168529 c.1090G>A p.E364K Chr.12: 13067244-13067244 on Build GRCh38
937074 c.1061C>G p.S354* Chr.12: 13067215-13067215 on Build GRCh38
7168528 c.1084G>A p.E362K Chr.12: 13067238-13067238 on Build GRCh38
3871110 c.1081G>A p.E361K Chr.12: 13067235-13067235 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191699676 Chr.12: 13067270-13067270 on Build GRCh38
rs187012151 Chr.12: 13067214-13067214 on Build GRCh38
rs768937655 Chr.12: 13067326-13067326 on Build GRCh38
rs781557871 Chr.12: 13067093-13067093 on Build GRCh38
rs759896574 Chr.12: 13067178-13067178 on Build GRCh38
rs368592708 Chr.12: 13067216-13067216 on Build GRCh38
rs763409085 Chr.12: 13067449-13067449 on Build GRCh38
rs192666585 Chr.12: 13067123-13067123 on Build GRCh38
rs983215736 Chr.12: 13067225-13067225 on Build GRCh38
rs747902754 Chr.12: 13067136-13067136 on Build GRCh38
rs544493343 Chr.12: 13067479-13067479 on Build GRCh38
rs764768667 Chr.12: 13067347-13067347 on Build GRCh38
rs771236568 Chr.12: 13067307-13067313 on Build GRCh38
rs764469409 Chr.12: 13067205-13067205 on Build GRCh38
rs775713239 Chr.12: 13067196-13067196 on Build GRCh38
rs928009825 Chr.12: 13067516-13067516 on Build GRCh38
rs969385645 Chr.12: 13067490-13067490 on Build GRCh38
rs201805527 Chr.12: 13067129-13067129 on Build GRCh38
rs771174891 Chr.12: 13067168-13067168 on Build GRCh38
rs989525956 Chr.12: 13067388-13067388 on Build GRCh38
rs147641983 Chr.12: 13067271-13067271 on Build GRCh38
rs778837714 Chr.12: 13067131-13067131 on Build GRCh38
rs567833009 Chr.12: 13067512-13067512 on Build GRCh38
rs746904223 Chr.12: 13067166-13067166 on Build GRCh38
rs200324213 Chr.12: 13067329-13067329 on Build GRCh38
rs773050321 Chr.12: 13067162-13067162 on Build GRCh38
rs771846702 Chr.12: 13067161-13067161 on Build GRCh38
rs958199551 Chr.12: 13067324-13067324 on Build GRCh38
rs184515827 Chr.12: 13067171-13067171 on Build GRCh38
rs746992165 Chr.12: 13067284-13067284 on Build GRCh38
rs932980354 Chr.12: 13067441-13067441 on Build GRCh38
rs940607950 Chr.12: 13067534-13067534 on Build GRCh38
rs781489451 Chr.12: 13067305-13067305 on Build GRCh38
rs368080351 Chr.12: 13067307-13067307 on Build GRCh38
rs770980630 Chr.12: 13067304-13067304 on Build GRCh38
rs202100036 Chr.12: 13067330-13067330 on Build GRCh38
rs573704729 Chr.12: 13067502-13067502 on Build GRCh38
rs765919769 Chr.12: 13067211-13067211 on Build GRCh38
rs151082428 Chr.12: 13067485-13067485 on Build GRCh38
rs747138323 Chr.12: 13067406-13067406 on Build GRCh38
rs148941252 Chr.12: 13067256-13067256 on Build GRCh38
rs902234718 Chr.12: 13067484-13067484 on Build GRCh38
rs770239569 Chr.12: 13067370-13067370 on Build GRCh38
rs184195986 Chr.12: 13067308-13067308 on Build GRCh38
rs921871617 Chr.12: 13067417-13067417 on Build GRCh38
rs750874285 Chr.12: 13067336-13067336 on Build GRCh38
rs888750537 Chr.12: 13067141-13067141 on Build GRCh38
rs762272987 Chr.12: 13067206-13067206 on Build GRCh38
rs768020130 Chr.12: 13067334-13067334 on Build GRCh38
rs577115710 Chr.12: 13067457-13067457 on Build GRCh38
rs200905746 Chr.12: 13067314-13067314 on Build GRCh38
rs758994987 Chr.12: 13067337-13067337 on Build GRCh38
rs772753758 Chr.12: 13067413-13067413 on Build GRCh38
rs142786932 Chr.12: 13067265-13067265 on Build GRCh38
rs763192106 Chr.12: 13067319-13067319 on Build GRCh38
rs374368584 Chr.12: 13067320-13067320 on Build GRCh38
rs761030760 Chr.12: 13067179-13067179 on Build GRCh38

View Full Product Details