Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 14614782-14615293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUCY2C
Entrez Gene ID
2984
Gene Name
guanylate cyclase 2C
Gene Aliases
DIAR6, GUC2C, MECIL, MUCIL, STAR
UniGene
Hs.524278
Gene Chromosome Location
Chr.12: 14612632-14696625 on Build GRCh38
Transcripts
NM_004963.3 XM_011520631.2 NR_120465.1

Gene Symbol
PLBD1-AS1
Entrez Gene ID
101928290
Gene Name
PLBD1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.131933
Gene Chromosome Location
Chr.12: 14567732-14619755 on Build GRCh38
Transcripts
NM_004963.3 XM_011520631.2 NR_120465.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4936230 c.3014A>G p.D1005G Chr.12: 14614900-14614900 on Build GRCh38
5361575 c.3010A>G p.K1004E Chr.12: 14614904-14614904 on Build GRCh38
6886226 c.3015C>T p.D1005D Chr.12: 14614899-14614899 on Build GRCh38
5768783 c.3045T>C p.T1015T Chr.12: 14614869-14614869 on Build GRCh38
6361893 c.3031C>T p.P1011S Chr.12: 14614883-14614883 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751895661 Chr.12: 14615133-14615133 on Build GRCh38
rs868348341 Chr.12: 14614936-14614936 on Build GRCh38
rs981631217 Chr.12: 14614923-14614923 on Build GRCh38
rs143930092 Chr.12: 14614918-14614918 on Build GRCh38
rs1008306124 Chr.12: 14615012-14615012 on Build GRCh38
rs142317986 Chr.12: 14615045-14615045 on Build GRCh38
rs749657039 Chr.12: 14614876-14614876 on Build GRCh38
rs948504686 Chr.12: 14614902-14614902 on Build GRCh38
rs779287047 Chr.12: 14614974-14614974 on Build GRCh38
rs746337343 Chr.12: 14614845-14614845 on Build GRCh38
rs770313258 Chr.12: 14614889-14614889 on Build GRCh38
rs367743549 Chr.12: 14614847-14614847 on Build GRCh38
rs765228667 Chr.12: 14614927-14614927 on Build GRCh38
rs759104608 Chr.12: 14614987-14614987 on Build GRCh38
rs974711482 Chr.12: 14615144-14615144 on Build GRCh38
rs757546787 Chr.12: 14614957-14614957 on Build GRCh38
rs757281171 Chr.12: 14614838-14614838 on Build GRCh38
rs776230327 Chr.12: 14614901-14614901 on Build GRCh38
rs113033825 Chr.12: 14614965-14614965 on Build GRCh38
rs78132598 Chr.12: 14615009-14615009 on Build GRCh38
rs981194722 Chr.12: 14615120-14615120 on Build GRCh38
rs770651016 Chr.12: 14614982-14614982 on Build GRCh38
rs781082842 Chr.12: 14614961-14614961 on Build GRCh38
rs556994031 Chr.12: 14615001-14615001 on Build GRCh38
rs928396358 Chr.12: 14615143-14615143 on Build GRCh38
rs771651235 Chr.12: 14614877-14614877 on Build GRCh38
rs777834072 Chr.12: 14615134-14615134 on Build GRCh38
rs751802716 Chr.12: 14614953-14614953 on Build GRCh38
rs1022789051 Chr.12: 14615055-14615055 on Build GRCh38
rs762828512 Chr.12: 14614929-14614929 on Build GRCh38
rs769487677 Chr.12: 14614993-14614993 on Build GRCh38
rs373571879 Chr.12: 14614834-14614834 on Build GRCh38
rs200646012 Chr.12: 14614969-14614969 on Build GRCh38
rs374138381 Chr.12: 14614861-14614861 on Build GRCh38
rs376786609 Chr.12: 14614933-14614933 on Build GRCh38
rs970333977 Chr.12: 14615074-14615074 on Build GRCh38
rs746596911 Chr.12: 14614980-14614980 on Build GRCh38
rs760439594 Chr.12: 14614880-14614880 on Build GRCh38
rs11830063 Chr.12: 14614964-14614964 on Build GRCh38
rs950838551 Chr.12: 14614878-14614878 on Build GRCh38
rs745330478 Chr.12: 14615033-14615034 on Build GRCh38
rs756301960 Chr.12: 14615046-14615046 on Build GRCh38
rs368245378 Chr.12: 14614998-14614998 on Build GRCh38
rs999511687 Chr.12: 14615040-14615040 on Build GRCh38
rs951266883 Chr.12: 14615104-14615104 on Build GRCh38
rs535349983 Chr.12: 14614879-14614879 on Build GRCh38
rs111793441 Chr.12: 14614972-14614972 on Build GRCh38
rs780356178 Chr.12: 14614870-14614870 on Build GRCh38
rs939637261 Chr.12: 14615225-14615225 on Build GRCh38
rs567926622 Chr.12: 14614810-14614810 on Build GRCh38
rs1031521326 Chr.12: 14615041-14615041 on Build GRCh38
rs749848252 Chr.12: 14614971-14614971 on Build GRCh38
rs554351598 Chr.12: 14615030-14615030 on Build GRCh38
rs933592589 Chr.12: 14615191-14615192 on Build GRCh38
rs983913987 Chr.12: 14614900-14614900 on Build GRCh38
rs764514831 Chr.12: 14615106-14615106 on Build GRCh38
rs183589636 Chr.12: 14615028-14615028 on Build GRCh38
rs921895155 Chr.12: 14615187-14615187 on Build GRCh38
rs757587183 Chr.12: 14615147-14615147 on Build GRCh38
rs781375157 Chr.12: 14614839-14614839 on Build GRCh38
rs775940799 Chr.12: 14614983-14614983 on Build GRCh38

View Full Product Details