Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 15515378-15515826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRO
Entrez Gene ID
5800
Gene Name
protein tyrosine phosphatase, receptor type O
Gene Aliases
GLEPP1, NPHS6, PTP-OC, PTP-U2, PTPROT, PTPU2, R-PTP-O
UniGene
Hs.160871
Gene Chromosome Location
Chr.12: 15322257-15598331 on Build GRCh38
Transcripts
NM_002848.3 NM_030667.2 XM_017019725.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4750329 c.1492A>T p.N498Y Chr.12: 15515525-15515525 on Build GRCh38
2003232 c.1486A>G p.I496V Chr.12: 15515519-15515519 on Build GRCh38
352434 c.1560T>G p.P520P Chr.12: 15515593-15515593 on Build GRCh38
5374747 c.1487T>A p.I496N Chr.12: 15515520-15515520 on Build GRCh38
5970626 c.1535G>A p.R512K Chr.12: 15515568-15515568 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144862328 Chr.12: 15515584-15515584 on Build GRCh38
rs929261770 Chr.12: 15515426-15515426 on Build GRCh38
rs777907233 Chr.12: 15515576-15515576 on Build GRCh38
rs769134771 Chr.12: 15515634-15515634 on Build GRCh38
rs778485282 Chr.12: 15515558-15515558 on Build GRCh38
rs187141826 Chr.12: 15515761-15515761 on Build GRCh38
rs746948260 Chr.12: 15515471-15515471 on Build GRCh38
rs899683064 Chr.12: 15515548-15515548 on Build GRCh38
rs776301808 Chr.12: 15515655-15515655 on Build GRCh38
rs762227927 Chr.12: 15515488-15515488 on Build GRCh38
rs772340649 Chr.12: 15515649-15515649 on Build GRCh38
rs141151359 Chr.12: 15515524-15515524 on Build GRCh38
rs1018484599 Chr.12: 15515535-15515535 on Build GRCh38
rs764673571 Chr.12: 15515517-15515517 on Build GRCh38
rs1054061572 Chr.12: 15515437-15515437 on Build GRCh38
rs758380243 Chr.12: 15515553-15515553 on Build GRCh38
rs759144420 Chr.12: 15515661-15515661 on Build GRCh38
rs575388792 Chr.12: 15515458-15515458 on Build GRCh38
rs576450204 Chr.12: 15515687-15515687 on Build GRCh38
rs917994331 Chr.12: 15515799-15515799 on Build GRCh38
rs997701758 Chr.12: 15515603-15515603 on Build GRCh38
rs545420600 Chr.12: 15515688-15515688 on Build GRCh38
rs774900992 Chr.12: 15515635-15515635 on Build GRCh38
rs774679970 Chr.12: 15515479-15515479 on Build GRCh38
rs377741968 Chr.12: 15515707-15515707 on Build GRCh38
rs764627972 Chr.12: 15515478-15515478 on Build GRCh38
rs769778581 Chr.12: 15515461-15515461 on Build GRCh38
rs118114059 Chr.12: 15515708-15515708 on Build GRCh38
rs201982128 Chr.12: 15515489-15515489 on Build GRCh38
rs763485057 Chr.12: 15515474-15515474 on Build GRCh38
rs200201580 Chr.12: 15515663-15515663 on Build GRCh38
rs144500453 Chr.12: 15515403-15515403 on Build GRCh38
rs892779825 Chr.12: 15515454-15515454 on Build GRCh38
rs74073225 Chr.12: 15515605-15515605 on Build GRCh38
rs968605130 Chr.12: 15515671-15515671 on Build GRCh38
rs769876192 Chr.12: 15515617-15515617 on Build GRCh38
rs749415800 Chr.12: 15515627-15515627 on Build GRCh38
rs779964662 Chr.12: 15515623-15515623 on Build GRCh38
rs546619745 Chr.12: 15515801-15515801 on Build GRCh38
rs955853223 Chr.12: 15515615-15515615 on Build GRCh38
rs745894878 Chr.12: 15515459-15515459 on Build GRCh38
rs367658795 Chr.12: 15515494-15515494 on Build GRCh38
rs753433762 Chr.12: 15515491-15515491 on Build GRCh38
rs573665503 Chr.12: 15515645-15515645 on Build GRCh38
rs372094226 Chr.12: 15515463-15515463 on Build GRCh38
rs190667799 Chr.12: 15515685-15515685 on Build GRCh38
rs960112272 Chr.12: 15515745-15515745 on Build GRCh38
rs147435149 Chr.12: 15515771-15515771 on Build GRCh38
rs951324585 Chr.12: 15515608-15515608 on Build GRCh38
rs183318824 Chr.12: 15515747-15515747 on Build GRCh38
rs1006449598 Chr.12: 15515500-15515500 on Build GRCh38
rs971186864 Chr.12: 15515672-15515672 on Build GRCh38
rs187873272 Chr.12: 15515670-15515670 on Build GRCh38
rs762103278 Chr.12: 15515643-15515643 on Build GRCh38
rs755074605 Chr.12: 15515611-15515611 on Build GRCh38

View Full Product Details