Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 26973951-26974458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TM7SF3
Entrez Gene ID
51768
Gene Name
transmembrane 7 superfamily member 3
Gene Aliases
-
UniGene
Hs.438641
Gene Chromosome Location
Chr.12: 26966519-27014427 on Build GRCh38
Transcripts
NM_016551.2 XM_005253391.3 XM_011520690.2 XM_017019463.1 XM_017019464.1 XM_017019465.1 XM_017019466.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3398640 c.1547C>T p.P516L Chr.12: 26974131-26974131 on Build GRCh38
4840060 c.1515A>C p.R505S Chr.12: 26974163-26974163 on Build GRCh38
315956 c.1576C>T p.R526C Chr.12: 26974102-26974102 on Build GRCh38
6802659 c.1476C>A p.G492G Chr.12: 26974202-26974202 on Build GRCh38
247847 c.1525C>T p.R509* Chr.12: 26974153-26974153 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs933357466 Chr.12: 26974423-26974423 on Build GRCh38
rs777588509 Chr.12: 26974063-26974063 on Build GRCh38
rs192369447 Chr.12: 26974355-26974355 on Build GRCh38
rs777498942 Chr.12: 26974009-26974009 on Build GRCh38
rs373188766 Chr.12: 26974241-26974241 on Build GRCh38
rs369276544 Chr.12: 26973981-26973981 on Build GRCh38
rs766558661 Chr.12: 26974358-26974358 on Build GRCh38
rs35443576 Chr.12: 26974035-26974035 on Build GRCh38
rs773159960 Chr.12: 26973979-26973979 on Build GRCh38
rs773625666 Chr.12: 26974137-26974137 on Build GRCh38
rs754057481 Chr.12: 26974372-26974372 on Build GRCh38
rs890968525 Chr.12: 26974290-26974290 on Build GRCh38
rs375220805 Chr.12: 26974101-26974101 on Build GRCh38
rs989437325 Chr.12: 26974356-26974356 on Build GRCh38
rs973770297 Chr.12: 26974016-26974016 on Build GRCh38
rs758707535 Chr.12: 26974025-26974025 on Build GRCh38
rs189626254 Chr.12: 26974335-26974335 on Build GRCh38
rs1004497789 Chr.12: 26974293-26974293 on Build GRCh38
rs376448339 Chr.12: 26974167-26974167 on Build GRCh38
rs981570219 Chr.12: 26974344-26974344 on Build GRCh38
rs765847526 Chr.12: 26973998-26973998 on Build GRCh38
rs769154313 Chr.12: 26974228-26974228 on Build GRCh38
rs973382762 Chr.12: 26974432-26974432 on Build GRCh38
rs777533569 Chr.12: 26974219-26974219 on Build GRCh38
rs761287905 Chr.12: 26974265-26974265 on Build GRCh38
rs902495039 Chr.12: 26974259-26974259 on Build GRCh38
rs1013645441 Chr.12: 26974204-26974204 on Build GRCh38
rs762353907 Chr.12: 26974159-26974159 on Build GRCh38
rs780172209 Chr.12: 26974057-26974057 on Build GRCh38
rs752026818 Chr.12: 26974156-26974156 on Build GRCh38
rs759168174 Chr.12: 26974010-26974010 on Build GRCh38
rs753062266 Chr.12: 26974015-26974015 on Build GRCh38
rs958276938 Chr.12: 26974312-26974312 on Build GRCh38
rs772023979 Chr.12: 26974156-26974157 on Build GRCh38
rs996679515 Chr.12: 26974271-26974271 on Build GRCh38
rs199657283 Chr.12: 26974142-26974142 on Build GRCh38
rs142751092 Chr.12: 26974329-26974329 on Build GRCh38
rs755262789 Chr.12: 26974200-26974200 on Build GRCh38
rs568130118 Chr.12: 26974340-26974340 on Build GRCh38
rs372452992 Chr.12: 26974148-26974148 on Build GRCh38
rs947350385 Chr.12: 26974379-26974379 on Build GRCh38
rs764798586 Chr.12: 26974011-26974011 on Build GRCh38
rs928763901 Chr.12: 26974346-26974346 on Build GRCh38
rs770813174 Chr.12: 26974222-26974222 on Build GRCh38
rs766368101 Chr.12: 26974275-26974275 on Build GRCh38
rs771077210 Chr.12: 26974135-26974135 on Build GRCh38
rs774434668 Chr.12: 26974154-26974154 on Build GRCh38
rs764548345 Chr.12: 26974154-26974154 on Build GRCh38
rs769435487 Chr.12: 26974136-26974136 on Build GRCh38
rs141442318 Chr.12: 26974044-26974044 on Build GRCh38
rs756081215 Chr.12: 26973985-26973985 on Build GRCh38
rs746160862 Chr.12: 26974155-26974156 on Build GRCh38
rs139476509 Chr.12: 26974097-26974097 on Build GRCh38
rs762705907 Chr.12: 26974152-26974152 on Build GRCh38
rs202189961 Chr.12: 26974036-26974036 on Build GRCh38
rs769399824 Chr.12: 26974002-26974002 on Build GRCh38
rs373434607 Chr.12: 26974041-26974041 on Build GRCh38
rs958433037 Chr.12: 26974028-26974028 on Build GRCh38
rs755076829 Chr.12: 26974425-26974425 on Build GRCh38
rs780233295 Chr.12: 26974192-26974192 on Build GRCh38
rs775132360 Chr.12: 26974239-26974239 on Build GRCh38
rs758995196 Chr.12: 26974127-26974127 on Build GRCh38
rs183473342 Chr.12: 26974392-26974392 on Build GRCh38
rs961681671 Chr.12: 26974421-26974421 on Build GRCh38
rs899033490 Chr.12: 26974130-26974130 on Build GRCh38
rs753980005 Chr.12: 26974198-26974198 on Build GRCh38
rs750873206 Chr.12: 26974264-26974264 on Build GRCh38
rs370925155 Chr.12: 26974220-26974220 on Build GRCh38
rs774791811 Chr.12: 26974255-26974255 on Build GRCh38
rs1024945847 Chr.12: 26973972-26973972 on Build GRCh38
rs764601817 Chr.12: 26974027-26974027 on Build GRCh38
rs1029943537 Chr.12: 26974282-26974282 on Build GRCh38
rs915675266 Chr.12: 26974018-26974018 on Build GRCh38
rs749097614 Chr.12: 26974112-26974113 on Build GRCh38
rs775680523 Chr.12: 26974230-26974230 on Build GRCh38
rs375122787 Chr.12: 26974246-26974246 on Build GRCh38
rs568869226 Chr.12: 26974428-26974428 on Build GRCh38
rs369292005 Chr.12: 26974102-26974102 on Build GRCh38
rs150826720 Chr.12: 26974058-26974058 on Build GRCh38
rs745864086 Chr.12: 26974226-26974226 on Build GRCh38
rs756171490 Chr.12: 26974179-26974179 on Build GRCh38
rs200243124 Chr.12: 26974261-26974261 on Build GRCh38
rs113677104 Chr.12: 26974300-26974300 on Build GRCh38
rs750485593 Chr.12: 26974043-26974043 on Build GRCh38
rs529911353 Chr.12: 26974301-26974301 on Build GRCh38
rs1039093097 Chr.12: 26974385-26974385 on Build GRCh38
rs200978035 Chr.12: 26974098-26974098 on Build GRCh38

View Full Product Details