Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 27658653-27659159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPFIBP1
Entrez Gene ID
8496
Gene Name
PPFIA binding protein 1
Gene Aliases
L2, SGT2, hSGT2, hSgt2p
UniGene
Hs.172445
Gene Chromosome Location
Chr.12: 27524112-27695564 on Build GRCh38
Transcripts
NM_001198915.1 NM_001198916.1 NM_003622.3 NM_177444.2 XM_005253505.1 XM_005253507.1 XM_005253508.1 XM_005253509.1 XM_006719159.3 XM_017020056.1 XM_017020057.1 XM_017020058.1 XM_017020059.1 XM_017020060.1 XM_017020061.1 XM_017020062.1 XM_017020063.1 XM_017020064.1 XM_017020065.1 XM_017020066.1 XM_017020067.1 XM_017020068.1 XM_017020069.1 XM_017020070.1 XM_017020071.1 XM_017020072.1 XM_017020073.1 XM_017020074.1 XM_017020075.1 XM_017020076.1 XM_017020077.1 XM_017020078.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7327386 c.812-2A>T p.? Chr.12: 27658814-27658814 on Build GRCh38
3704050 c.828G>A p.K276K Chr.12: 27658832-27658832 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs866694700 Chr.12: 27658711-27658711 on Build GRCh38
rs757139866 Chr.12: 27658863-27658863 on Build GRCh38
rs778804391 Chr.12: 27658818-27658818 on Build GRCh38
rs199832697 Chr.12: 27658897-27658897 on Build GRCh38
rs755219478 Chr.12: 27658854-27658854 on Build GRCh38
rs748899367 Chr.12: 27658880-27658880 on Build GRCh38
rs200344303 Chr.12: 27658784-27658784 on Build GRCh38
rs60888459 Chr.12: 27658988-27658988 on Build GRCh38
rs764513573 Chr.12: 27658781-27658781 on Build GRCh38
rs368899945 Chr.12: 27658765-27658765 on Build GRCh38
rs890526029 Chr.12: 27658793-27658793 on Build GRCh38
rs547077073 Chr.12: 27659065-27659065 on Build GRCh38
rs192749192 Chr.12: 27659095-27659095 on Build GRCh38
rs932141563 Chr.12: 27658741-27658741 on Build GRCh38
rs142181591 Chr.12: 27658947-27658947 on Build GRCh38
rs547460145 Chr.12: 27658727-27658728 on Build GRCh38
rs376337581 Chr.12: 27658857-27658857 on Build GRCh38
rs781249596 Chr.12: 27658869-27658869 on Build GRCh38
rs571032560 Chr.12: 27659029-27659029 on Build GRCh38
rs745348035 Chr.12: 27658873-27658873 on Build GRCh38
rs1001378455 Chr.12: 27659124-27659124 on Build GRCh38
rs768070470 Chr.12: 27658810-27658810 on Build GRCh38
rs930172205 Chr.12: 27659092-27659092 on Build GRCh38
rs771211045 Chr.12: 27658772-27658776 on Build GRCh38
rs11049083 Chr.12: 27658879-27658879 on Build GRCh38
rs375232567 Chr.12: 27658998-27658998 on Build GRCh38
rs184998939 Chr.12: 27659018-27659018 on Build GRCh38
rs10771354 Chr.12: 27658716-27658716 on Build GRCh38
rs766560161 Chr.12: 27658843-27658843 on Build GRCh38
rs976933359 Chr.12: 27659082-27659082 on Build GRCh38
rs745452107 Chr.12: 27658773-27658773 on Build GRCh38
rs926192575 Chr.12: 27658965-27658965 on Build GRCh38
rs946242389 Chr.12: 27658693-27658693 on Build GRCh38
rs956709320 Chr.12: 27658991-27658991 on Build GRCh38
rs767162184 Chr.12: 27659020-27659020 on Build GRCh38
rs1012685130 Chr.12: 27658900-27658900 on Build GRCh38
rs201545962 Chr.12: 27658898-27658898 on Build GRCh38
rs761394831 Chr.12: 27658951-27658951 on Build GRCh38
rs918772568 Chr.12: 27659090-27659090 on Build GRCh38
rs192664204 Chr.12: 27658838-27658838 on Build GRCh38
rs932200956 Chr.12: 27658973-27658973 on Build GRCh38
rs775330116 Chr.12: 27658778-27658778 on Build GRCh38
rs576230759 Chr.12: 27658733-27658733 on Build GRCh38
rs754024307 Chr.12: 27658847-27658847 on Build GRCh38
rs750461958 Chr.12: 27658836-27658836 on Build GRCh38
rs548911988 Chr.12: 27658915-27658915 on Build GRCh38
rs992511801 Chr.12: 27658946-27658946 on Build GRCh38
rs769467276 Chr.12: 27658875-27658875 on Build GRCh38
rs906461171 Chr.12: 27658949-27658950 on Build GRCh38
rs762165190 Chr.12: 27658800-27658800 on Build GRCh38
rs76435342 Chr.12: 27659113-27659113 on Build GRCh38
rs368219215 Chr.12: 27659017-27659017 on Build GRCh38
rs774797236 Chr.12: 27658798-27658798 on Build GRCh38
rs576508430 Chr.12: 27658859-27658859 on Build GRCh38
rs763376839 Chr.12: 27658780-27658780 on Build GRCh38
rs897084285 Chr.12: 27658853-27658853 on Build GRCh38
rs879010567 Chr.12: 27658963-27658963 on Build GRCh38
rs201947912 Chr.12: 27658772-27658772 on Build GRCh38
rs981970176 Chr.12: 27658684-27658684 on Build GRCh38
rs148657383 Chr.12: 27658744-27658744 on Build GRCh38
rs913505313 Chr.12: 27658685-27658685 on Build GRCh38
rs1043255727 Chr.12: 27658695-27658695 on Build GRCh38
rs967021216 Chr.12: 27658948-27658948 on Build GRCh38
rs373667603 Chr.12: 27658681-27658681 on Build GRCh38
rs768433560 Chr.12: 27658889-27658889 on Build GRCh38
rs764448100 Chr.12: 27658917-27658917 on Build GRCh38
rs772442386 Chr.12: 27658899-27658899 on Build GRCh38
rs187509646 Chr.12: 27659034-27659034 on Build GRCh38
rs897854835 Chr.12: 27659064-27659064 on Build GRCh38

View Full Product Details