Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 46186648-46187189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC38A1
Entrez Gene ID
81539
Gene Name
solute carrier family 38 member 1
Gene Aliases
ATA1, NAT2, SAT1, SNAT1
UniGene
Hs.533770
Gene Chromosome Location
Chr.12: 46183055-46269425 on Build GRCh38
Transcripts
NM_001077484.1 NM_001278387.1 NM_001278388.1 NM_001278389.1 NM_030674.3

SNP Detail

SNP ID Chromosome Location
rs942278936 Chr.12: 46186687-46186687 on Build GRCh38
rs560348892 Chr.12: 46186768-46186768 on Build GRCh38
rs889155308 Chr.12: 46187098-46187098 on Build GRCh38
rs919384866 Chr.12: 46186812-46186812 on Build GRCh38
rs1011444166 Chr.12: 46186774-46186774 on Build GRCh38
rs899415377 Chr.12: 46187160-46187160 on Build GRCh38
rs1007550341 Chr.12: 46187109-46187109 on Build GRCh38
rs1041552701 Chr.12: 46187034-46187034 on Build GRCh38
rs948083265 Chr.12: 46187132-46187132 on Build GRCh38
rs1044271745 Chr.12: 46187154-46187154 on Build GRCh38
rs996364355 Chr.12: 46187046-46187046 on Build GRCh38
rs924014789 Chr.12: 46187064-46187064 on Build GRCh38
rs905690410 Chr.12: 46186796-46186796 on Build GRCh38
rs997586449 Chr.12: 46187077-46187078 on Build GRCh38
rs2241960 Chr.12: 46186835-46186835 on Build GRCh38
rs1016340680 Chr.12: 46186844-46186844 on Build GRCh38
rs1053912094 Chr.12: 46187117-46187117 on Build GRCh38
rs767920077 Chr.12: 46186899-46186899 on Build GRCh38
rs1008248937 Chr.12: 46186770-46186770 on Build GRCh38
rs922087707 Chr.12: 46186703-46186703 on Build GRCh38
rs902996416 Chr.12: 46187063-46187063 on Build GRCh38
rs779359508 Chr.12: 46186798-46186798 on Build GRCh38
rs1051829419 Chr.12: 46186727-46186727 on Build GRCh38
rs1016514174 Chr.12: 46186773-46186773 on Build GRCh38
rs1043729200 Chr.12: 46186689-46186689 on Build GRCh38
rs573667570 Chr.12: 46187091-46187091 on Build GRCh38
rs796309759 Chr.12: 46186973-46186973 on Build GRCh38
rs543081921 Chr.12: 46186939-46186939 on Build GRCh38
rs11183386 Chr.12: 46186867-46186867 on Build GRCh38
rs914504468 Chr.12: 46186968-46186968 on Build GRCh38
rs1051778698 Chr.12: 46187083-46187085 on Build GRCh38
rs1002629299 Chr.12: 46187163-46187163 on Build GRCh38
rs922204286 Chr.12: 46186870-46186870 on Build GRCh38
rs893011864 Chr.12: 46186769-46186769 on Build GRCh38
rs553111191 Chr.12: 46186737-46186737 on Build GRCh38
rs1035417969 Chr.12: 46186799-46186799 on Build GRCh38
rs935317508 Chr.12: 46186736-46186736 on Build GRCh38
rs937022401 Chr.12: 46186892-46186892 on Build GRCh38
rs191313231 Chr.12: 46186784-46186784 on Build GRCh38
rs933506047 Chr.12: 46186726-46186726 on Build GRCh38
rs1030302927 Chr.12: 46186754-46186754 on Build GRCh38
rs963074582 Chr.12: 46186845-46186845 on Build GRCh38
rs950530356 Chr.12: 46186808-46186808 on Build GRCh38
rs997534192 Chr.12: 46186738-46186738 on Build GRCh38
rs905802269 Chr.12: 46187157-46187157 on Build GRCh38
rs935416500 Chr.12: 46187066-46187066 on Build GRCh38
rs902614033 Chr.12: 46186730-46186730 on Build GRCh38
rs369501401 Chr.12: 46187128-46187128 on Build GRCh38

View Full Product Details