Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48064987-48065497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SENP1
Entrez Gene ID
29843
Gene Name
SUMO1/sentrin specific peptidase 1
Gene Aliases
SuPr-2
UniGene
Hs.371957
Gene Chromosome Location
Chr.12: 48042893-48106308 on Build GRCh38
Transcripts
NM_001267594.1 NM_001267595.1 NR_051991.1 NR_051992.1 XM_006719361.3 XM_006719362.2 XM_011538244.2 XM_011538245.2 XM_017019229.1 XM_017019230.1 XM_017019231.1 XM_017019232.1 XM_017019233.1 XM_017019234.1 XM_017019235.1 XM_017019236.1 XM_017019237.1 XM_017019238.1 XM_017019239.1 XM_017019240.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5492949 c.1226_1227insA p.G410fs*3 Chr.12: 48065113-48065114 on Build GRCh38
1225205 c.1195C>T p.P399S Chr.12: 48065145-48065145 on Build GRCh38
693799 c.1243G>C p.D415H Chr.12: 48065097-48065097 on Build GRCh38
4893714 c.1188G>A p.K396K Chr.12: 48065152-48065152 on Build GRCh38
3461293 c.1231C>T p.H411Y Chr.12: 48065109-48065109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762166238 Chr.12: 48065028-48065028 on Build GRCh38
rs769424838 Chr.12: 48065197-48065197 on Build GRCh38
rs777583203 Chr.12: 48065174-48065174 on Build GRCh38
rs748859551 Chr.12: 48065436-48065436 on Build GRCh38
rs917214159 Chr.12: 48065320-48065320 on Build GRCh38
rs766711163 Chr.12: 48065241-48065241 on Build GRCh38
rs750622021 Chr.12: 48065038-48065038 on Build GRCh38
rs867650460 Chr.12: 48065146-48065146 on Build GRCh38
rs767321661 Chr.12: 48065030-48065030 on Build GRCh38
rs184803349 Chr.12: 48065346-48065346 on Build GRCh38
rs779609176 Chr.12: 48065206-48065206 on Build GRCh38
rs747305136 Chr.12: 48065326-48065326 on Build GRCh38
rs780934653 Chr.12: 48065192-48065192 on Build GRCh38
rs768158385 Chr.12: 48065208-48065208 on Build GRCh38
rs954348904 Chr.12: 48065273-48065273 on Build GRCh38
rs770700027 Chr.12: 48065073-48065073 on Build GRCh38
rs552005846 Chr.12: 48065324-48065324 on Build GRCh38
rs1057491940 Chr.12: 48065459-48065459 on Build GRCh38
rs764569555 Chr.12: 48065015-48065015 on Build GRCh38
rs755018829 Chr.12: 48065160-48065160 on Build GRCh38
rs947735059 Chr.12: 48065414-48065414 on Build GRCh38
rs753229707 Chr.12: 48065265-48065265 on Build GRCh38
rs745483333 Chr.12: 48065094-48065094 on Build GRCh38
rs935818557 Chr.12: 48065397-48065397 on Build GRCh38
rs963355228 Chr.12: 48065167-48065167 on Build GRCh38
rs774960025 Chr.12: 48065098-48065098 on Build GRCh38
rs756291188 Chr.12: 48065039-48065039 on Build GRCh38
rs961584843 Chr.12: 48065287-48065287 on Build GRCh38
rs751463995 Chr.12: 48065175-48065175 on Build GRCh38
rs927536858 Chr.12: 48065439-48065439 on Build GRCh38
rs768180722 Chr.12: 48065441-48065441 on Build GRCh38
rs776936075 Chr.12: 48065260-48065260 on Build GRCh38
rs750651342 Chr.12: 48065112-48065113 on Build GRCh38
rs761007018 Chr.12: 48065055-48065055 on Build GRCh38
rs1016301727 Chr.12: 48065138-48065138 on Build GRCh38
rs188821058 Chr.12: 48065155-48065155 on Build GRCh38
rs772612036 Chr.12: 48065218-48065218 on Build GRCh38
rs192874712 Chr.12: 48065328-48065328 on Build GRCh38
rs905653266 Chr.12: 48065085-48065085 on Build GRCh38
rs745571027 Chr.12: 48065194-48065194 on Build GRCh38
rs779328437 Chr.12: 48065119-48065119 on Build GRCh38
rs766651853 Chr.12: 48065080-48065080 on Build GRCh38
rs774746421 Chr.12: 48065209-48065209 on Build GRCh38
rs927182431 Chr.12: 48065347-48065347 on Build GRCh38
rs758434980 Chr.12: 48065113-48065113 on Build GRCh38
rs769334883 Chr.12: 48065095-48065095 on Build GRCh38
rs377624808 Chr.12: 48065115-48065115 on Build GRCh38
rs754031932 Chr.12: 48065111-48065111 on Build GRCh38
rs182987633 Chr.12: 48065129-48065129 on Build GRCh38
rs150073869 Chr.12: 48065233-48065233 on Build GRCh38
rs1000156233 Chr.12: 48065096-48065096 on Build GRCh38
rs562467363 Chr.12: 48065458-48065458 on Build GRCh38
rs151323587 Chr.12: 48065230-48065230 on Build GRCh38
rs140956490 Chr.12: 48065464-48065464 on Build GRCh38
rs760953184 Chr.12: 48065128-48065128 on Build GRCh38
rs144379408 Chr.12: 48065239-48065239 on Build GRCh38
rs769447536 Chr.12: 48065355-48065355 on Build GRCh38
rs779663119 Chr.12: 48065378-48065378 on Build GRCh38
rs752727836 Chr.12: 48065054-48065054 on Build GRCh38
rs750192775 Chr.12: 48065040-48065040 on Build GRCh38
rs965796661 Chr.12: 48065213-48065213 on Build GRCh38
rs780846848 Chr.12: 48065087-48065087 on Build GRCh38
rs552244426 Chr.12: 48065416-48065416 on Build GRCh38
rs777689984 Chr.12: 48065065-48065065 on Build GRCh38
rs1044871820 Chr.12: 48065339-48065339 on Build GRCh38
rs769043662 Chr.12: 48065110-48065110 on Build GRCh38
rs75637252 Chr.12: 48065282-48065282 on Build GRCh38
rs1004994243 Chr.12: 48065467-48065467 on Build GRCh38
rs370707175 Chr.12: 48065176-48065176 on Build GRCh38
rs758253671 Chr.12: 48065056-48065056 on Build GRCh38
rs765387135 Chr.12: 48065032-48065034 on Build GRCh38
rs191336797 Chr.12: 48065399-48065399 on Build GRCh38
rs189214541 Chr.12: 48065385-48065385 on Build GRCh38
rs751860069 Chr.12: 48065026-48065026 on Build GRCh38
rs763513613 Chr.12: 48065104-48065104 on Build GRCh38
rs748755383 Chr.12: 48065207-48065207 on Build GRCh38
rs910119286 Chr.12: 48065283-48065283 on Build GRCh38
rs373291017 Chr.12: 48065310-48065310 on Build GRCh38
rs748654193 Chr.12: 48065212-48065212 on Build GRCh38
rs192825742 Chr.12: 48065113-48065113 on Build GRCh38

View Full Product Details