Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48981387-48981882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WNT1
Entrez Gene ID
7471
Gene Name
Wnt family member 1
Gene Aliases
BMND16, INT1, OI15
UniGene
Hs.248164
Gene Chromosome Location
Chr.12: 48978453-48982613 on Build GRCh38
Transcripts
NM_005430.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6863017 c.910C>T p.R304C Chr.12: 48981437-48981437 on Build GRCh38
3461484 c.991G>A p.G331S Chr.12: 48981518-48981518 on Build GRCh38
7397913 c.896G>A p.C299Y Chr.12: 48981423-48981423 on Build GRCh38
5033805 c.994A>G p.R332G Chr.12: 48981521-48981521 on Build GRCh38
6455395 c.963G>A p.A321A Chr.12: 48981490-48981490 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150664507 Chr.12: 48981687-48981687 on Build GRCh38
rs1012073084 Chr.12: 48981695-48981695 on Build GRCh38
rs555496663 Chr.12: 48981814-48981814 on Build GRCh38
rs748636854 Chr.12: 48981602-48981602 on Build GRCh38
rs370460025 Chr.12: 48981730-48981730 on Build GRCh38
rs989555065 Chr.12: 48981757-48981757 on Build GRCh38
rs976018078 Chr.12: 48981480-48981480 on Build GRCh38
rs754298019 Chr.12: 48981569-48981569 on Build GRCh38
rs759464320 Chr.12: 48981467-48981467 on Build GRCh38
rs201599168 Chr.12: 48981649-48981649 on Build GRCh38
rs761901163 Chr.12: 48981521-48981521 on Build GRCh38
rs148154047 Chr.12: 48981534-48981534 on Build GRCh38
rs61942212 Chr.12: 48981520-48981520 on Build GRCh38
rs924169589 Chr.12: 48981437-48981437 on Build GRCh38
rs776468931 Chr.12: 48981440-48981440 on Build GRCh38
rs387907358 Chr.12: 48981590-48981590 on Build GRCh38
rs148513034 Chr.12: 48981859-48981859 on Build GRCh38
rs958130273 Chr.12: 48981441-48981441 on Build GRCh38
rs767267224 Chr.12: 48981717-48981717 on Build GRCh38
rs561689863 Chr.12: 48981622-48981622 on Build GRCh38
rs751568269 Chr.12: 48981502-48981502 on Build GRCh38
rs188075130 Chr.12: 48981862-48981862 on Build GRCh38
rs754821470 Chr.12: 48981689-48981692 on Build GRCh38
rs114287565 Chr.12: 48981756-48981756 on Build GRCh38
rs387907357 Chr.12: 48981476-48981477 on Build GRCh38
rs765259238 Chr.12: 48981497-48981497 on Build GRCh38
rs1011942272 Chr.12: 48981703-48981703 on Build GRCh38
rs766350929 Chr.12: 48981817-48981817 on Build GRCh38
rs137998844 Chr.12: 48981783-48981783 on Build GRCh38
rs922873456 Chr.12: 48981591-48981591 on Build GRCh38
rs61751606 Chr.12: 48981523-48981523 on Build GRCh38
rs756414060 Chr.12: 48981552-48981552 on Build GRCh38
rs1025189952 Chr.12: 48981717-48981717 on Build GRCh38
rs976735424 Chr.12: 48981810-48981810 on Build GRCh38
rs79110091 Chr.12: 48981509-48981509 on Build GRCh38
rs755420699 Chr.12: 48981586-48981586 on Build GRCh38
rs771546038 Chr.12: 48981603-48981603 on Build GRCh38
rs922725473 Chr.12: 48981829-48981829 on Build GRCh38
rs79565269 Chr.12: 48981659-48981659 on Build GRCh38
rs967788160 Chr.12: 48981749-48981749 on Build GRCh38
rs964602627 Chr.12: 48981804-48981804 on Build GRCh38
rs779430216 Chr.12: 48981600-48981600 on Build GRCh38
rs897040674 Chr.12: 48981635-48981635 on Build GRCh38
rs727505392 Chr.12: 48981552-48981552 on Build GRCh38
rs750603706 Chr.12: 48981546-48981546 on Build GRCh38
rs886076291 Chr.12: 48981648-48981648 on Build GRCh38
rs567911014 Chr.12: 48981758-48981758 on Build GRCh38
rs761673348 Chr.12: 48981663-48981663 on Build GRCh38
rs999821734 Chr.12: 48981747-48981747 on Build GRCh38
rs930178892 Chr.12: 48981609-48981609 on Build GRCh38
rs141970485 Chr.12: 48981668-48981668 on Build GRCh38
rs930254651 Chr.12: 48981859-48981859 on Build GRCh38
rs183334037 Chr.12: 48981755-48981755 on Build GRCh38
rs780565493 Chr.12: 48981554-48981554 on Build GRCh38
rs539687282 Chr.12: 48981704-48981704 on Build GRCh38
rs1047433600 Chr.12: 48981647-48981647 on Build GRCh38
rs387907356 Chr.12: 48981411-48981411 on Build GRCh38
rs531388810 Chr.12: 48981426-48981426 on Build GRCh38
rs932036970 Chr.12: 48981517-48981517 on Build GRCh38
rs941151208 Chr.12: 48981560-48981560 on Build GRCh38

View Full Product Details