Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 3475995-3476497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF16
Entrez Gene ID
27237
Gene Name
Rho guanine nucleotide exchange factor 16
Gene Aliases
GEF16, NBR
UniGene
Hs.87435
Gene Chromosome Location
Chr.1: 3454583-3481115 on Build GRCh38
Transcripts
NM_014448.3 XM_017001049.1 XM_017001050.1 XM_017001051.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4643780 c.575C>T p.T192M Chr.1: 3476028-3476028 on Build GRCh38
4847939 c.529G>C p.E177Q Chr.1: 3476006-3476006 on Build GRCh38
4847937 c.1417G>C p.E473Q Chr.1: 3476006-3476006 on Build GRCh38
6422719 c.1424T>A p.M475K Chr.1: 3476013-3476013 on Build GRCh38
5977810 c.562C>T p.Q188* Chr.1: 3476039-3476039 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140578437 Chr.1: 3476328-3476328 on Build GRCh38
rs929004325 Chr.1: 3476244-3476244 on Build GRCh38
rs111372778 Chr.1: 3476378-3476378 on Build GRCh38
rs1057048956 Chr.1: 3476019-3476019 on Build GRCh38
rs1053854537 Chr.1: 3476336-3476336 on Build GRCh38
rs963560687 Chr.1: 3476219-3476219 on Build GRCh38
rs774598957 Chr.1: 3476037-3476037 on Build GRCh38
rs767740272 Chr.1: 3476305-3476305 on Build GRCh38
rs570754038 Chr.1: 3476467-3476467 on Build GRCh38
rs998571453 Chr.1: 3476139-3476139 on Build GRCh38
rs201584820 Chr.1: 3476024-3476024 on Build GRCh38
rs776739628 Chr.1: 3476028-3476028 on Build GRCh38
rs768901451 Chr.1: 3476033-3476033 on Build GRCh38
rs928929036 Chr.1: 3476018-3476018 on Build GRCh38
rs893741003 Chr.1: 3476091-3476091 on Build GRCh38
rs757428959 Chr.1: 3476384-3476384 on Build GRCh38
rs1003162476 Chr.1: 3476154-3476154 on Build GRCh38
rs766385189 Chr.1: 3476385-3476385 on Build GRCh38
rs935388679 Chr.1: 3476329-3476329 on Build GRCh38
rs985615682 Chr.1: 3476193-3476193 on Build GRCh38
rs372181466 Chr.1: 3476218-3476218 on Build GRCh38
rs886881521 Chr.1: 3476403-3476403 on Build GRCh38
rs569899111 Chr.1: 3476362-3476362 on Build GRCh38
rs919024441 Chr.1: 3476392-3476392 on Build GRCh38
rs1053212378 Chr.1: 3476327-3476327 on Build GRCh38
rs925838103 Chr.1: 3476471-3476471 on Build GRCh38
rs369558947 Chr.1: 3476426-3476426 on Build GRCh38
rs910050509 Chr.1: 3476217-3476217 on Build GRCh38
rs150781453 Chr.1: 3476422-3476422 on Build GRCh38
rs954358153 Chr.1: 3476338-3476339 on Build GRCh38
rs1044288748 Chr.1: 3476275-3476275 on Build GRCh38
rs917198919 Chr.1: 3476256-3476256 on Build GRCh38
rs947902250 Chr.1: 3476369-3476369 on Build GRCh38
rs974868071 Chr.1: 3476460-3476460 on Build GRCh38
rs1005737634 Chr.1: 3476413-3476413 on Build GRCh38
rs12073933 Chr.1: 3476079-3476079 on Build GRCh38
rs1045002423 Chr.1: 3476371-3476371 on Build GRCh38
rs377591919 Chr.1: 3476156-3476156 on Build GRCh38
rs762005626 Chr.1: 3476062-3476062 on Build GRCh38
rs553015615 Chr.1: 3476243-3476243 on Build GRCh38
rs1057170514 Chr.1: 3476251-3476251 on Build GRCh38
rs773394281 Chr.1: 3476075-3476075 on Build GRCh38
rs767914765 Chr.1: 3476067-3476067 on Build GRCh38
rs904649080 Chr.1: 3476057-3476057 on Build GRCh38
rs956068183 Chr.1: 3476127-3476127 on Build GRCh38
rs549317612 Chr.1: 3476410-3476410 on Build GRCh38
rs576215689 Chr.1: 3476165-3476165 on Build GRCh38
rs904367968 Chr.1: 3476302-3476302 on Build GRCh38
rs1012599439 Chr.1: 3476105-3476105 on Build GRCh38
rs148787654 Chr.1: 3476029-3476029 on Build GRCh38
rs950505368 Chr.1: 3476397-3476397 on Build GRCh38
rs12124163 Chr.1: 3476473-3476473 on Build GRCh38
rs549878034 Chr.1: 3476347-3476347 on Build GRCh38
rs948644052 Chr.1: 3476269-3476269 on Build GRCh38
rs527295205 Chr.1: 3476234-3476234 on Build GRCh38
rs114390535 Chr.1: 3476167-3476167 on Build GRCh38
rs970398431 Chr.1: 3476130-3476130 on Build GRCh38
rs74050523 Chr.1: 3476303-3476303 on Build GRCh38
rs922176397 Chr.1: 3476247-3476247 on Build GRCh38
rs948337114 Chr.1: 3476429-3476429 on Build GRCh38
rs563919451 Chr.1: 3476325-3476325 on Build GRCh38
rs921951906 Chr.1: 3476017-3476017 on Build GRCh38
rs963915598 Chr.1: 3476437-3476437 on Build GRCh38
rs556368936 Chr.1: 3476128-3476128 on Build GRCh38
rs933309902 Chr.1: 3476253-3476253 on Build GRCh38
rs200535005 Chr.1: 3476313-3476314 on Build GRCh38
rs143811823 Chr.1: 3476020-3476020 on Build GRCh38
rs139137037 Chr.1: 3476461-3476461 on Build GRCh38

View Full Product Details