Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 3682778-3683292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TP73
Entrez Gene ID
7161
Gene Name
tumor protein p73
Gene Aliases
P73
UniGene
Hs.192132
Gene Chromosome Location
Chr.1: 3652565-3736201 on Build GRCh38
Transcripts
NM_001204184.1 NM_001204185.1 NM_001204186.1 NM_001204187.1 NM_001204188.1 NM_005427.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6063263 c.89A>C p.D30A Chr.1: 3683083-3683083 on Build GRCh38
2080493 c.130G>A p.G44R Chr.1: 3683124-3683124 on Build GRCh38
4187333 c.115A>G p.N39D Chr.1: 3683109-3683109 on Build GRCh38
2080494 c.148G>T p.D50Y Chr.1: 3683142-3683142 on Build GRCh38
338254 c.163G>A p.E55K Chr.1: 3683157-3683157 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs796864922 Chr.1: 3683193-3683193 on Build GRCh38
rs1041451258 Chr.1: 3682961-3682961 on Build GRCh38
rs749038392 Chr.1: 3683013-3683013 on Build GRCh38
rs374777755 Chr.1: 3682995-3682995 on Build GRCh38
rs773568340 Chr.1: 3683186-3683187 on Build GRCh38
rs57492244 Chr.1: 3683187-3683187 on Build GRCh38
rs1041593021 Chr.1: 3683177-3683177 on Build GRCh38
rs371016737 Chr.1: 3683192-3683192 on Build GRCh38
rs762022048 Chr.1: 3683190-3683190 on Build GRCh38
rs1019815648 Chr.1: 3683169-3683170 on Build GRCh38
rs753600448 Chr.1: 3683150-3683150 on Build GRCh38
rs965634081 Chr.1: 3682952-3682952 on Build GRCh38
rs975388384 Chr.1: 3683165-3683165 on Build GRCh38
rs772113750 Chr.1: 3683221-3683221 on Build GRCh38
rs200152559 Chr.1: 3682909-3682909 on Build GRCh38
rs544753424 Chr.1: 3683187-3683188 on Build GRCh38
rs201553822 Chr.1: 3683191-3683191 on Build GRCh38
rs755616910 Chr.1: 3683144-3683144 on Build GRCh38
rs34147948 Chr.1: 3683155-3683156 on Build GRCh38
rs745328021 Chr.1: 3683066-3683066 on Build GRCh38
rs751836786 Chr.1: 3683129-3683129 on Build GRCh38
rs764517004 Chr.1: 3683049-3683049 on Build GRCh38
rs559819794 Chr.1: 3682861-3682861 on Build GRCh38
rs577907372 Chr.1: 3682902-3682902 on Build GRCh38
rs751091879 Chr.1: 3682830-3682830 on Build GRCh38
rs57596460 Chr.1: 3682950-3682950 on Build GRCh38
rs933725393 Chr.1: 3682951-3682951 on Build GRCh38
rs768727497 Chr.1: 3683020-3683020 on Build GRCh38
rs146385419 Chr.1: 3683130-3683130 on Build GRCh38
rs148777835 Chr.1: 3683128-3683128 on Build GRCh38
rs146411154 Chr.1: 3683166-3683166 on Build GRCh38
rs745954458 Chr.1: 3683168-3683168 on Build GRCh38
rs918580790 Chr.1: 3682857-3682857 on Build GRCh38
rs777629878 Chr.1: 3683167-3683167 on Build GRCh38
rs766034421 Chr.1: 3683196-3683196 on Build GRCh38
rs540531760 Chr.1: 3683039-3683039 on Build GRCh38
rs776805418 Chr.1: 3683115-3683115 on Build GRCh38
rs778557179 Chr.1: 3683101-3683101 on Build GRCh38
rs747859737 Chr.1: 3683159-3683159 on Build GRCh38
rs780791352 Chr.1: 3683060-3683060 on Build GRCh38
rs775770570 Chr.1: 3683254-3683254 on Build GRCh38
rs577760486 Chr.1: 3682814-3682814 on Build GRCh38
rs370965402 Chr.1: 3683114-3683114 on Build GRCh38
rs748164046 Chr.1: 3683105-3683105 on Build GRCh38
rs1012906415 Chr.1: 3683229-3683229 on Build GRCh38
rs377512486 Chr.1: 3683155-3683155 on Build GRCh38
rs146218192 Chr.1: 3683145-3683145 on Build GRCh38
rs202188925 Chr.1: 3683001-3683001 on Build GRCh38
rs752551379 Chr.1: 3683225-3683225 on Build GRCh38
rs557506665 Chr.1: 3682876-3682876 on Build GRCh38
rs975640636 Chr.1: 3682981-3682981 on Build GRCh38
rs769931912 Chr.1: 3683180-3683180 on Build GRCh38
rs942661590 Chr.1: 3682933-3682933 on Build GRCh38
rs199819715 Chr.1: 3683193-3683193 on Build GRCh38
rs200965855 Chr.1: 3683217-3683217 on Build GRCh38
rs114167646 Chr.1: 3683029-3683029 on Build GRCh38
rs754692674 Chr.1: 3683204-3683204 on Build GRCh38
rs200224584 Chr.1: 3683032-3683032 on Build GRCh38
rs1039923648 Chr.1: 3682941-3682941 on Build GRCh38
rs770017801 Chr.1: 3683118-3683118 on Build GRCh38
rs748631019 Chr.1: 3683100-3683101 on Build GRCh38
rs199602778 Chr.1: 3683187-3683187 on Build GRCh38
rs770878554 Chr.1: 3683142-3683142 on Build GRCh38
rs958295810 Chr.1: 3682809-3682809 on Build GRCh38
rs142260199 Chr.1: 3683033-3683033 on Build GRCh38
rs759564140 Chr.1: 3683117-3683117 on Build GRCh38
rs775452853 Chr.1: 3683011-3683011 on Build GRCh38
rs931297593 Chr.1: 3683006-3683006 on Build GRCh38
rs772069446 Chr.1: 3683161-3683161 on Build GRCh38
rs774799047 Chr.1: 3683189-3683189 on Build GRCh38
rs766536962 Chr.1: 3683027-3683027 on Build GRCh38
rs755848902 Chr.1: 3683069-3683069 on Build GRCh38
rs982697297 Chr.1: 3683092-3683092 on Build GRCh38
rs186470936 Chr.1: 3683124-3683124 on Build GRCh38
rs138505844 Chr.1: 3682869-3682869 on Build GRCh38
rs749098599 Chr.1: 3683085-3683085 on Build GRCh38
rs779792003 Chr.1: 3683081-3683081 on Build GRCh38
rs1033989028 Chr.1: 3682817-3682817 on Build GRCh38
rs753365526 Chr.1: 3683202-3683202 on Build GRCh38
rs75902087 Chr.1: 3682844-3682844 on Build GRCh38
rs557894865 Chr.1: 3683007-3683007 on Build GRCh38
rs774278400 Chr.1: 3683024-3683024 on Build GRCh38
rs543693711 Chr.1: 3682906-3682906 on Build GRCh38
rs768562905 Chr.1: 3683100-3683100 on Build GRCh38
rs753083283 Chr.1: 3683037-3683037 on Build GRCh38
rs754563142 Chr.1: 3683151-3683151 on Build GRCh38
rs200781521 Chr.1: 3683160-3683160 on Build GRCh38
rs761650125 Chr.1: 3683025-3683025 on Build GRCh38
rs752179148 Chr.1: 3683055-3683055 on Build GRCh38
rs971201658 Chr.1: 3682834-3682834 on Build GRCh38

View Full Product Details