Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6206051-6206555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF207
Entrez Gene ID
388591
Gene Name
ring finger protein 207
Gene Aliases
C1orf188
UniGene
Hs.716549
Gene Chromosome Location
Chr.1: 6205475-6221299 on Build GRCh38
Transcripts
NM_207396.2 XM_011541439.2 XM_017001259.1

SNP Detail

SNP ID Chromosome Location
rs761234514 Chr.1: 6206512-6206512 on Build GRCh38
rs747508143 Chr.1: 6206253-6206253 on Build GRCh38
rs866684303 Chr.1: 6206303-6206303 on Build GRCh38
rs953826891 Chr.1: 6206511-6206511 on Build GRCh38
rs993619206 Chr.1: 6206108-6206108 on Build GRCh38
rs1031794790 Chr.1: 6206275-6206275 on Build GRCh38
rs902732591 Chr.1: 6206436-6206436 on Build GRCh38
rs1057227864 Chr.1: 6206135-6206135 on Build GRCh38
rs906247425 Chr.1: 6206120-6206120 on Build GRCh38
rs766742539 Chr.1: 6206513-6206513 on Build GRCh38
rs748573944 Chr.1: 6206489-6206489 on Build GRCh38
rs772536253 Chr.1: 6206494-6206494 on Build GRCh38
rs961239434 Chr.1: 6206497-6206497 on Build GRCh38
rs1036659754 Chr.1: 6206333-6206333 on Build GRCh38
rs1019197178 Chr.1: 6206440-6206440 on Build GRCh38
rs953993689 Chr.1: 6206301-6206301 on Build GRCh38
rs967004616 Chr.1: 6206180-6206180 on Build GRCh38
rs561978490 Chr.1: 6206163-6206163 on Build GRCh38
rs1054920157 Chr.1: 6206136-6206137 on Build GRCh38
rs919282897 Chr.1: 6206364-6206364 on Build GRCh38
rs1022952316 Chr.1: 6206167-6206167 on Build GRCh38
rs942298467 Chr.1: 6206348-6206348 on Build GRCh38
rs758768877 Chr.1: 6206190-6206190 on Build GRCh38
rs1022780213 Chr.1: 6206205-6206205 on Build GRCh38
rs974796862 Chr.1: 6206298-6206298 on Build GRCh38
rs976354559 Chr.1: 6206309-6206309 on Build GRCh38
rs986277733 Chr.1: 6206342-6206342 on Build GRCh38
rs556727314 Chr.1: 6206392-6206392 on Build GRCh38
rs774667250 Chr.1: 6206488-6206488 on Build GRCh38
rs996154831 Chr.1: 6206454-6206454 on Build GRCh38
rs535295651 Chr.1: 6206354-6206354 on Build GRCh38
rs966063979 Chr.1: 6206206-6206206 on Build GRCh38
rs1044647991 Chr.1: 6206441-6206441 on Build GRCh38
rs922076891 Chr.1: 6206301-6206301 on Build GRCh38
rs1000100757 Chr.1: 6206244-6206244 on Build GRCh38
rs942269395 Chr.1: 6206071-6206071 on Build GRCh38
rs1054868737 Chr.1: 6206359-6206359 on Build GRCh38
rs765438329 Chr.1: 6206431-6206431 on Build GRCh38
rs1038135230 Chr.1: 6206072-6206072 on Build GRCh38
rs574180986 Chr.1: 6206350-6206350 on Build GRCh38
rs955920235 Chr.1: 6206474-6206474 on Build GRCh38
rs778649762 Chr.1: 6206107-6206107 on Build GRCh38
rs895295367 Chr.1: 6206156-6206157 on Build GRCh38
rs1000556011 Chr.1: 6206459-6206459 on Build GRCh38
rs984324808 Chr.1: 6206473-6206473 on Build GRCh38
rs575183123 Chr.1: 6206421-6206421 on Build GRCh38
rs983988406 Chr.1: 6206316-6206316 on Build GRCh38
rs1016769729 Chr.1: 6206490-6206490 on Build GRCh38
rs955725536 Chr.1: 6206282-6206282 on Build GRCh38
rs969051709 Chr.1: 6206235-6206235 on Build GRCh38
rs534442392 Chr.1: 6206119-6206119 on Build GRCh38
rs773925337 Chr.1: 6206502-6206502 on Build GRCh38
rs953775089 Chr.1: 6206331-6206331 on Build GRCh38
rs780466452 Chr.1: 6206250-6206250 on Build GRCh38
rs1012757615 Chr.1: 6206191-6206191 on Build GRCh38
rs750729093 Chr.1: 6206141-6206141 on Build GRCh38
rs897805401 Chr.1: 6206100-6206100 on Build GRCh38
rs922361693 Chr.1: 6206330-6206330 on Build GRCh38
rs1012976112 Chr.1: 6206438-6206438 on Build GRCh38
rs146041008 Chr.1: 6206448-6206448 on Build GRCh38
rs752846258 Chr.1: 6206236-6206237 on Build GRCh38
rs1036709845 Chr.1: 6206115-6206115 on Build GRCh38
rs1057115811 Chr.1: 6206399-6206399 on Build GRCh38
rs776213991 Chr.1: 6206516-6206516 on Build GRCh38
rs61760805 Chr.1: 6206462-6206462 on Build GRCh38
rs758933573 Chr.1: 6206528-6206528 on Build GRCh38
rs552999849 Chr.1: 6206264-6206264 on Build GRCh38
rs997828173 Chr.1: 6206480-6206480 on Build GRCh38
rs910709948 Chr.1: 6206346-6206346 on Build GRCh38
rs929297546 Chr.1: 6206391-6206391 on Build GRCh38

View Full Product Details