Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49564657-49565165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCRS1
Entrez Gene ID
10445
Gene Name
microspherule protein 1
Gene Aliases
ICP22BP, INO80Q, MCRS2, MSP58, P78
UniGene
Hs.25313
Gene Chromosome Location
Chr.12: 49558294-49572157 on Build GRCh38
Transcripts
NM_001012300.1 NM_001278341.1 NM_006337.4 XM_005268572.3 XM_011537760.2 XM_017018689.1 XM_017018690.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5510596 c.347C>T p.A116V Chr.12: 49564849-49564849 on Build GRCh38
3746304 c.339A>G p.P113P Chr.12: 49564857-49564857 on Build GRCh38
5510594 c.374C>T p.A125V Chr.12: 49564849-49564849 on Build GRCh38
4783067 c.301C>A p.P101T Chr.12: 49564883-49564883 on Build GRCh38
1362125 c.340C>A p.P114T Chr.12: 49564883-49564883 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369689131 Chr.12: 49565065-49565065 on Build GRCh38
rs528882226 Chr.12: 49564683-49564683 on Build GRCh38
rs771819023 Chr.12: 49564748-49564748 on Build GRCh38
rs1011970216 Chr.12: 49565110-49565110 on Build GRCh38
rs768002163 Chr.12: 49564903-49564903 on Build GRCh38
rs565448881 Chr.12: 49564873-49564873 on Build GRCh38
rs578135237 Chr.12: 49565086-49565086 on Build GRCh38
rs376824824 Chr.12: 49565108-49565108 on Build GRCh38
rs778466964 Chr.12: 49564846-49564846 on Build GRCh38
rs919645437 Chr.12: 49565035-49565035 on Build GRCh38
rs961263794 Chr.12: 49565005-49565005 on Build GRCh38
rs867592617 Chr.12: 49565116-49565116 on Build GRCh38
rs1011649180 Chr.12: 49564938-49564938 on Build GRCh38
rs982367061 Chr.12: 49565070-49565070 on Build GRCh38
rs534500411 Chr.12: 49564901-49564901 on Build GRCh38
rs762317153 Chr.12: 49564791-49564791 on Build GRCh38
rs973776997 Chr.12: 49564993-49564993 on Build GRCh38
rs1028170677 Chr.12: 49564959-49564959 on Build GRCh38
rs773698066 Chr.12: 49564723-49564723 on Build GRCh38
rs774499196 Chr.12: 49564898-49564898 on Build GRCh38
rs773904964 Chr.12: 49564783-49564783 on Build GRCh38
rs377377448 Chr.12: 49564707-49564707 on Build GRCh38
rs760592258 Chr.12: 49564764-49564764 on Build GRCh38
rs952598672 Chr.12: 49564972-49564972 on Build GRCh38
rs948499706 Chr.12: 49565037-49565037 on Build GRCh38
rs996150409 Chr.12: 49564949-49564949 on Build GRCh38
rs777539470 Chr.12: 49564870-49564870 on Build GRCh38
rs767984197 Chr.12: 49564800-49564800 on Build GRCh38
rs764534770 Chr.12: 49564691-49564691 on Build GRCh38
rs984076689 Chr.12: 49564985-49564985 on Build GRCh38
rs753235617 Chr.12: 49564831-49564831 on Build GRCh38
rs892061858 Chr.12: 49565109-49565109 on Build GRCh38
rs73305008 Chr.12: 49565128-49565128 on Build GRCh38
rs551333425 Chr.12: 49564906-49564906 on Build GRCh38
rs557235912 Chr.12: 49565138-49565138 on Build GRCh38
rs866253636 Chr.12: 49564986-49564986 on Build GRCh38
rs754686958 Chr.12: 49564837-49564837 on Build GRCh38
rs745904292 Chr.12: 49564684-49564684 on Build GRCh38
rs764183714 Chr.12: 49564784-49564784 on Build GRCh38
rs766702634 Chr.12: 49564881-49564881 on Build GRCh38
rs907415453 Chr.12: 49565074-49565074 on Build GRCh38
rs371399995 Chr.12: 49564685-49564685 on Build GRCh38
rs747853716 Chr.12: 49564743-49564743 on Build GRCh38
rs775362560 Chr.12: 49564774-49564774 on Build GRCh38
rs761284538 Chr.12: 49564914-49564914 on Build GRCh38
rs761231514 Chr.12: 49564908-49564908 on Build GRCh38
rs766752922 Chr.12: 49564919-49564919 on Build GRCh38
rs149206309 Chr.12: 49564989-49564990 on Build GRCh38
rs927712732 Chr.12: 49565071-49565071 on Build GRCh38
rs769926444 Chr.12: 49564771-49564771 on Build GRCh38
rs751775111 Chr.12: 49564785-49564785 on Build GRCh38
rs965174473 Chr.12: 49564942-49564942 on Build GRCh38
rs756626355 Chr.12: 49564815-49564815 on Build GRCh38
rs867237275 Chr.12: 49564755-49564755 on Build GRCh38
rs765338179 Chr.12: 49564702-49564702 on Build GRCh38
rs750948617 Chr.12: 49564717-49564717 on Build GRCh38
rs757870907 Chr.12: 49564700-49564700 on Build GRCh38
rs763439779 Chr.12: 49564812-49564813 on Build GRCh38
rs891815411 Chr.12: 49564937-49564937 on Build GRCh38
rs751041711 Chr.12: 49564803-49564803 on Build GRCh38
rs751822785 Chr.12: 49564695-49564695 on Build GRCh38
rs964843516 Chr.12: 49564950-49564950 on Build GRCh38
rs370265546 Chr.12: 49564728-49564728 on Build GRCh38
rs746952087 Chr.12: 49564871-49564871 on Build GRCh38
rs775672369 Chr.12: 49564877-49564877 on Build GRCh38
rs768759179 Chr.12: 49564893-49564893 on Build GRCh38
rs768134075 Chr.12: 49564731-49564731 on Build GRCh38
rs955269106 Chr.12: 49564976-49564976 on Build GRCh38
rs771911241 Chr.12: 49564866-49564866 on Build GRCh38
rs749367910 Chr.12: 49564884-49564884 on Build GRCh38
rs540983167 Chr.12: 49564934-49564936 on Build GRCh38
rs779632630 Chr.12: 49564816-49564816 on Build GRCh38
rs755747854 Chr.12: 49564726-49564726 on Build GRCh38
rs1021631740 Chr.12: 49565113-49565113 on Build GRCh38
rs138752838 Chr.12: 49564758-49564758 on Build GRCh38
rs1027663341 Chr.12: 49564958-49564958 on Build GRCh38
rs373415416 Chr.12: 49565100-49565100 on Build GRCh38
rs144889219 Chr.12: 49564753-49564753 on Build GRCh38
rs747813843 Chr.12: 49564864-49564864 on Build GRCh38
rs754491594 Chr.12: 49564940-49564940 on Build GRCh38
rs754335674 Chr.12: 49564933-49564933 on Build GRCh38
rs867433308 Chr.12: 49564928-49564928 on Build GRCh38
rs1044508392 Chr.12: 49565038-49565039 on Build GRCh38
rs778788546 Chr.12: 49564740-49564740 on Build GRCh38

View Full Product Details