Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49889390-49889896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAIM2
Entrez Gene ID
23017
Gene Name
Fas apoptotic inhibitory molecule 2
Gene Aliases
LFG, LFG2, NGP35, NMP35, TMBIM2
UniGene
Hs.567424
Gene Chromosome Location
Chr.12: 49866896-49904275 on Build GRCh38
Transcripts
NM_012306.3 XM_005268730.3 XM_017019040.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3376432 c.647C>T p.T216I Chr.12: 49889485-49889485 on Build GRCh38
4754963 c.583G>A p.V195M Chr.12: 49889549-49889549 on Build GRCh38
7162642 c.630C>T p.T210T Chr.12: 49889502-49889502 on Build GRCh38
4943676 c.576C>T p.T192T Chr.12: 49889556-49889556 on Build GRCh38
1362196 c.621C>G p.L207L Chr.12: 49889511-49889511 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758363486 Chr.12: 49889453-49889453 on Build GRCh38
rs762826221 Chr.12: 49889606-49889606 on Build GRCh38
rs988944997 Chr.12: 49889789-49889789 on Build GRCh38
rs766414434 Chr.12: 49889577-49889577 on Build GRCh38
rs200576279 Chr.12: 49889578-49889578 on Build GRCh38
rs910249502 Chr.12: 49889823-49889823 on Build GRCh38
rs888870729 Chr.12: 49889416-49889416 on Build GRCh38
rs150732432 Chr.12: 49889501-49889501 on Build GRCh38
rs528385393 Chr.12: 49889855-49889855 on Build GRCh38
rs776551275 Chr.12: 49889434-49889434 on Build GRCh38
rs750572780 Chr.12: 49889559-49889559 on Build GRCh38
rs970170781 Chr.12: 49889712-49889712 on Build GRCh38
rs756424544 Chr.12: 49889467-49889467 on Build GRCh38
rs749469222 Chr.12: 49889480-49889480 on Build GRCh38
rs746490841 Chr.12: 49889503-49889503 on Build GRCh38
rs748479032 Chr.12: 49889585-49889585 on Build GRCh38
rs763025973 Chr.12: 49889550-49889550 on Build GRCh38
rs79538930 Chr.12: 49889586-49889586 on Build GRCh38
rs1032903504 Chr.12: 49889623-49889623 on Build GRCh38
rs1020701249 Chr.12: 49889799-49889799 on Build GRCh38
rs375275250 Chr.12: 49889511-49889511 on Build GRCh38
rs968772829 Chr.12: 49889819-49889819 on Build GRCh38
rs770378669 Chr.12: 49889587-49889587 on Build GRCh38
rs750437175 Chr.12: 49889458-49889458 on Build GRCh38
rs139044559 Chr.12: 49889502-49889502 on Build GRCh38
rs191393510 Chr.12: 49889777-49889777 on Build GRCh38
rs546762745 Chr.12: 49889852-49889852 on Build GRCh38
rs1027220354 Chr.12: 49889703-49889703 on Build GRCh38
rs561016522 Chr.12: 49889827-49889827 on Build GRCh38
rs748673361 Chr.12: 49889493-49889493 on Build GRCh38
rs878900068 Chr.12: 49889640-49889640 on Build GRCh38
rs779296836 Chr.12: 49889582-49889582 on Build GRCh38
rs77687604 Chr.12: 49889752-49889752 on Build GRCh38
rs775403937 Chr.12: 49889549-49889549 on Build GRCh38
rs765147386 Chr.12: 49889526-49889526 on Build GRCh38
rs139491459 Chr.12: 49889603-49889603 on Build GRCh38
rs768436409 Chr.12: 49889608-49889608 on Build GRCh38
rs995695025 Chr.12: 49889686-49889686 on Build GRCh38
rs769543071 Chr.12: 49889602-49889602 on Build GRCh38
rs774156733 Chr.12: 49889614-49889614 on Build GRCh38
rs985807404 Chr.12: 49889782-49889782 on Build GRCh38
rs941845658 Chr.12: 49889417-49889417 on Build GRCh38
rs780796918 Chr.12: 49889863-49889863 on Build GRCh38
rs643971 Chr.12: 49889682-49889682 on Build GRCh38
rs919078372 Chr.12: 49889845-49889845 on Build GRCh38
rs756161392 Chr.12: 49889564-49889564 on Build GRCh38
rs80246888 Chr.12: 49889524-49889524 on Build GRCh38
rs761770156 Chr.12: 49889619-49889619 on Build GRCh38
rs1000984461 Chr.12: 49889595-49889595 on Build GRCh38
rs187052883 Chr.12: 49889487-49889487 on Build GRCh38
rs765486396 Chr.12: 49889486-49889486 on Build GRCh38
rs759267308 Chr.12: 49889443-49889443 on Build GRCh38
rs776158913 Chr.12: 49889589-49889589 on Build GRCh38
rs1037925997 Chr.12: 49889463-49889463 on Build GRCh38
rs181154832 Chr.12: 49889412-49889412 on Build GRCh38
rs989219867 Chr.12: 49889422-49889422 on Build GRCh38
rs745568447 Chr.12: 49889591-49889591 on Build GRCh38
rs925204789 Chr.12: 49889842-49889842 on Build GRCh38
rs1037115613 Chr.12: 49889639-49889639 on Build GRCh38
rs529880460 Chr.12: 49889831-49889831 on Build GRCh38
rs57891777 Chr.12: 49889658-49889658 on Build GRCh38
rs776157297 Chr.12: 49889523-49889523 on Build GRCh38
rs764121293 Chr.12: 49889551-49889551 on Build GRCh38
rs988789939 Chr.12: 49889724-49889724 on Build GRCh38
rs765057686 Chr.12: 49889452-49889452 on Build GRCh38

View Full Product Details