Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51269950-51270472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMAGP
Entrez Gene ID
57228
Gene Name
small cell adhesion glycoprotein
Gene Aliases
hSMAGP
UniGene
Hs.652389
Gene Chromosome Location
Chr.12: 51245349-51270418 on Build GRCh38
Transcripts
NM_001031628.1 NM_001033873.1

SNP Detail

SNP ID Chromosome Location
rs905013845 Chr.12: 51270103-51270103 on Build GRCh38
rs566263070 Chr.12: 51270365-51270365 on Build GRCh38
rs1032083277 Chr.12: 51269969-51269969 on Build GRCh38
rs951636122 Chr.12: 51270003-51270003 on Build GRCh38
rs964358961 Chr.12: 51270423-51270423 on Build GRCh38
rs911505994 Chr.12: 51270243-51270243 on Build GRCh38
rs905150198 Chr.12: 51270322-51270322 on Build GRCh38
rs896492153 Chr.12: 51270093-51270093 on Build GRCh38
rs987859203 Chr.12: 51270219-51270219 on Build GRCh38
rs914533727 Chr.12: 51270251-51270251 on Build GRCh38
rs974434402 Chr.12: 51270288-51270288 on Build GRCh38
rs987073890 Chr.12: 51270048-51270048 on Build GRCh38
rs1038799781 Chr.12: 51270102-51270103 on Build GRCh38
rs998338873 Chr.12: 51270134-51270134 on Build GRCh38
rs971203738 Chr.12: 51270056-51270056 on Build GRCh38
rs1004831892 Chr.12: 51270143-51270143 on Build GRCh38
rs990464538 Chr.12: 51270247-51270247 on Build GRCh38
rs962743245 Chr.12: 51270045-51270046 on Build GRCh38
rs375397631 Chr.12: 51270231-51270231 on Build GRCh38
rs956489896 Chr.12: 51270141-51270141 on Build GRCh38
rs1022989350 Chr.12: 51270179-51270179 on Build GRCh38
rs1013879697 Chr.12: 51270094-51270094 on Build GRCh38
rs564907954 Chr.12: 51270296-51270296 on Build GRCh38
rs148977360 Chr.12: 51270235-51270235 on Build GRCh38
rs1039025750 Chr.12: 51270360-51270360 on Build GRCh38
rs530611920 Chr.12: 51270304-51270304 on Build GRCh38
rs930009579 Chr.12: 51270276-51270276 on Build GRCh38
rs974275666 Chr.12: 51270260-51270260 on Build GRCh38
rs573758664 Chr.12: 51270055-51270055 on Build GRCh38
rs1047124455 Chr.12: 51270089-51270089 on Build GRCh38
rs954420930 Chr.12: 51270236-51270236 on Build GRCh38
rs1017196451 Chr.12: 51270145-51270145 on Build GRCh38
rs1023567772 Chr.12: 51270102-51270102 on Build GRCh38
rs934204653 Chr.12: 51270043-51270043 on Build GRCh38
rs971483439 Chr.12: 51270229-51270229 on Build GRCh38
rs1018939735 Chr.12: 51270404-51270404 on Build GRCh38
rs1031750238 Chr.12: 51270428-51270428 on Build GRCh38
rs528060798 Chr.12: 51270240-51270240 on Build GRCh38
rs954512711 Chr.12: 51270370-51270370 on Build GRCh38
rs535365052 Chr.12: 51270380-51270380 on Build GRCh38
rs942895009 Chr.12: 51270051-51270051 on Build GRCh38
rs901616537 Chr.12: 51270111-51270111 on Build GRCh38
rs974327872 Chr.12: 51270440-51270440 on Build GRCh38
rs544998739 Chr.12: 51270271-51270271 on Build GRCh38
rs934544646 Chr.12: 51270082-51270082 on Build GRCh38
rs924517723 Chr.12: 51270071-51270071 on Build GRCh38
rs913477993 Chr.12: 51270042-51270042 on Build GRCh38
rs910727120 Chr.12: 51270016-51270027 on Build GRCh38
rs1047179852 Chr.12: 51270287-51270287 on Build GRCh38
rs556735731 Chr.12: 51270050-51270050 on Build GRCh38
rs1004480145 Chr.12: 51270016-51270021 on Build GRCh38
rs1012961594 Chr.12: 51270174-51270174 on Build GRCh38
rs543554531 Chr.12: 51270033-51270033 on Build GRCh38
rs920051486 Chr.12: 51270446-51270446 on Build GRCh38
rs981267068 Chr.12: 51270064-51270064 on Build GRCh38
rs1022892756 Chr.12: 51270367-51270367 on Build GRCh38
rs188070189 Chr.12: 51270336-51270336 on Build GRCh38
rs542867146 Chr.12: 51270066-51270066 on Build GRCh38
rs942843748 Chr.12: 51270256-51270256 on Build GRCh38
rs892304284 Chr.12: 51270353-51270353 on Build GRCh38
rs1032051189 Chr.12: 51270136-51270136 on Build GRCh38
rs999376264 Chr.12: 51270065-51270065 on Build GRCh38
rs2271821 Chr.12: 51270363-51270363 on Build GRCh38
rs1038449882 Chr.12: 51270054-51270054 on Build GRCh38
rs1000680074 Chr.12: 51270113-51270113 on Build GRCh38
rs919997836 Chr.12: 51270273-51270273 on Build GRCh38
rs963028096 Chr.12: 51270147-51270147 on Build GRCh38
rs73307788 Chr.12: 51270324-51270324 on Build GRCh38
rs879378845 Chr.12: 51270027-51270028 on Build GRCh38
rs956357119 Chr.12: 51269976-51269976 on Build GRCh38
rs929957075 Chr.12: 51270077-51270077 on Build GRCh38
rs529472421 Chr.12: 51270355-51270355 on Build GRCh38
rs981504915 Chr.12: 51270232-51270232 on Build GRCh38
rs577702860 Chr.12: 51270010-51270010 on Build GRCh38
rs917884626 Chr.12: 51270294-51270294 on Build GRCh38

View Full Product Details